Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 46756721-46757218 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FKRP
Entrez Gene ID
79147
Gene Name
fukutin related protein
Gene Aliases
LGMD2I, MDC1C, MDDGA5, MDDGB5, MDDGC5
UniGene
Hs.515493
Gene Chromosome Location
Chr.19: 46746015-46758575 on Build GRCh38
Transcripts
NM_001039885.2 NM_024301.4 XM_005259247.2 XM_005259248.2 XM_005259249.4 XM_011527306.1 XM_011527307.1 XM_017027297.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4684806 c.1382C>T p.A461V Chr.19: 46756832-46756832 on Build GRCh38
5484316 c.1338G>A p.L446L Chr.19: 46756788-46756788 on Build GRCh38
5498351 c.1307G>A p.R436Q Chr.19: 46756757-46756757 on Build GRCh38
5499375 c.1327G>A p.E443K Chr.19: 46756777-46756777 on Build GRCh38
3389162 c.1424C>T p.P475L Chr.19: 46756874-46756874 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749109905 Chr.19: 46756826-46756826 on Build GRCh38
rs104894681 Chr.19: 46756793-46756793 on Build GRCh38
rs755722104 Chr.19: 46756871-46756871 on Build GRCh38
rs948074617 Chr.19: 46757069-46757069 on Build GRCh38
rs940897679 Chr.19: 46757160-46757160 on Build GRCh38
rs764311774 Chr.19: 46756757-46756757 on Build GRCh38
rs752243337 Chr.19: 46756868-46756868 on Build GRCh38
rs912051695 Chr.19: 46757024-46757024 on Build GRCh38
rs886044542 Chr.19: 46756802-46756802 on Build GRCh38
rs763422045 Chr.19: 46756847-46756847 on Build GRCh38
rs747785577 Chr.19: 46756813-46756813 on Build GRCh38
rs752153053 Chr.19: 46756940-46756940 on Build GRCh38
rs759929539 Chr.19: 46756933-46756933 on Build GRCh38
rs930285524 Chr.19: 46756876-46756876 on Build GRCh38
rs762008480 Chr.19: 46756908-46756908 on Build GRCh38
rs989542748 Chr.19: 46757030-46757030 on Build GRCh38
rs930317962 Chr.19: 46757177-46757177 on Build GRCh38
rs727502844 Chr.19: 46756892-46756892 on Build GRCh38
rs745774108 Chr.19: 46756891-46756891 on Build GRCh38
rs754002921 Chr.19: 46756766-46756766 on Build GRCh38
rs763299165 Chr.19: 46756927-46756927 on Build GRCh38
rs1031041128 Chr.19: 46756966-46756966 on Build GRCh38
rs936735225 Chr.19: 46756788-46756788 on Build GRCh38
rs928180718 Chr.19: 46757113-46757113 on Build GRCh38
rs780545437 Chr.19: 46756863-46756863 on Build GRCh38
rs121908110 Chr.19: 46756837-46756837 on Build GRCh38
rs1046858778 Chr.19: 46757091-46757091 on Build GRCh38
rs758936275 Chr.19: 46756776-46756776 on Build GRCh38
rs369666163 Chr.19: 46756889-46756889 on Build GRCh38
rs927775318 Chr.19: 46757118-46757118 on Build GRCh38
rs958282186 Chr.19: 46756979-46756979 on Build GRCh38
rs115365212 Chr.19: 46756890-46756890 on Build GRCh38
rs759403448 Chr.19: 46757097-46757099 on Build GRCh38
rs754446767 Chr.19: 46756794-46756794 on Build GRCh38
rs772959356 Chr.19: 46756817-46756817 on Build GRCh38
rs780445235 Chr.19: 46756785-46756785 on Build GRCh38
rs759585825 Chr.19: 46756833-46756833 on Build GRCh38
rs953608731 Chr.19: 46757010-46757010 on Build GRCh38
rs1036061592 Chr.19: 46756829-46756829 on Build GRCh38
rs768606230 Chr.19: 46756834-46756834 on Build GRCh38
rs185647378 Chr.19: 46757004-46757004 on Build GRCh38
rs148318216 Chr.19: 46756775-46756775 on Build GRCh38
rs920322240 Chr.19: 46757173-46757173 on Build GRCh38
rs1000011265 Chr.19: 46756955-46756955 on Build GRCh38
rs987624962 Chr.19: 46757015-46757015 on Build GRCh38
rs144236975 Chr.19: 46756756-46756756 on Build GRCh38
rs104894682 Chr.19: 46756936-46756936 on Build GRCh38
rs766839420 Chr.19: 46756850-46756850 on Build GRCh38
rs916693739 Chr.19: 46757051-46757051 on Build GRCh38
rs972223917 Chr.19: 46757046-46757046 on Build GRCh38
rs886044520 Chr.19: 46756773-46756773 on Build GRCh38
rs776825755 Chr.19: 46756836-46756836 on Build GRCh38
rs373244438 Chr.19: 46756932-46756932 on Build GRCh38
rs543603798 Chr.19: 46757172-46757172 on Build GRCh38
rs746787862 Chr.19: 46756887-46756887 on Build GRCh38
rs757615255 Chr.19: 46756767-46756767 on Build GRCh38
rs750771321 Chr.19: 46756842-46756842 on Build GRCh38
rs977327820 Chr.19: 46757003-46757003 on Build GRCh38
rs780817621 Chr.19: 46756796-46756796 on Build GRCh38
rs765678360 Chr.19: 46756770-46756770 on Build GRCh38
rs774161137 Chr.19: 46756832-46756832 on Build GRCh38
rs961858508 Chr.19: 46757034-46757034 on Build GRCh38
rs765591278 Chr.19: 46756839-46756839 on Build GRCh38
rs776815140 Chr.19: 46756904-46756904 on Build GRCh38
rs756983814 Chr.19: 46756983-46756983 on Build GRCh38
rs770845177 Chr.19: 46756827-46756827 on Build GRCh38
rs772089991 Chr.19: 46756899-46756899 on Build GRCh38
rs112819526 Chr.19: 46756915-46756915 on Build GRCh38
rs773569711 Chr.19: 46756919-46756919 on Build GRCh38
rs753476178 Chr.19: 46756974-46756974 on Build GRCh38
rs28937903 Chr.19: 46756814-46756814 on Build GRCh38
rs868310782 Chr.19: 46756869-46756869 on Build GRCh38
rs766343049 Chr.19: 46756748-46756748 on Build GRCh38
rs574731209 Chr.19: 46757053-46757053 on Build GRCh38
rs541355514 Chr.19: 46756971-46756971 on Build GRCh38
rs143129484 Chr.19: 46756855-46756855 on Build GRCh38
rs375326964 Chr.19: 46756877-46756877 on Build GRCh38

View Full Product Details