Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47266522-47266943 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC9
Entrez Gene ID
26093
Gene Name
coiled-coil domain containing 9
Gene Aliases
-
UniGene
Hs.227782
Gene Chromosome Location
Chr.19: 47256474-47271958 on Build GRCh38
Transcripts
NM_015603.2 XM_017026578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5481536 c.782G>A p.R261Q Chr.19: 47266672-47266672 on Build GRCh38
7148997 c.822C>T p.I274I Chr.19: 47266712-47266712 on Build GRCh38
4132192 c.885C>T p.A295A Chr.19: 47266775-47266775 on Build GRCh38
7294578 c.775C>T p.R259W Chr.19: 47266665-47266665 on Build GRCh38
5645679 c.746C>G p.A249G Chr.19: 47266636-47266636 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756202754 Chr.19: 47266641-47266641 on Build GRCh38
rs746176928 Chr.19: 47266766-47266766 on Build GRCh38
rs771309913 Chr.19: 47266573-47266573 on Build GRCh38
rs760057485 Chr.19: 47266574-47266574 on Build GRCh38
rs751412718 Chr.19: 47266812-47266812 on Build GRCh38
rs751476635 Chr.19: 47266747-47266747 on Build GRCh38
rs753909244 Chr.19: 47266832-47266832 on Build GRCh38
rs779070299 Chr.19: 47266764-47266764 on Build GRCh38
rs1016547944 Chr.19: 47266753-47266753 on Build GRCh38
rs143618657 Chr.19: 47266784-47266784 on Build GRCh38
rs368410576 Chr.19: 47266562-47266562 on Build GRCh38
rs748238443 Chr.19: 47266563-47266563 on Build GRCh38
rs762763043 Chr.19: 47266809-47266809 on Build GRCh38
rs199690358 Chr.19: 47266700-47266700 on Build GRCh38
rs973675316 Chr.19: 47266692-47266692 on Build GRCh38
rs757457032 Chr.19: 47266763-47266763 on Build GRCh38
rs758135209 Chr.19: 47266712-47266712 on Build GRCh38
rs746276188 Chr.19: 47266662-47266662 on Build GRCh38
rs779139148 Chr.19: 47266864-47266864 on Build GRCh38
rs747498759 Chr.19: 47266781-47266781 on Build GRCh38
rs748194748 Chr.19: 47266618-47266618 on Build GRCh38
rs141001862 Chr.19: 47266685-47266685 on Build GRCh38
rs996754716 Chr.19: 47266880-47266880 on Build GRCh38
rs949899701 Chr.19: 47266887-47266887 on Build GRCh38
rs60383222 Chr.19: 47266775-47266775 on Build GRCh38
rs755164287 Chr.19: 47266839-47266840 on Build GRCh38
rs766120752 Chr.19: 47266811-47266811 on Build GRCh38
rs779137034 Chr.19: 47266834-47266834 on Build GRCh38
rs552367352 Chr.19: 47266581-47266581 on Build GRCh38
rs376713157 Chr.19: 47266921-47266921 on Build GRCh38
rs563523153 Chr.19: 47266549-47266549 on Build GRCh38
rs1055853612 Chr.19: 47266862-47266863 on Build GRCh38
rs144274363 Chr.19: 47266650-47266650 on Build GRCh38
rs951085279 Chr.19: 47266550-47266550 on Build GRCh38
rs775050861 Chr.19: 47266578-47266578 on Build GRCh38
rs754110218 Chr.19: 47266601-47266602 on Build GRCh38
rs866950905 Chr.19: 47266738-47266738 on Build GRCh38
rs936072145 Chr.19: 47266853-47266853 on Build GRCh38
rs936060847 Chr.19: 47266615-47266615 on Build GRCh38
rs962423320 Chr.19: 47266570-47266570 on Build GRCh38
rs201519530 Chr.19: 47266686-47266686 on Build GRCh38
rs766285297 Chr.19: 47266722-47266722 on Build GRCh38
rs779926142 Chr.19: 47266614-47266614 on Build GRCh38
rs891061385 Chr.19: 47266910-47266910 on Build GRCh38
rs200221689 Chr.19: 47266682-47266682 on Build GRCh38
rs532686088 Chr.19: 47266561-47266561 on Build GRCh38
rs9304666 Chr.19: 47266582-47266582 on Build GRCh38
rs777924596 Chr.19: 47266648-47266648 on Build GRCh38
rs955411828 Chr.19: 47266639-47266639 on Build GRCh38
rs772627183 Chr.19: 47266666-47266666 on Build GRCh38
rs1046203687 Chr.19: 47266548-47266548 on Build GRCh38
rs772756625 Chr.19: 47266808-47266808 on Build GRCh38
rs146588636 Chr.19: 47266679-47266679 on Build GRCh38
rs760168719 Chr.19: 47266580-47266580 on Build GRCh38
rs774676187 Chr.19: 47266660-47266660 on Build GRCh38
rs749544651 Chr.19: 47266649-47266649 on Build GRCh38
rs893167056 Chr.19: 47266858-47266858 on Build GRCh38
rs753968748 Chr.19: 47266762-47266762 on Build GRCh38
rs757332323 Chr.19: 47266833-47266833 on Build GRCh38
rs758347882 Chr.19: 47266604-47266604 on Build GRCh38
rs758519801 Chr.19: 47266845-47266845 on Build GRCh38
rs759447821 Chr.19: 47266814-47266814 on Build GRCh38
rs770054138 Chr.19: 47266566-47266566 on Build GRCh38
rs376149307 Chr.19: 47266675-47266675 on Build GRCh38
rs777801182 Chr.19: 47266761-47266761 on Build GRCh38
rs749599636 Chr.19: 47266571-47266571 on Build GRCh38
rs776254737 Chr.19: 47266785-47266785 on Build GRCh38
rs894322185 Chr.19: 47266862-47266862 on Build GRCh38
rs765151613 Chr.19: 47266598-47266598 on Build GRCh38
rs556966941 Chr.19: 47266884-47266884 on Build GRCh38
rs773430275 Chr.19: 47266569-47266569 on Build GRCh38
rs10413121 Chr.19: 47266758-47266758 on Build GRCh38
rs750220803 Chr.19: 47266602-47266602 on Build GRCh38
rs369751478 Chr.19: 47266791-47266791 on Build GRCh38
rs767500734 Chr.19: 47266828-47266828 on Build GRCh38
rs761485244 Chr.19: 47266801-47266801 on Build GRCh38
rs373431331 Chr.19: 47266807-47266807 on Build GRCh38
rs975095517 Chr.19: 47266760-47266760 on Build GRCh38
rs757067315 Chr.19: 47266587-47266587 on Build GRCh38
rs144789929 Chr.19: 47266705-47266705 on Build GRCh38

View Full Product Details