Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47270326-47270831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC9
Entrez Gene ID
26093
Gene Name
coiled-coil domain containing 9
Gene Aliases
-
UniGene
Hs.227782
Gene Chromosome Location
Chr.19: 47256474-47271958 on Build GRCh38
Transcripts
NM_015603.2 XM_017026578.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
327389 c.1043A>G p.K348R Chr.19: 47270646-47270646 on Build GRCh38
6277929 c.903-3C>T p.? Chr.19: 47270404-47270404 on Build GRCh38
712180 c.992G>C p.G331A Chr.19: 47270595-47270595 on Build GRCh38
3106185 c.981C>T p.P327P Chr.19: 47270584-47270584 on Build GRCh38
5641326 c.908A>G p.K303R Chr.19: 47270412-47270412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747221939 Chr.19: 47270734-47270734 on Build GRCh38
rs374668118 Chr.19: 47270364-47270364 on Build GRCh38
rs778078938 Chr.19: 47270602-47270602 on Build GRCh38
rs758523905 Chr.19: 47270441-47270441 on Build GRCh38
rs558396002 Chr.19: 47270609-47270609 on Build GRCh38
rs1051605316 Chr.19: 47270776-47270776 on Build GRCh38
rs773996092 Chr.19: 47270479-47270479 on Build GRCh38
rs118041406 Chr.19: 47270471-47270471 on Build GRCh38
rs750554274 Chr.19: 47270433-47270433 on Build GRCh38
rs774616334 Chr.19: 47270612-47270612 on Build GRCh38
rs775240151 Chr.19: 47270577-47270577 on Build GRCh38
rs777857979 Chr.19: 47270721-47270721 on Build GRCh38
rs754630659 Chr.19: 47270714-47270714 on Build GRCh38
rs767965640 Chr.19: 47270523-47270523 on Build GRCh38
rs748680082 Chr.19: 47270530-47270530 on Build GRCh38
rs977196829 Chr.19: 47270780-47270780 on Build GRCh38
rs1024183042 Chr.19: 47270743-47270743 on Build GRCh38
rs146630699 Chr.19: 47270753-47270753 on Build GRCh38
rs759811174 Chr.19: 47270615-47270615 on Build GRCh38
rs147599888 Chr.19: 47270498-47270499 on Build GRCh38
rs992661765 Chr.19: 47270511-47270511 on Build GRCh38
rs188221636 Chr.19: 47270674-47270674 on Build GRCh38
rs764267487 Chr.19: 47270678-47270678 on Build GRCh38
rs764449548 Chr.19: 47270632-47270632 on Build GRCh38
rs888838 Chr.19: 47270529-47270529 on Build GRCh38
rs760495782 Chr.19: 47270489-47270489 on Build GRCh38
rs888837 Chr.19: 47270716-47270716 on Build GRCh38
rs978888180 Chr.19: 47270450-47270450 on Build GRCh38
rs563132818 Chr.19: 47270696-47270696 on Build GRCh38
rs756686739 Chr.19: 47270451-47270451 on Build GRCh38
rs932013669 Chr.19: 47270754-47270754 on Build GRCh38
rs755357473 Chr.19: 47270518-47270518 on Build GRCh38
rs551514778 Chr.19: 47270601-47270601 on Build GRCh38
rs543129639 Chr.19: 47270685-47270685 on Build GRCh38
rs764138396 Chr.19: 47270493-47270493 on Build GRCh38
rs147977663 Chr.19: 47270413-47270413 on Build GRCh38
rs758747507 Chr.19: 47270367-47270367 on Build GRCh38
rs780956231 Chr.19: 47270655-47270655 on Build GRCh38
rs376597834 Chr.19: 47270452-47270452 on Build GRCh38
rs753163249 Chr.19: 47270528-47270528 on Build GRCh38
rs569465656 Chr.19: 47270400-47270400 on Build GRCh38
rs762896813 Chr.19: 47270585-47270585 on Build GRCh38
rs745548599 Chr.19: 47270571-47270571 on Build GRCh38
rs958656284 Chr.19: 47270731-47270731 on Build GRCh38
rs199892243 Chr.19: 47270673-47270673 on Build GRCh38
rs371160012 Chr.19: 47270582-47270582 on Build GRCh38
rs374725472 Chr.19: 47270708-47270708 on Build GRCh38
rs776631253 Chr.19: 47270584-47270584 on Build GRCh38
rs372768345 Chr.19: 47270407-47270407 on Build GRCh38
rs765788216 Chr.19: 47270698-47270698 on Build GRCh38
rs771751378 Chr.19: 47270576-47270576 on Build GRCh38
rs534274243 Chr.19: 47270649-47270649 on Build GRCh38
rs747423442 Chr.19: 47270382-47270382 on Build GRCh38
rs752643878 Chr.19: 47270419-47270419 on Build GRCh38
rs774005372 Chr.19: 47270404-47270404 on Build GRCh38
rs374156415 Chr.19: 47270740-47270740 on Build GRCh38
rs758961738 Chr.19: 47270504-47270505 on Build GRCh38
rs769851329 Chr.19: 47270719-47270719 on Build GRCh38
rs144536796 Chr.19: 47270568-47270568 on Build GRCh38
rs73061366 Chr.19: 47270390-47270390 on Build GRCh38
rs138726882 Chr.19: 47270448-47270448 on Build GRCh38
rs768808563 Chr.19: 47270738-47270738 on Build GRCh38
rs71363731 Chr.19: 47270499-47270499 on Build GRCh38
rs746876329 Chr.19: 47270578-47270578 on Build GRCh38
rs143949045 Chr.19: 47270654-47270654 on Build GRCh38
rs900622975 Chr.19: 47270748-47270748 on Build GRCh38
rs774561026 Chr.19: 47270668-47270668 on Build GRCh38
rs372250323 Chr.19: 47270351-47270351 on Build GRCh38
rs571729241 Chr.19: 47270616-47270616 on Build GRCh38
rs759392795 Chr.19: 47270411-47270411 on Build GRCh38
rs772300699 Chr.19: 47270732-47270732 on Build GRCh38
rs147165767 Chr.19: 47270430-47270430 on Build GRCh38
rs925738058 Chr.19: 47270788-47270788 on Build GRCh38
rs771874321 Chr.19: 47270486-47270486 on Build GRCh38
rs567723154 Chr.19: 47270432-47270432 on Build GRCh38
rs200778283 Chr.19: 47270570-47270570 on Build GRCh38
rs777037057 Chr.19: 47270739-47270739 on Build GRCh38
rs565297764 Chr.19: 47270391-47270391 on Build GRCh38
rs996267465 Chr.19: 47270642-47270642 on Build GRCh38
rs990933050 Chr.19: 47270464-47270464 on Build GRCh38
rs747607469 Chr.19: 47270385-47270385 on Build GRCh38
rs958476074 Chr.19: 47270677-47270677 on Build GRCh38
rs781766879 Chr.19: 47270383-47270383 on Build GRCh38
rs778362376 Chr.19: 47270634-47270634 on Build GRCh38
rs745887102 Chr.19: 47270725-47270725 on Build GRCh38
rs748090179 Chr.19: 47270661-47270661 on Build GRCh38
rs545740296 Chr.19: 47270710-47270710 on Build GRCh38
rs201850534 Chr.19: 47270697-47270697 on Build GRCh38
rs770485049 Chr.19: 47270476-47270476 on Build GRCh38
rs753108097 Chr.19: 47270624-47270624 on Build GRCh38
rs775534185 Chr.19: 47270733-47270733 on Build GRCh38
rs376220544 Chr.19: 47270640-47270640 on Build GRCh38
rs760944969 Chr.19: 47270370-47270370 on Build GRCh38
rs770667368 Chr.19: 47270401-47270401 on Build GRCh38
rs769904342 Chr.19: 47270665-47270665 on Build GRCh38
rs757904822 Chr.19: 47270706-47270706 on Build GRCh38
rs751963757 Chr.19: 47270447-47270447 on Build GRCh38
rs779744692 Chr.19: 47270647-47270647 on Build GRCh38
rs201743544 Chr.19: 47270608-47270608 on Build GRCh38
rs766677996 Chr.19: 47270442-47270442 on Build GRCh38
rs768469405 Chr.19: 47270581-47270581 on Build GRCh38
rs751857280 Chr.19: 47270507-47270507 on Build GRCh38
rs761106079 Chr.19: 47270629-47270629 on Build GRCh38
rs368199034 Chr.19: 47270715-47270715 on Build GRCh38
rs780247055 Chr.19: 47270379-47270379 on Build GRCh38
rs749928362 Chr.19: 47270701-47270701 on Build GRCh38
rs754391036 Chr.19: 47270633-47270633 on Build GRCh38
rs373016166 Chr.19: 47270485-47270485 on Build GRCh38

View Full Product Details