Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 47725733-47726238 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EHD2
Entrez Gene ID
30846
Gene Name
EH domain containing 2
Gene Aliases
PAST2
UniGene
Hs.744963
Gene Chromosome Location
Chr.19: 47713344-47743134 on Build GRCh38
Transcripts
NM_014601.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
998787 c.585G>A p.E195E Chr.19: 47725894-47725894 on Build GRCh38
5486064 c.621G>A p.L207L Chr.19: 47725930-47725930 on Build GRCh38
1394947 c.550C>T p.L184F Chr.19: 47725859-47725859 on Build GRCh38
4127748 c.556A>T p.I186F Chr.19: 47725865-47725865 on Build GRCh38
5516849 c.617C>T p.A206V Chr.19: 47725926-47725926 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs149479756 Chr.19: 47725829-47725829 on Build GRCh38
rs763394764 Chr.19: 47725922-47725922 on Build GRCh38
rs765525694 Chr.19: 47725913-47725913 on Build GRCh38
rs924389369 Chr.19: 47725911-47725912 on Build GRCh38
rs948649576 Chr.19: 47725984-47725984 on Build GRCh38
rs775649453 Chr.19: 47725795-47725795 on Build GRCh38
rs773625785 Chr.19: 47726166-47726166 on Build GRCh38
rs754240911 Chr.19: 47725824-47725824 on Build GRCh38
rs376420269 Chr.19: 47725876-47725876 on Build GRCh38
rs1006684929 Chr.19: 47725985-47725985 on Build GRCh38
rs752344653 Chr.19: 47726102-47726102 on Build GRCh38
rs754731075 Chr.19: 47725857-47725857 on Build GRCh38
rs751158458 Chr.19: 47725934-47725934 on Build GRCh38
rs771422226 Chr.19: 47726169-47726169 on Build GRCh38
rs969536924 Chr.19: 47726138-47726138 on Build GRCh38
rs760909203 Chr.19: 47725814-47725814 on Build GRCh38
rs553547501 Chr.19: 47725828-47725828 on Build GRCh38
rs367574070 Chr.19: 47725986-47725986 on Build GRCh38
rs775008579 Chr.19: 47726071-47726071 on Build GRCh38
rs147314520 Chr.19: 47725932-47725932 on Build GRCh38
rs547172741 Chr.19: 47726076-47726076 on Build GRCh38
rs928139468 Chr.19: 47726160-47726160 on Build GRCh38
rs748350898 Chr.19: 47726018-47726018 on Build GRCh38
rs774957397 Chr.19: 47726183-47726183 on Build GRCh38
rs202143335 Chr.19: 47725921-47725921 on Build GRCh38
rs951479746 Chr.19: 47726020-47726020 on Build GRCh38
rs965645306 Chr.19: 47726216-47726216 on Build GRCh38
rs772046356 Chr.19: 47725786-47725786 on Build GRCh38
rs760139648 Chr.19: 47726205-47726205 on Build GRCh38
rs779463819 Chr.19: 47725850-47725850 on Build GRCh38
rs533779192 Chr.19: 47725765-47725765 on Build GRCh38
rs755769922 Chr.19: 47726104-47726104 on Build GRCh38
rs757859585 Chr.19: 47725848-47725848 on Build GRCh38
rs781274712 Chr.19: 47726146-47726146 on Build GRCh38
rs749225110 Chr.19: 47725966-47725966 on Build GRCh38
rs190123450 Chr.19: 47725936-47725936 on Build GRCh38
rs143975739 Chr.19: 47725889-47725889 on Build GRCh38
rs769953115 Chr.19: 47726159-47726159 on Build GRCh38
rs137935701 Chr.19: 47725812-47725812 on Build GRCh38
rs1028472701 Chr.19: 47726099-47726099 on Build GRCh38
rs760855823 Chr.19: 47725897-47725897 on Build GRCh38
rs770150064 Chr.19: 47726035-47726035 on Build GRCh38
rs909945966 Chr.19: 47725785-47725785 on Build GRCh38
rs148558427 Chr.19: 47725892-47725892 on Build GRCh38
rs200207567 Chr.19: 47725816-47725816 on Build GRCh38
rs777450158 Chr.19: 47725955-47725955 on Build GRCh38
rs577413598 Chr.19: 47725759-47725759 on Build GRCh38
rs763536721 Chr.19: 47725790-47725790 on Build GRCh38
rs753293483 Chr.19: 47726214-47726214 on Build GRCh38
rs1025530886 Chr.19: 47726014-47726014 on Build GRCh38
rs544914999 Chr.19: 47726011-47726011 on Build GRCh38
rs781127268 Chr.19: 47725867-47725867 on Build GRCh38
rs150300804 Chr.19: 47726147-47726147 on Build GRCh38
rs757181096 Chr.19: 47725976-47725976 on Build GRCh38
rs199961822 Chr.19: 47726152-47726152 on Build GRCh38
rs369840650 Chr.19: 47725943-47725943 on Build GRCh38
rs755011408 Chr.19: 47726141-47726141 on Build GRCh38
rs984101500 Chr.19: 47726084-47726084 on Build GRCh38
rs769487418 Chr.19: 47725881-47725881 on Build GRCh38
rs763918440 Chr.19: 47726109-47726109 on Build GRCh38
rs777696254 Chr.19: 47725887-47725887 on Build GRCh38
rs767216948 Chr.19: 47725940-47725940 on Build GRCh38
rs556634967 Chr.19: 47725783-47725783 on Build GRCh38
rs781570240 Chr.19: 47726007-47726007 on Build GRCh38
rs769833491 Chr.19: 47725761-47725761 on Build GRCh38
rs767163876 Chr.19: 47726098-47726098 on Build GRCh38
rs762224358 Chr.19: 47725825-47725825 on Build GRCh38
rs757175078 Chr.19: 47726114-47726114 on Build GRCh38
rs749553791 Chr.19: 47726168-47726168 on Build GRCh38
rs755895230 Chr.19: 47725948-47725948 on Build GRCh38
rs996023845 Chr.19: 47726096-47726096 on Build GRCh38
rs747150177 Chr.19: 47725996-47725996 on Build GRCh38
rs560354382 Chr.19: 47726111-47726111 on Build GRCh38
rs759598202 Chr.19: 47725766-47725766 on Build GRCh38
rs751209627 Chr.19: 47725851-47725851 on Build GRCh38
rs576144192 Chr.19: 47726008-47726008 on Build GRCh38
rs767063664 Chr.19: 47726063-47726063 on Build GRCh38
rs533522326 Chr.19: 47726062-47726062 on Build GRCh38
rs8111184 Chr.19: 47725912-47725912 on Build GRCh38
rs768934406 Chr.19: 47725900-47725900 on Build GRCh38
rs776967729 Chr.19: 47725904-47725904 on Build GRCh38
rs778765086 Chr.19: 47726122-47726122 on Build GRCh38
rs564807856 Chr.19: 47726056-47726056 on Build GRCh38
rs867611192 Chr.19: 47726126-47726126 on Build GRCh38
rs753302685 Chr.19: 47725776-47725777 on Build GRCh38
rs745812114 Chr.19: 47726132-47726132 on Build GRCh38
rs1019831845 Chr.19: 47726158-47726158 on Build GRCh38
rs1043214730 Chr.19: 47725954-47725954 on Build GRCh38

View Full Product Details