Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48303855-48304332 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC114
Entrez Gene ID
93233
Gene Name
coiled-coil domain containing 114
Gene Aliases
CILD20
UniGene
Hs.112645
Gene Chromosome Location
Chr.19: 48296452-48322793 on Build GRCh38
Transcripts
NM_144577.3 XM_005259413.3 XM_005259414.3 XM_005259415.3 XM_005259416.4 XM_011527515.2 XM_011527516.2 XM_011527517.1 XM_017027483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7127303 c.681C>T p.L227L Chr.19: 48304014-48304014 on Build GRCh38
3766392 c.555-4G>A p.? Chr.19: 48304144-48304144 on Build GRCh38
1494357 c.84G>A p.Q28Q Chr.19: 48303990-48303990 on Build GRCh38
6322317 c.657C>A p.H219Q Chr.19: 48304038-48304038 on Build GRCh38
7127977 c.592C>T p.R198C Chr.19: 48304103-48304103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749886957 Chr.19: 48304212-48304212 on Build GRCh38
rs761545928 Chr.19: 48304122-48304122 on Build GRCh38
rs367561911 Chr.19: 48303950-48303950 on Build GRCh38
rs749128355 Chr.19: 48303911-48303911 on Build GRCh38
rs56177753 Chr.19: 48304144-48304144 on Build GRCh38
rs936558738 Chr.19: 48304232-48304232 on Build GRCh38
rs564694520 Chr.19: 48304044-48304044 on Build GRCh38
rs533885449 Chr.19: 48303972-48303972 on Build GRCh38
rs886038729 Chr.19: 48304155-48304155 on Build GRCh38
rs200808081 Chr.19: 48303999-48303999 on Build GRCh38
rs368483838 Chr.19: 48304101-48304101 on Build GRCh38
rs28582401 Chr.19: 48304110-48304110 on Build GRCh38
rs374990723 Chr.19: 48304099-48304099 on Build GRCh38
rs185349587 Chr.19: 48304179-48304179 on Build GRCh38
rs746508737 Chr.19: 48304112-48304112 on Build GRCh38
rs538126564 Chr.19: 48304156-48304156 on Build GRCh38
rs114078313 Chr.19: 48304076-48304076 on Build GRCh38
rs1041880653 Chr.19: 48303879-48303879 on Build GRCh38
rs755043090 Chr.19: 48304001-48304001 on Build GRCh38
rs201877590 Chr.19: 48304132-48304132 on Build GRCh38
rs575514518 Chr.19: 48304021-48304021 on Build GRCh38
rs755132968 Chr.19: 48304098-48304098 on Build GRCh38
rs147718607 Chr.19: 48303953-48303953 on Build GRCh38
rs1029945467 Chr.19: 48304131-48304131 on Build GRCh38
rs977493681 Chr.19: 48304256-48304256 on Build GRCh38
rs148382103 Chr.19: 48303987-48303987 on Build GRCh38
rs1013618358 Chr.19: 48303877-48303877 on Build GRCh38
rs754039474 Chr.19: 48304149-48304149 on Build GRCh38
rs867613985 Chr.19: 48304017-48304017 on Build GRCh38
rs534010981 Chr.19: 48304159-48304159 on Build GRCh38
rs775044044 Chr.19: 48304111-48304111 on Build GRCh38
rs192848164 Chr.19: 48304100-48304100 on Build GRCh38
rs761608877 Chr.19: 48304079-48304079 on Build GRCh38
rs145629439 Chr.19: 48303998-48303998 on Build GRCh38
rs765089177 Chr.19: 48304089-48304089 on Build GRCh38
rs113520498 Chr.19: 48304103-48304103 on Build GRCh38
rs935740499 Chr.19: 48304276-48304276 on Build GRCh38
rs773791686 Chr.19: 48303965-48303965 on Build GRCh38
rs142846668 Chr.19: 48304024-48304024 on Build GRCh38
rs34628933 Chr.19: 48303981-48303981 on Build GRCh38
rs991341986 Chr.19: 48304207-48304207 on Build GRCh38
rs773119584 Chr.19: 48304139-48304139 on Build GRCh38
rs760357218 Chr.19: 48304072-48304072 on Build GRCh38
rs201621335 Chr.19: 48304007-48304007 on Build GRCh38
rs763841580 Chr.19: 48303990-48303990 on Build GRCh38
rs746224399 Chr.19: 48304167-48304167 on Build GRCh38
rs1029658127 Chr.19: 48303968-48303968 on Build GRCh38
rs958561432 Chr.19: 48304190-48304190 on Build GRCh38
rs747107469 Chr.19: 48303935-48303935 on Build GRCh38
rs200168343 Chr.19: 48304109-48304109 on Build GRCh38
rs145848971 Chr.19: 48303889-48303889 on Build GRCh38
rs145107907 Chr.19: 48304295-48304295 on Build GRCh38
rs767173437 Chr.19: 48303984-48303984 on Build GRCh38
rs34888501 Chr.19: 48304144-48304145 on Build GRCh38
rs199960441 Chr.19: 48304108-48304108 on Build GRCh38
rs762551198 Chr.19: 48304040-48304040 on Build GRCh38
rs773803395 Chr.19: 48303974-48303974 on Build GRCh38
rs746503231 Chr.19: 48304188-48304188 on Build GRCh38
rs371432008 Chr.19: 48304095-48304095 on Build GRCh38
rs571414944 Chr.19: 48304211-48304211 on Build GRCh38
rs776279484 Chr.19: 48304121-48304121 on Build GRCh38
rs986502176 Chr.19: 48303977-48303977 on Build GRCh38
rs752887454 Chr.19: 48304145-48304145 on Build GRCh38
rs764230566 Chr.19: 48304148-48304148 on Build GRCh38
rs866968571 Chr.19: 48303962-48303962 on Build GRCh38
rs200495728 Chr.19: 48303934-48303934 on Build GRCh38
rs774954250 Chr.19: 48304066-48304066 on Build GRCh38
rs925201151 Chr.19: 48304038-48304038 on Build GRCh38
rs115621120 Chr.19: 48303910-48303910 on Build GRCh38
rs757192666 Chr.19: 48303928-48303928 on Build GRCh38
rs758961278 Chr.19: 48304170-48304170 on Build GRCh38
rs955384540 Chr.19: 48303980-48303980 on Build GRCh38
rs1054982940 Chr.19: 48304243-48304243 on Build GRCh38
rs149194036 Chr.19: 48303964-48303964 on Build GRCh38
rs547074727 Chr.19: 48304077-48304077 on Build GRCh38
rs776898770 Chr.19: 48303994-48303994 on Build GRCh38
rs138256591 Chr.19: 48304008-48304008 on Build GRCh38
rs768205609 Chr.19: 48304119-48304119 on Build GRCh38
rs963121561 Chr.19: 48304181-48304181 on Build GRCh38
rs925233835 Chr.19: 48304231-48304231 on Build GRCh38
rs749664448 Chr.19: 48304032-48304032 on Build GRCh38
rs949711483 Chr.19: 48304235-48304235 on Build GRCh38
rs771518813 Chr.19: 48304065-48304065 on Build GRCh38
rs756163196 Chr.19: 48304147-48304147 on Build GRCh38
rs759075558 Chr.19: 48304052-48304052 on Build GRCh38
rs374808728 Chr.19: 48303947-48303947 on Build GRCh38
rs760854519 Chr.19: 48304085-48304087 on Build GRCh38
rs772744749 Chr.19: 48304189-48304189 on Build GRCh38
rs766667522 Chr.19: 48303927-48303928 on Build GRCh38
rs766995142 Chr.19: 48304270-48304270 on Build GRCh38
rs774866725 Chr.19: 48304051-48304051 on Build GRCh38
rs760482173 Chr.19: 48303988-48303988 on Build GRCh38
rs949503248 Chr.19: 48304305-48304305 on Build GRCh38
rs544515139 Chr.19: 48304023-48304023 on Build GRCh38
rs141590182 Chr.19: 48303995-48303995 on Build GRCh38

View Full Product Details