Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48306146-48306647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC114
Entrez Gene ID
93233
Gene Name
coiled-coil domain containing 114
Gene Aliases
CILD20
UniGene
Hs.112645
Gene Chromosome Location
Chr.19: 48296452-48322793 on Build GRCh38
Transcripts
NM_144577.3 XM_005259413.3 XM_005259414.3 XM_005259415.3 XM_005259416.4 XM_011527515.2 XM_011527516.2 XM_011527517.1 XM_017027483.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3362939 c.546C>T p.Y182Y Chr.19: 48306264-48306264 on Build GRCh38
998928 c.550G>A p.V184I Chr.19: 48306260-48306260 on Build GRCh38
3823538 c.535A>C p.T179P Chr.19: 48306275-48306275 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs961609745 Chr.19: 48306510-48306510 on Build GRCh38
rs567924125 Chr.19: 48306250-48306250 on Build GRCh38
rs923203517 Chr.19: 48306575-48306575 on Build GRCh38
rs956704954 Chr.19: 48306201-48306201 on Build GRCh38
rs558965790 Chr.19: 48306409-48306409 on Build GRCh38
rs994361953 Chr.19: 48306520-48306520 on Build GRCh38
rs1053333723 Chr.19: 48306387-48306387 on Build GRCh38
rs1012034183 Chr.19: 48306419-48306419 on Build GRCh38
rs953433588 Chr.19: 48306278-48306278 on Build GRCh38
rs10416590 Chr.19: 48306328-48306328 on Build GRCh38
rs563328584 Chr.19: 48306612-48306612 on Build GRCh38
rs565536263 Chr.19: 48306198-48306198 on Build GRCh38
rs1048049367 Chr.19: 48306439-48306439 on Build GRCh38
rs1011195858 Chr.19: 48306618-48306618 on Build GRCh38
rs753203701 Chr.19: 48306369-48306369 on Build GRCh38
rs934554781 Chr.19: 48306598-48306598 on Build GRCh38
rs759674548 Chr.19: 48306224-48306224 on Build GRCh38
rs762832505 Chr.19: 48306264-48306264 on Build GRCh38
rs569437283 Chr.19: 48306208-48306208 on Build GRCh38
rs934523693 Chr.19: 48306376-48306376 on Build GRCh38
rs765299412 Chr.19: 48306237-48306240 on Build GRCh38
rs773209379 Chr.19: 48306263-48306263 on Build GRCh38
rs757562950 Chr.19: 48306349-48306349 on Build GRCh38
rs961579491 Chr.19: 48306242-48306242 on Build GRCh38
rs606231239 Chr.19: 48306325-48306325 on Build GRCh38
rs574369936 Chr.19: 48306556-48306556 on Build GRCh38
rs760703306 Chr.19: 48306309-48306309 on Build GRCh38
rs773608080 Chr.19: 48306613-48306613 on Build GRCh38
rs918506361 Chr.19: 48306413-48306413 on Build GRCh38
rs753828189 Chr.19: 48306329-48306329 on Build GRCh38
rs970339985 Chr.19: 48306196-48306196 on Build GRCh38
rs566331011 Chr.19: 48306524-48306524 on Build GRCh38
rs1004133745 Chr.19: 48306599-48306599 on Build GRCh38
rs769301117 Chr.19: 48306260-48306260 on Build GRCh38
rs142314893 Chr.19: 48306429-48306429 on Build GRCh38
rs1044818108 Chr.19: 48306541-48306541 on Build GRCh38
rs923172494 Chr.19: 48306356-48306356 on Build GRCh38
rs372479683 Chr.19: 48306458-48306458 on Build GRCh38
rs1022343513 Chr.19: 48306267-48306267 on Build GRCh38
rs576523977 Chr.19: 48306392-48306392 on Build GRCh38
rs905894303 Chr.19: 48306426-48306426 on Build GRCh38
rs1002869413 Chr.19: 48306430-48306430 on Build GRCh38
rs975706967 Chr.19: 48306570-48306570 on Build GRCh38
rs538756035 Chr.19: 48306397-48306397 on Build GRCh38
rs150851681 Chr.19: 48306567-48306567 on Build GRCh38
rs866776174 Chr.19: 48306311-48306311 on Build GRCh38
rs929617684 Chr.19: 48306421-48306421 on Build GRCh38
rs536856228 Chr.19: 48306302-48306302 on Build GRCh38
rs1017347064 Chr.19: 48306563-48306563 on Build GRCh38
rs747885546 Chr.19: 48306245-48306245 on Build GRCh38
rs548117033 Chr.19: 48306244-48306244 on Build GRCh38
rs755454266 Chr.19: 48306207-48306207 on Build GRCh38
rs919043631 Chr.19: 48306294-48306294 on Build GRCh38
rs367799104 Chr.19: 48306295-48306295 on Build GRCh38
rs534550383 Chr.19: 48306206-48306206 on Build GRCh38
rs547313988 Chr.19: 48306523-48306523 on Build GRCh38
rs146512076 Chr.19: 48306476-48306476 on Build GRCh38
rs943886051 Chr.19: 48306353-48306353 on Build GRCh38

View Full Product Details