Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48325252-48325762 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMP3
Entrez Gene ID
2014
Gene Name
epithelial membrane protein 3
Gene Aliases
YMP
UniGene
Hs.9999
Gene Chromosome Location
Chr.19: 48325372-48330555 on Build GRCh38
Transcripts
NM_001425.2 XM_011526605.2

SNP Detail

SNP ID Chromosome Location
rs867931764 Chr.19: 48325476-48325476 on Build GRCh38
rs903136990 Chr.19: 48325602-48325603 on Build GRCh38
rs936953632 Chr.19: 48325587-48325587 on Build GRCh38
rs773645295 Chr.19: 48325688-48325688 on Build GRCh38
rs543162407 Chr.19: 48325400-48325400 on Build GRCh38
rs574201102 Chr.19: 48325398-48325398 on Build GRCh38
rs991876039 Chr.19: 48325547-48325547 on Build GRCh38
rs1039084852 Chr.19: 48325388-48325388 on Build GRCh38
rs967280165 Chr.19: 48325553-48325553 on Build GRCh38
rs945612818 Chr.19: 48325333-48325333 on Build GRCh38
rs1033323010 Chr.19: 48325516-48325516 on Build GRCh38
rs898440269 Chr.19: 48325494-48325494 on Build GRCh38
rs959073709 Chr.19: 48325522-48325522 on Build GRCh38
rs974671950 Chr.19: 48325548-48325548 on Build GRCh38
rs941486266 Chr.19: 48325297-48325297 on Build GRCh38
rs762364169 Chr.19: 48325446-48325446 on Build GRCh38
rs995550398 Chr.19: 48325511-48325511 on Build GRCh38
rs890118280 Chr.19: 48325399-48325400 on Build GRCh38
rs978744449 Chr.19: 48325335-48325335 on Build GRCh38
rs1008775693 Chr.19: 48325400-48325403 on Build GRCh38
rs886548899 Chr.19: 48325662-48325662 on Build GRCh38
rs959127422 Chr.19: 48325307-48325307 on Build GRCh38
rs917124950 Chr.19: 48325519-48325519 on Build GRCh38
rs900560039 Chr.19: 48325391-48325391 on Build GRCh38
rs1050147060 Chr.19: 48325576-48325576 on Build GRCh38
rs948526228 Chr.19: 48325523-48325523 on Build GRCh38
rs890120576 Chr.19: 48325577-48325577 on Build GRCh38
rs908713694 Chr.19: 48325282-48325282 on Build GRCh38
rs936651493 Chr.19: 48325352-48325352 on Build GRCh38
rs939836544 Chr.19: 48325684-48325684 on Build GRCh38
rs563673256 Chr.19: 48325739-48325739 on Build GRCh38
rs879276648 Chr.19: 48325459-48325459 on Build GRCh38
rs1000049741 Chr.19: 48325625-48325625 on Build GRCh38
rs999956380 Chr.19: 48325441-48325441 on Build GRCh38
rs867006077 Chr.19: 48325593-48325593 on Build GRCh38
rs558539089 Chr.19: 48325474-48325474 on Build GRCh38
rs908767734 Chr.19: 48325532-48325532 on Build GRCh38
rs549755296 Chr.19: 48325290-48325290 on Build GRCh38
rs992261170 Chr.19: 48325309-48325309 on Build GRCh38
rs1015907523 Chr.19: 48325455-48325455 on Build GRCh38
rs1050555779 Chr.19: 48325365-48325365 on Build GRCh38
rs1033296622 Chr.19: 48325626-48325626 on Build GRCh38
rs548788840 Chr.19: 48325429-48325431 on Build GRCh38
rs918086641 Chr.19: 48325320-48325320 on Build GRCh38
rs185841956 Chr.19: 48325444-48325444 on Build GRCh38
rs80096621 Chr.19: 48325568-48325568 on Build GRCh38
rs1050876579 Chr.19: 48325433-48325433 on Build GRCh38
rs944350066 Chr.19: 48325595-48325595 on Build GRCh38
rs767909176 Chr.19: 48325702-48325702 on Build GRCh38
rs113582815 Chr.19: 48325633-48325633 on Build GRCh38
rs978310666 Chr.19: 48325560-48325560 on Build GRCh38
rs925598721 Chr.19: 48325340-48325340 on Build GRCh38
rs1051841423 Chr.19: 48325656-48325656 on Build GRCh38
rs921932136 Chr.19: 48325607-48325607 on Build GRCh38
rs979879275 Chr.19: 48325534-48325534 on Build GRCh38
rs538934325 Chr.19: 48325464-48325464 on Build GRCh38
rs933358850 Chr.19: 48325623-48325623 on Build GRCh38
rs1024710824 Chr.19: 48325549-48325549 on Build GRCh38
rs73587206 Chr.19: 48325600-48325600 on Build GRCh38
rs776610115 Chr.19: 48325371-48325371 on Build GRCh38
rs921441490 Chr.19: 48325564-48325564 on Build GRCh38

View Full Product Details