Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48636768-48637260 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBP
Entrez Gene ID
1628
Gene Name
D-box binding PAR bZIP transcription factor
Gene Aliases
DABP
UniGene
Hs.414480
Gene Chromosome Location
Chr.19: 48630560-48637550 on Build GRCh38
Transcripts
NM_001352.4 XM_017026388.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5833805 c.68delC p.P23fs*27 Chr.19: 48636927-48636927 on Build GRCh38
4521401 c.110G>A p.G37E Chr.19: 48636885-48636885 on Build GRCh38
6640290 c.55G>A p.A19T Chr.19: 48636940-48636940 on Build GRCh38
4079891 c.78A>G p.G26G Chr.19: 48636917-48636917 on Build GRCh38
4676302 c.57C>A p.A19A Chr.19: 48636938-48636938 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs540846669 Chr.19: 48637111-48637111 on Build GRCh38
rs567875372 Chr.19: 48636873-48636873 on Build GRCh38
rs555252046 Chr.19: 48637087-48637087 on Build GRCh38
rs769874117 Chr.19: 48636884-48636884 on Build GRCh38
rs777295484 Chr.19: 48636820-48636820 on Build GRCh38
rs756874787 Chr.19: 48636929-48636929 on Build GRCh38
rs187361350 Chr.19: 48637120-48637120 on Build GRCh38
rs948109274 Chr.19: 48637069-48637069 on Build GRCh38
rs891086617 Chr.19: 48636789-48636789 on Build GRCh38
rs150228428 Chr.19: 48636843-48636843 on Build GRCh38
rs763245265 Chr.19: 48636895-48636895 on Build GRCh38
rs778734663 Chr.19: 48636944-48636944 on Build GRCh38
rs546355807 Chr.19: 48637025-48637025 on Build GRCh38
rs371234571 Chr.19: 48636922-48636922 on Build GRCh38
rs780823876 Chr.19: 48636806-48636806 on Build GRCh38
rs1057020 Chr.19: 48637052-48637052 on Build GRCh38
rs777035889 Chr.19: 48637233-48637233 on Build GRCh38
rs758141677 Chr.19: 48636961-48636961 on Build GRCh38
rs1031273808 Chr.19: 48637227-48637227 on Build GRCh38
rs769159834 Chr.19: 48637154-48637154 on Build GRCh38
rs551268466 Chr.19: 48636927-48636927 on Build GRCh38
rs774659051 Chr.19: 48637008-48637008 on Build GRCh38
rs956758144 Chr.19: 48636972-48636972 on Build GRCh38
rs773393801 Chr.19: 48637000-48637000 on Build GRCh38
rs755615176 Chr.19: 48636923-48636923 on Build GRCh38
rs377129998 Chr.19: 48636812-48636812 on Build GRCh38
rs897046127 Chr.19: 48637147-48637147 on Build GRCh38
rs375881110 Chr.19: 48636939-48636939 on Build GRCh38
rs745848804 Chr.19: 48636857-48636857 on Build GRCh38
rs1048933828 Chr.19: 48637226-48637226 on Build GRCh38
rs1048508752 Chr.19: 48637160-48637160 on Build GRCh38
rs35982754 Chr.19: 48637095-48637095 on Build GRCh38
rs994226903 Chr.19: 48636819-48636819 on Build GRCh38
rs1057078784 Chr.19: 48637137-48637137 on Build GRCh38
rs745673197 Chr.19: 48636960-48636960 on Build GRCh38
rs748177167 Chr.19: 48636881-48636881 on Build GRCh38
rs1013217386 Chr.19: 48636937-48636937 on Build GRCh38
rs201132033 Chr.19: 48636924-48636924 on Build GRCh38
rs994708145 Chr.19: 48637205-48637205 on Build GRCh38
rs576049417 Chr.19: 48637015-48637015 on Build GRCh38
rs753181059 Chr.19: 48636917-48636917 on Build GRCh38
rs755508163 Chr.19: 48636805-48636805 on Build GRCh38
rs1037143121 Chr.19: 48637157-48637157 on Build GRCh38
rs771266714 Chr.19: 48637007-48637007 on Build GRCh38
rs761226449 Chr.19: 48637036-48637036 on Build GRCh38
rs772479170 Chr.19: 48636907-48636907 on Build GRCh38
rs771888798 Chr.19: 48636865-48636865 on Build GRCh38
rs774849448 Chr.19: 48636905-48636905 on Build GRCh38
rs36008846 Chr.19: 48636925-48636925 on Build GRCh38
rs901374318 Chr.19: 48637221-48637221 on Build GRCh38
rs539393245 Chr.19: 48636940-48636940 on Build GRCh38
rs200399861 Chr.19: 48636886-48636886 on Build GRCh38
rs1057420277 Chr.19: 48636794-48636794 on Build GRCh38
rs763637827 Chr.19: 48637043-48637043 on Build GRCh38
rs370768785 Chr.19: 48636976-48636976 on Build GRCh38
rs779089789 Chr.19: 48637170-48637170 on Build GRCh38
rs1006607125 Chr.19: 48637177-48637177 on Build GRCh38
rs373978426 Chr.19: 48636838-48636838 on Build GRCh38
rs767923749 Chr.19: 48636916-48636916 on Build GRCh38
rs1018035078 Chr.19: 48637186-48637186 on Build GRCh38
rs569307976 Chr.19: 48636936-48636936 on Build GRCh38
rs531074622 Chr.19: 48636813-48636813 on Build GRCh38
rs747100539 Chr.19: 48637034-48637034 on Build GRCh38
rs921550007 Chr.19: 48637048-48637048 on Build GRCh38
rs746065208 Chr.19: 48636900-48636900 on Build GRCh38
rs990273421 Chr.19: 48637041-48637041 on Build GRCh38
rs779883115 Chr.19: 48636970-48636970 on Build GRCh38
rs373877433 Chr.19: 48637002-48637002 on Build GRCh38
rs748904050 Chr.19: 48636825-48636825 on Build GRCh38
rs573464315 Chr.19: 48637096-48637096 on Build GRCh38
rs557745939 Chr.19: 48636998-48636998 on Build GRCh38
rs766889742 Chr.19: 48636902-48636902 on Build GRCh38

View Full Product Details