Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 48960720-48961231 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BAX
Entrez Gene ID
581
Gene Name
BCL2 associated X, apoptosis regulator
Gene Aliases
BCL2L4
UniGene
Hs.624291
Gene Chromosome Location
Chr.19: 48954825-48961798 on Build GRCh38
Transcripts
NM_001291428.1 NM_001291429.1 NM_001291430.1 NM_001291431.1 NM_004324.3 NM_138761.3 NM_138763.3 NM_138764.4 NR_027882.1 XM_017027077.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5856349 c.452G>A p.W151* Chr.19: 48960892-48960892 on Build GRCh38
3108311 c.406A>C p.I136L Chr.19: 48960846-48960846 on Build GRCh38
3108315 c.430C>A p.L144I Chr.19: 48960870-48960870 on Build GRCh38
3536711 c.371C>T p.A124V Chr.19: 48960811-48960811 on Build GRCh38
6575294 c.581G>T p.S194I Chr.19: 48961021-48961021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755234872 Chr.19: 48960790-48960790 on Build GRCh38
rs759203778 Chr.19: 48961119-48961119 on Build GRCh38
rs773244034 Chr.19: 48961024-48961024 on Build GRCh38
rs538327173 Chr.19: 48960948-48960948 on Build GRCh38
rs767380946 Chr.19: 48961137-48961137 on Build GRCh38
rs751880977 Chr.19: 48961015-48961025 on Build GRCh38
rs755625957 Chr.19: 48960765-48960765 on Build GRCh38
rs77328789 Chr.19: 48960759-48960759 on Build GRCh38
rs781625726 Chr.19: 48960791-48960791 on Build GRCh38
rs887035918 Chr.19: 48961090-48961090 on Build GRCh38
rs569560296 Chr.19: 48961022-48961022 on Build GRCh38
rs753795347 Chr.19: 48960972-48960972 on Build GRCh38
rs887750546 Chr.19: 48960983-48960983 on Build GRCh38
rs746173568 Chr.19: 48960930-48960930 on Build GRCh38
rs929319102 Chr.19: 48961065-48961065 on Build GRCh38
rs747429913 Chr.19: 48961002-48961002 on Build GRCh38
rs776348320 Chr.19: 48961013-48961013 on Build GRCh38
rs750553916 Chr.19: 48961073-48961073 on Build GRCh38
rs752989631 Chr.19: 48960885-48960885 on Build GRCh38
rs780659742 Chr.19: 48961084-48961084 on Build GRCh38
rs749444930 Chr.19: 48960915-48960915 on Build GRCh38
rs759411396 Chr.19: 48961045-48961045 on Build GRCh38
rs774444543 Chr.19: 48961117-48961117 on Build GRCh38
rs754503498 Chr.19: 48961141-48961141 on Build GRCh38
rs753290709 Chr.19: 48961092-48961092 on Build GRCh38
rs536564321 Chr.19: 48961031-48961031 on Build GRCh38
rs900623653 Chr.19: 48960960-48960960 on Build GRCh38
rs750177497 Chr.19: 48960874-48960874 on Build GRCh38
rs372506919 Chr.19: 48960873-48960873 on Build GRCh38
rs755378794 Chr.19: 48961082-48961082 on Build GRCh38
rs769968939 Chr.19: 48961079-48961079 on Build GRCh38
rs762786630 Chr.19: 48960956-48960956 on Build GRCh38
rs375291870 Chr.19: 48960840-48960840 on Build GRCh38
rs773804974 Chr.19: 48960803-48960803 on Build GRCh38
rs781505807 Chr.19: 48960884-48960884 on Build GRCh38
rs1021296039 Chr.19: 48960752-48960752 on Build GRCh38
rs763965186 Chr.19: 48960774-48960774 on Build GRCh38
rs760132642 Chr.19: 48960846-48960846 on Build GRCh38
rs745721410 Chr.19: 48960789-48960789 on Build GRCh38
rs375097351 Chr.19: 48960976-48960976 on Build GRCh38
rs150314785 Chr.19: 48961145-48961145 on Build GRCh38
rs761631646 Chr.19: 48960871-48960871 on Build GRCh38
rs765332911 Chr.19: 48961070-48961070 on Build GRCh38
rs1035163287 Chr.19: 48961173-48961173 on Build GRCh38
rs777681381 Chr.19: 48960896-48960896 on Build GRCh38
rs922653645 Chr.19: 48961017-48961017 on Build GRCh38
rs760014274 Chr.19: 48960832-48960832 on Build GRCh38
rs772610950 Chr.19: 48960989-48960989 on Build GRCh38
rs774285236 Chr.19: 48961038-48961038 on Build GRCh38
rs762950892 Chr.19: 48961027-48961027 on Build GRCh38
rs190602136 Chr.19: 48960880-48960880 on Build GRCh38
rs369660551 Chr.19: 48960829-48960829 on Build GRCh38
rs573226978 Chr.19: 48961078-48961078 on Build GRCh38
rs566931225 Chr.19: 48961087-48961087 on Build GRCh38
rs538863351 Chr.19: 48961201-48961201 on Build GRCh38
rs778538796 Chr.19: 48960779-48960779 on Build GRCh38
rs763328495 Chr.19: 48960965-48960978 on Build GRCh38
rs753745806 Chr.19: 48960776-48960776 on Build GRCh38
rs768187175 Chr.19: 48961007-48961007 on Build GRCh38
rs779698557 Chr.19: 48961078-48961088 on Build GRCh38
rs552694023 Chr.19: 48961107-48961107 on Build GRCh38
rs1034046509 Chr.19: 48960797-48960797 on Build GRCh38
rs1025860658 Chr.19: 48961146-48961146 on Build GRCh38
rs778124480 Chr.19: 48960969-48960969 on Build GRCh38
rs772452241 Chr.19: 48960934-48960934 on Build GRCh38
rs150330824 Chr.19: 48960962-48960962 on Build GRCh38
rs375752615 Chr.19: 48960879-48960879 on Build GRCh38
rs960464394 Chr.19: 48961181-48961181 on Build GRCh38
rs142107485 Chr.19: 48960990-48960990 on Build GRCh38
rs755785366 Chr.19: 48961072-48961074 on Build GRCh38
rs369919235 Chr.19: 48960977-48960977 on Build GRCh38
rs769370752 Chr.19: 48961089-48961089 on Build GRCh38
rs910082433 Chr.19: 48961184-48961184 on Build GRCh38
rs768343456 Chr.19: 48960942-48960942 on Build GRCh38
rs904267924 Chr.19: 48961085-48961085 on Build GRCh38
rs779479574 Chr.19: 48960927-48960927 on Build GRCh38
rs767216467 Chr.19: 48960760-48960760 on Build GRCh38
rs751438320 Chr.19: 48960963-48960963 on Build GRCh38
rs753430330 Chr.19: 48960860-48960860 on Build GRCh38
rs1027659970 Chr.19: 48960974-48960974 on Build GRCh38
rs1001314589 Chr.19: 48961102-48961102 on Build GRCh38
rs747303365 Chr.19: 48961080-48961080 on Build GRCh38
rs772824562 Chr.19: 48961093-48961093 on Build GRCh38
rs764401066 Chr.19: 48960983-48960984 on Build GRCh38
rs943470258 Chr.19: 48960997-48960997 on Build GRCh38
rs372856198 Chr.19: 48960805-48960805 on Build GRCh38
rs757248443 Chr.19: 48961062-48961062 on Build GRCh38
rs998145625 Chr.19: 48960749-48960749 on Build GRCh38
rs200010531 Chr.19: 48960809-48960809 on Build GRCh38
rs775087033 Chr.19: 48960938-48960938 on Build GRCh38
rs951626301 Chr.19: 48961159-48961159 on Build GRCh38
rs763627965 Chr.19: 48960848-48960848 on Build GRCh38
rs988433784 Chr.19: 48960973-48960973 on Build GRCh38
rs371947281 Chr.19: 48960830-48960830 on Build GRCh38
rs750387674 Chr.19: 48960951-48960951 on Build GRCh38
rs763002429 Chr.19: 48961131-48961131 on Build GRCh38
rs764165341 Chr.19: 48961054-48961054 on Build GRCh38
rs769368417 Chr.19: 48961023-48961023 on Build GRCh38
rs746408654 Chr.19: 48960985-48960985 on Build GRCh38
rs747988714 Chr.19: 48960748-48960748 on Build GRCh38
rs760292142 Chr.19: 48960939-48960939 on Build GRCh38
rs753142726 Chr.19: 48960869-48960869 on Build GRCh38
rs4645900 Chr.19: 48961106-48961106 on Build GRCh38
rs765013477 Chr.19: 48960950-48960950 on Build GRCh38
rs373780166 Chr.19: 48961058-48961058 on Build GRCh38
rs376997564 Chr.19: 48960810-48960810 on Build GRCh38
rs369570399 Chr.19: 48960841-48960841 on Build GRCh38
rs988224920 Chr.19: 48961203-48961203 on Build GRCh38
rs752200085 Chr.19: 48960761-48960761 on Build GRCh38
rs756221659 Chr.19: 48960886-48960886 on Build GRCh38
rs137993558 Chr.19: 48960967-48960967 on Build GRCh38
rs368527574 Chr.19: 48960798-48960798 on Build GRCh38
rs141194773 Chr.19: 48960949-48960949 on Build GRCh38
rs771339205 Chr.19: 48960924-48960924 on Build GRCh38

View Full Product Details