Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49043834-49044340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CGB5
Entrez Gene ID
93659
Gene Name
chorionic gonadotropin beta subunit 5
Gene Aliases
CGB, HCG, hCGB
UniGene
Hs.172944
Gene Chromosome Location
Chr.19: 49043845-49045311 on Build GRCh38
Transcripts
NM_033043.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3692951 c.11T>A p.F4Y Chr.19: 49044220-49044220 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs182501842 Chr.19: 49044132-49044132 on Build GRCh38
rs533841470 Chr.19: 49044206-49044206 on Build GRCh38
rs552368146 Chr.19: 49044099-49044099 on Build GRCh38
rs1044936049 Chr.19: 49044053-49044053 on Build GRCh38
rs560080839 Chr.19: 49044312-49044312 on Build GRCh38
rs192602502 Chr.19: 49043919-49043919 on Build GRCh38
rs776595826 Chr.19: 49044158-49044158 on Build GRCh38
rs146912876 Chr.19: 49044220-49044220 on Build GRCh38
rs201518833 Chr.19: 49043945-49043945 on Build GRCh38
rs577086059 Chr.19: 49044014-49044014 on Build GRCh38
rs545869227 Chr.19: 49044317-49044317 on Build GRCh38
rs757085288 Chr.19: 49044182-49044182 on Build GRCh38
rs370110240 Chr.19: 49044154-49044154 on Build GRCh38
rs1058651 Chr.19: 49044104-49044104 on Build GRCh38
rs779903729 Chr.19: 49044163-49044164 on Build GRCh38
rs376431435 Chr.19: 49044269-49044269 on Build GRCh38
rs924699760 Chr.19: 49044147-49044147 on Build GRCh38
rs1007172347 Chr.19: 49044279-49044279 on Build GRCh38
rs1031808851 Chr.19: 49044309-49044309 on Build GRCh38
rs1054423848 Chr.19: 49044046-49044046 on Build GRCh38
rs748249167 Chr.19: 49044191-49044191 on Build GRCh38
rs541840801 Chr.19: 49044296-49044296 on Build GRCh38
rs771752650 Chr.19: 49044240-49044240 on Build GRCh38
rs62126046 Chr.19: 49043982-49043982 on Build GRCh38
rs375590697 Chr.19: 49044153-49044153 on Build GRCh38
rs564747089 Chr.19: 49044318-49044318 on Build GRCh38
rs1019194187 Chr.19: 49044288-49044288 on Build GRCh38
rs767891521 Chr.19: 49044101-49044101 on Build GRCh38
rs779282078 Chr.19: 49044237-49044237 on Build GRCh38
rs558759229 Chr.19: 49044001-49044001 on Build GRCh38
rs12610392 Chr.19: 49044189-49044189 on Build GRCh38
rs1045034347 Chr.19: 49044192-49044192 on Build GRCh38
rs576942503 Chr.19: 49044215-49044215 on Build GRCh38
rs554559103 Chr.19: 49044173-49044173 on Build GRCh38
rs915888377 Chr.19: 49044169-49044169 on Build GRCh38
rs375396542 Chr.19: 49043929-49043929 on Build GRCh38
rs536262481 Chr.19: 49044161-49044161 on Build GRCh38
rs948708295 Chr.19: 49044185-49044185 on Build GRCh38
rs1014661167 Chr.19: 49043972-49043972 on Build GRCh38
rs1054876301 Chr.19: 49044159-49044159 on Build GRCh38
rs893721021 Chr.19: 49044047-49044047 on Build GRCh38
rs556192631 Chr.19: 49044235-49044235 on Build GRCh38
rs1041812814 Chr.19: 49044016-49044016 on Build GRCh38
rs374966503 Chr.19: 49044239-49044239 on Build GRCh38
rs986376306 Chr.19: 49044113-49044113 on Build GRCh38
rs1058657 Chr.19: 49044121-49044121 on Build GRCh38
rs142628262 Chr.19: 49044208-49044208 on Build GRCh38
rs903215703 Chr.19: 49043930-49043930 on Build GRCh38
rs944758839 Chr.19: 49043907-49043907 on Build GRCh38
rs568998749 Chr.19: 49043966-49043966 on Build GRCh38
rs755636942 Chr.19: 49044162-49044162 on Build GRCh38
rs755168209 Chr.19: 49044226-49044226 on Build GRCh38
rs868763019 Chr.19: 49044209-49044209 on Build GRCh38
rs745495736 Chr.19: 49044074-49044074 on Build GRCh38
rs570939829 Chr.19: 49044112-49044112 on Build GRCh38
rs749776100 Chr.19: 49044234-49044234 on Build GRCh38
rs1014718309 Chr.19: 49044077-49044077 on Build GRCh38
rs750188047 Chr.19: 49044224-49044224 on Build GRCh38
rs778486675 Chr.19: 49044200-49044200 on Build GRCh38
rs1054147651 Chr.19: 49043940-49043940 on Build GRCh38
rs369920988 Chr.19: 49044183-49044183 on Build GRCh38
rs866155360 Chr.19: 49044030-49044030 on Build GRCh38
rs201199416 Chr.19: 49044175-49044175 on Build GRCh38
rs1023440880 Chr.19: 49043958-49043958 on Build GRCh38
rs544165307 Chr.19: 49044032-49044032 on Build GRCh38
rs944858579 Chr.19: 49044015-49044015 on Build GRCh38
rs33914343 Chr.19: 49043882-49043882 on Build GRCh38
rs538961984 Chr.19: 49043860-49043860 on Build GRCh38
rs775164727 Chr.19: 49044241-49044241 on Build GRCh38
rs973083093 Chr.19: 49043984-49043984 on Build GRCh38
rs994914418 Chr.19: 49044093-49044093 on Build GRCh38
rs527749182 Chr.19: 49044081-49044081 on Build GRCh38
rs527814089 Chr.19: 49044315-49044315 on Build GRCh38
rs754924374 Chr.19: 49044221-49044222 on Build GRCh38
rs540432391 Chr.19: 49043993-49043993 on Build GRCh38
rs912132360 Chr.19: 49044118-49044118 on Build GRCh38
rs1003273379 Chr.19: 49043967-49043967 on Build GRCh38
rs573803893 Chr.19: 49044050-49044050 on Build GRCh38
rs372528003 Chr.19: 49044152-49044152 on Build GRCh38
rs867669539 Chr.19: 49044025-49044025 on Build GRCh38
rs780565947 Chr.19: 49044066-49044066 on Build GRCh38
rs920882951 Chr.19: 49043992-49043992 on Build GRCh38
rs986711960 Chr.19: 49044002-49044002 on Build GRCh38
rs760156777 Chr.19: 49044250-49044250 on Build GRCh38
rs966226485 Chr.19: 49044298-49044298 on Build GRCh38
rs758607531 Chr.19: 49044151-49044151 on Build GRCh38
rs749218854 Chr.19: 49044195-49044195 on Build GRCh38
rs778398694 Chr.19: 49044233-49044233 on Build GRCh38
rs375003905 Chr.19: 49044168-49044168 on Build GRCh38
rs768106210 Chr.19: 49044264-49044264 on Build GRCh38
rs890542657 Chr.19: 49043871-49043871 on Build GRCh38
rs961732642 Chr.19: 49043979-49043979 on Build GRCh38
rs765040760 Chr.19: 49044222-49044222 on Build GRCh38
rs560213421 Chr.19: 49044072-49044072 on Build GRCh38
rs201347231 Chr.19: 49043886-49043886 on Build GRCh38
rs755990336 Chr.19: 49044176-49044176 on Build GRCh38
rs764079257 Chr.19: 49044178-49044178 on Build GRCh38
rs906390605 Chr.19: 49044062-49044062 on Build GRCh38
rs371475564 Chr.19: 49044219-49044219 on Build GRCh38
rs369314198 Chr.19: 49043942-49043942 on Build GRCh38
rs756297627 Chr.19: 49044231-49044231 on Build GRCh38
rs566597050 Chr.19: 49044188-49044188 on Build GRCh38
rs866898732 Chr.19: 49044134-49044134 on Build GRCh38
rs541779442 Chr.19: 49044067-49044067 on Build GRCh38
rs749967837 Chr.19: 49044160-49044160 on Build GRCh38
rs536400087 Chr.19: 49043870-49043870 on Build GRCh38
rs1027288370 Chr.19: 49044093-49044093 on Build GRCh38
rs753627879 Chr.19: 49044180-49044180 on Build GRCh38
rs770408103 Chr.19: 49044194-49044194 on Build GRCh38
rs953772033 Chr.19: 49044098-49044098 on Build GRCh38
rs763467014 Chr.19: 49044165-49044165 on Build GRCh38
rs550204532 Chr.19: 49044138-49044138 on Build GRCh38
rs752817899 Chr.19: 49044229-49044229 on Build GRCh38
rs770426766 Chr.19: 49044164-49044164 on Build GRCh38
rs766122270 Chr.19: 49044167-49044167 on Build GRCh38
rs562930586 Chr.19: 49044038-49044038 on Build GRCh38
rs912031635 Chr.19: 49044010-49044010 on Build GRCh38
rs568736052 Chr.19: 49043867-49043867 on Build GRCh38
rs977565377 Chr.19: 49044307-49044307 on Build GRCh38
rs1000188968 Chr.19: 49043932-49043932 on Build GRCh38
rs555782069 Chr.19: 49044017-49044017 on Build GRCh38
rs14207 Chr.19: 49044201-49044201 on Build GRCh38

View Full Product Details