Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49364390-49364901 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DKKL1
Entrez Gene ID
27120
Gene Name
dickkopf like acrosomal protein 1
Gene Aliases
CT34, SGY, SGY-1, SGY1
UniGene
Hs.515855
Gene Chromosome Location
Chr.19: 49360520-49375132 on Build GRCh38
Transcripts
NM_001197301.1 NM_001197302.1 NM_014419.3 XM_006723142.2 XM_011526724.1 XM_011526725.1 XM_011526726.2 XM_011526727.2 XM_011526728.2 XM_011526729.1 XM_017026593.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1712586 c.73G>A p.V25M Chr.19: 49364644-49364644 on Build GRCh38
3938207 c.170G>A p.R57Q Chr.19: 49364741-49364741 on Build GRCh38
5001917 c.85G>A p.A29T Chr.19: 49364656-49364656 on Build GRCh38
6289842 c.135C>T p.L45L Chr.19: 49364706-49364706 on Build GRCh38
6398923 c.53T>C p.L18P Chr.19: 49364624-49364624 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774912440 Chr.19: 49364567-49364567 on Build GRCh38
rs752266473 Chr.19: 49364570-49364570 on Build GRCh38
rs544282844 Chr.19: 49364494-49364494 on Build GRCh38
rs766861522 Chr.19: 49364566-49364566 on Build GRCh38
rs140685187 Chr.19: 49364779-49364779 on Build GRCh38
rs1004945070 Chr.19: 49364815-49364815 on Build GRCh38
rs1042863095 Chr.19: 49364443-49364443 on Build GRCh38
rs755667514 Chr.19: 49364576-49364576 on Build GRCh38
rs750312885 Chr.19: 49364704-49364704 on Build GRCh38
rs372918287 Chr.19: 49364766-49364766 on Build GRCh38
rs533834968 Chr.19: 49364840-49364840 on Build GRCh38
rs369926852 Chr.19: 49364838-49364838 on Build GRCh38
rs768938333 Chr.19: 49364544-49364544 on Build GRCh38
rs544396001 Chr.19: 49364568-49364568 on Build GRCh38
rs562607286 Chr.19: 49364525-49364525 on Build GRCh38
rs527976440 Chr.19: 49364644-49364644 on Build GRCh38
rs761338559 Chr.19: 49364772-49364772 on Build GRCh38
rs575714787 Chr.19: 49364493-49364493 on Build GRCh38
rs116223719 Chr.19: 49364655-49364655 on Build GRCh38
rs141910577 Chr.19: 49364727-49364727 on Build GRCh38
rs763792602 Chr.19: 49364682-49364682 on Build GRCh38
rs1016393450 Chr.19: 49364582-49364582 on Build GRCh38
rs771632482 Chr.19: 49364649-49364649 on Build GRCh38
rs372117146 Chr.19: 49364663-49364663 on Build GRCh38
rs758170786 Chr.19: 49364629-49364629 on Build GRCh38
rs776519239 Chr.19: 49364766-49364767 on Build GRCh38
rs974991699 Chr.19: 49364592-49364592 on Build GRCh38
rs764688429 Chr.19: 49364774-49364774 on Build GRCh38
rs754770671 Chr.19: 49364798-49364798 on Build GRCh38
rs758245650 Chr.19: 49364710-49364710 on Build GRCh38
rs968861594 Chr.19: 49364419-49364419 on Build GRCh38
rs774693540 Chr.19: 49364697-49364698 on Build GRCh38
rs772487863 Chr.19: 49364760-49364760 on Build GRCh38
rs190836480 Chr.19: 49364828-49364828 on Build GRCh38
rs761285343 Chr.19: 49364694-49364694 on Build GRCh38
rs764458646 Chr.19: 49364803-49364803 on Build GRCh38
rs919364 Chr.19: 49364656-49364656 on Build GRCh38
rs763470169 Chr.19: 49364578-49364578 on Build GRCh38
rs199831575 Chr.19: 49364740-49364740 on Build GRCh38
rs748281161 Chr.19: 49364636-49364636 on Build GRCh38
rs368445433 Chr.19: 49364531-49364531 on Build GRCh38
rs765211326 Chr.19: 49364701-49364701 on Build GRCh38
rs775236738 Chr.19: 49364667-49364667 on Build GRCh38
rs779649917 Chr.19: 49364715-49364715 on Build GRCh38
rs913787134 Chr.19: 49364842-49364842 on Build GRCh38
rs1025860764 Chr.19: 49364716-49364716 on Build GRCh38
rs750262253 Chr.19: 49364626-49364626 on Build GRCh38
rs778820244 Chr.19: 49364606-49364606 on Build GRCh38
rs376073063 Chr.19: 49364490-49364490 on Build GRCh38
rs753776234 Chr.19: 49364585-49364585 on Build GRCh38
rs79220639 Chr.19: 49364478-49364478 on Build GRCh38
rs375995405 Chr.19: 49364689-49364689 on Build GRCh38
rs781499893 Chr.19: 49364632-49364632 on Build GRCh38
rs766326095 Chr.19: 49364792-49364792 on Build GRCh38
rs763487900 Chr.19: 49364564-49364564 on Build GRCh38
rs756347586 Chr.19: 49364731-49364731 on Build GRCh38
rs773801801 Chr.19: 49364558-49364558 on Build GRCh38
rs939293013 Chr.19: 49364594-49364594 on Build GRCh38
rs945905915 Chr.19: 49364432-49364432 on Build GRCh38
rs891837616 Chr.19: 49364613-49364613 on Build GRCh38
rs1018884184 Chr.19: 49364688-49364688 on Build GRCh38
rs776690008 Chr.19: 49364549-49364549 on Build GRCh38
rs1052296313 Chr.19: 49364598-49364598 on Build GRCh38
rs927801047 Chr.19: 49364587-49364587 on Build GRCh38
rs749317800 Chr.19: 49364588-49364589 on Build GRCh38
rs769258136 Chr.19: 49364631-49364632 on Build GRCh38
rs760175066 Chr.19: 49364671-49364671 on Build GRCh38
rs757246751 Chr.19: 49364593-49364593 on Build GRCh38
rs748309077 Chr.19: 49364553-49364553 on Build GRCh38
rs147276980 Chr.19: 49364777-49364777 on Build GRCh38
rs988022861 Chr.19: 49364839-49364839 on Build GRCh38
rs372470212 Chr.19: 49364554-49364554 on Build GRCh38
rs1007436543 Chr.19: 49364470-49364470 on Build GRCh38
rs2303757 Chr.19: 49364642-49364642 on Build GRCh38
rs778135086 Chr.19: 49364739-49364739 on Build GRCh38
rs753213646 Chr.19: 49364770-49364770 on Build GRCh38
rs771111490 Chr.19: 49364741-49364741 on Build GRCh38
rs746332148 Chr.19: 49364755-49364755 on Build GRCh38
rs868233204 Chr.19: 49364467-49364467 on Build GRCh38
rs1013523601 Chr.19: 49364501-49364501 on Build GRCh38
rs868020018 Chr.19: 49364428-49364428 on Build GRCh38
rs779581383 Chr.19: 49364754-49364754 on Build GRCh38
rs918287227 Chr.19: 49364516-49364516 on Build GRCh38
rs745652625 Chr.19: 49364532-49364532 on Build GRCh38
rs576586238 Chr.19: 49364761-49364761 on Build GRCh38
rs751374702 Chr.19: 49364797-49364797 on Build GRCh38
rs375743381 Chr.19: 49364767-49364767 on Build GRCh38

View Full Product Details