Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49406833-49407340 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC155
Entrez Gene ID
147872
Gene Name
coiled-coil domain containing 155
Gene Aliases
KASH5
UniGene
Hs.662085
Gene Chromosome Location
Chr.19: 49388218-49418002 on Build GRCh38
Transcripts
NM_144688.4 XM_005258549.3 XM_011526488.2 XM_011526489.2 XM_011526490.2 XM_011526494.2 XM_011526495.2 XM_011526496.2 XM_011526497.2 XM_017026333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2752492 c.821G>A p.R274Q Chr.19: 49406908-49406908 on Build GRCh38
6898304 c.916G>A p.D306N Chr.19: 49407279-49407279 on Build GRCh38
5775488 c.846G>C p.Q282H Chr.19: 49406933-49406933 on Build GRCh38
6292924 c.885C>T p.L295L Chr.19: 49407248-49407248 on Build GRCh38
1325136 c.823G>A p.D275N Chr.19: 49406910-49406910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773234684 Chr.19: 49406908-49406908 on Build GRCh38
rs1033373118 Chr.19: 49407125-49407125 on Build GRCh38
rs367605166 Chr.19: 49407222-49407222 on Build GRCh38
rs772644534 Chr.19: 49406926-49406926 on Build GRCh38
rs772501452 Chr.19: 49406885-49406885 on Build GRCh38
rs759541524 Chr.19: 49407263-49407263 on Build GRCh38
rs750125339 Chr.19: 49407026-49407026 on Build GRCh38
rs751148932 Chr.19: 49407238-49407238 on Build GRCh38
rs61732834 Chr.19: 49406888-49406888 on Build GRCh38
rs962669983 Chr.19: 49407129-49407129 on Build GRCh38
rs1055037357 Chr.19: 49406861-49406861 on Build GRCh38
rs117030842 Chr.19: 49406862-49406862 on Build GRCh38
rs777175342 Chr.19: 49407221-49407221 on Build GRCh38
rs761129244 Chr.19: 49406907-49406907 on Build GRCh38
rs566432841 Chr.19: 49407149-49407149 on Build GRCh38
rs757894941 Chr.19: 49407145-49407145 on Build GRCh38
rs747407268 Chr.19: 49406897-49406897 on Build GRCh38
rs750593202 Chr.19: 49407297-49407297 on Build GRCh38
rs779176054 Chr.19: 49406997-49406997 on Build GRCh38
rs200360301 Chr.19: 49407308-49407308 on Build GRCh38
rs202070364 Chr.19: 49406922-49406922 on Build GRCh38
rs933058578 Chr.19: 49407254-49407254 on Build GRCh38
rs766162301 Chr.19: 49406876-49406878 on Build GRCh38
rs374609604 Chr.19: 49407285-49407285 on Build GRCh38
rs200851261 Chr.19: 49407287-49407287 on Build GRCh38
rs57234750 Chr.19: 49407260-49407260 on Build GRCh38
rs547981952 Chr.19: 49407112-49407112 on Build GRCh38
rs769352429 Chr.19: 49407223-49407223 on Build GRCh38
rs752539755 Chr.19: 49407265-49407265 on Build GRCh38
rs900380872 Chr.19: 49407025-49407025 on Build GRCh38
rs764515946 Chr.19: 49406932-49406932 on Build GRCh38
rs147642514 Chr.19: 49407081-49407081 on Build GRCh38
rs527307252 Chr.19: 49407154-49407154 on Build GRCh38
rs751428531 Chr.19: 49406964-49406964 on Build GRCh38
rs752197722 Chr.19: 49407138-49407138 on Build GRCh38
rs1005217415 Chr.19: 49407272-49407272 on Build GRCh38
rs111577017 Chr.19: 49407216-49407216 on Build GRCh38
rs572388108 Chr.19: 49407319-49407319 on Build GRCh38
rs1028643506 Chr.19: 49407001-49407001 on Build GRCh38
rs370996029 Chr.19: 49407264-49407264 on Build GRCh38
rs749458669 Chr.19: 49406873-49406873 on Build GRCh38
rs531766681 Chr.19: 49406909-49406909 on Build GRCh38
rs959552349 Chr.19: 49407144-49407144 on Build GRCh38
rs757514112 Chr.19: 49406992-49406992 on Build GRCh38
rs570790266 Chr.19: 49407171-49407171 on Build GRCh38
rs112074780 Chr.19: 49406882-49406882 on Build GRCh38
rs755590313 Chr.19: 49407309-49407309 on Build GRCh38
rs752488483 Chr.19: 49406986-49406986 on Build GRCh38
rs377455499 Chr.19: 49407303-49407303 on Build GRCh38
rs886236285 Chr.19: 49407268-49407268 on Build GRCh38
rs369333334 Chr.19: 49407008-49407008 on Build GRCh38
rs369003961 Chr.19: 49406903-49406903 on Build GRCh38
rs774527096 Chr.19: 49406869-49406869 on Build GRCh38
rs764911357 Chr.19: 49406918-49406918 on Build GRCh38
rs899405531 Chr.19: 49406933-49406933 on Build GRCh38
rs754777243 Chr.19: 49406973-49406973 on Build GRCh38
rs886371195 Chr.19: 49407102-49407102 on Build GRCh38
rs1026079928 Chr.19: 49407162-49407171 on Build GRCh38
rs773833096 Chr.19: 49407241-49407241 on Build GRCh38
rs1030279534 Chr.19: 49407073-49407073 on Build GRCh38
rs780430941 Chr.19: 49407199-49407199 on Build GRCh38
rs758175295 Chr.19: 49407065-49407065 on Build GRCh38
rs376011371 Chr.19: 49407005-49407005 on Build GRCh38
rs778923068 Chr.19: 49406879-49406879 on Build GRCh38
rs748551898 Chr.19: 49407318-49407318 on Build GRCh38
rs568170851 Chr.19: 49407240-49407240 on Build GRCh38
rs370261651 Chr.19: 49407305-49407305 on Build GRCh38
rs926786809 Chr.19: 49406889-49406889 on Build GRCh38
rs562232016 Chr.19: 49406940-49406940 on Build GRCh38
rs762371553 Chr.19: 49407233-49407233 on Build GRCh38
rs529750728 Chr.19: 49407061-49407061 on Build GRCh38
rs1016014867 Chr.19: 49407126-49407126 on Build GRCh38
rs781745478 Chr.19: 49407311-49407311 on Build GRCh38
rs372992146 Chr.19: 49406939-49406939 on Build GRCh38
rs767361842 Chr.19: 49406979-49406979 on Build GRCh38
rs552486049 Chr.19: 49407167-49407167 on Build GRCh38
rs974189082 Chr.19: 49407134-49407134 on Build GRCh38

View Full Product Details