Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49407122-49407624 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC155
Entrez Gene ID
147872
Gene Name
coiled-coil domain containing 155
Gene Aliases
KASH5
UniGene
Hs.662085
Gene Chromosome Location
Chr.19: 49388218-49418002 on Build GRCh38
Transcripts
NM_144688.4 XM_005258549.3 XM_011526488.2 XM_011526489.2 XM_011526490.2 XM_011526494.2 XM_011526495.2 XM_011526496.2 XM_011526497.2 XM_017026333.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4285304 c.934-5C>T p.? Chr.19: 49407607-49407607 on Build GRCh38
6898304 c.916G>A p.D306N Chr.19: 49407279-49407279 on Build GRCh38
1395229 c.934C>T p.R312C Chr.19: 49407612-49407612 on Build GRCh38
6621747 c.941G>A p.R314H Chr.19: 49407619-49407619 on Build GRCh38
6292924 c.885C>T p.L295L Chr.19: 49407248-49407248 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754387518 Chr.19: 49407432-49407432 on Build GRCh38
rs756922082 Chr.19: 49407350-49407350 on Build GRCh38
rs973871018 Chr.19: 49407494-49407494 on Build GRCh38
rs367605166 Chr.19: 49407222-49407222 on Build GRCh38
rs778263346 Chr.19: 49407346-49407346 on Build GRCh38
rs867596953 Chr.19: 49407398-49407398 on Build GRCh38
rs397810241 Chr.19: 49407448-49407449 on Build GRCh38
rs759541524 Chr.19: 49407263-49407263 on Build GRCh38
rs940765647 Chr.19: 49407481-49407481 on Build GRCh38
rs751148932 Chr.19: 49407238-49407238 on Build GRCh38
rs751710516 Chr.19: 49407336-49407336 on Build GRCh38
rs759028552 Chr.19: 49407328-49407328 on Build GRCh38
rs531597186 Chr.19: 49407540-49407540 on Build GRCh38
rs560026795 Chr.19: 49407448-49407448 on Build GRCh38
rs777175342 Chr.19: 49407221-49407221 on Build GRCh38
rs755067324 Chr.19: 49407337-49407337 on Build GRCh38
rs753186940 Chr.19: 49407338-49407338 on Build GRCh38
rs541872252 Chr.19: 49407445-49407445 on Build GRCh38
rs566432841 Chr.19: 49407149-49407149 on Build GRCh38
rs570753153 Chr.19: 49407538-49407538 on Build GRCh38
rs757894941 Chr.19: 49407145-49407145 on Build GRCh38
rs750593202 Chr.19: 49407297-49407297 on Build GRCh38
rs763892064 Chr.19: 49407334-49407334 on Build GRCh38
rs552252270 Chr.19: 49407475-49407475 on Build GRCh38
rs958502615 Chr.19: 49407344-49407344 on Build GRCh38
rs760393501 Chr.19: 49407333-49407333 on Build GRCh38
rs527259950 Chr.19: 49407455-49407455 on Build GRCh38
rs200360301 Chr.19: 49407308-49407308 on Build GRCh38
rs370226167 Chr.19: 49407339-49407339 on Build GRCh38
rs933058578 Chr.19: 49407254-49407254 on Build GRCh38
rs374609604 Chr.19: 49407285-49407285 on Build GRCh38
rs182108044 Chr.19: 49407439-49407439 on Build GRCh38
rs200851261 Chr.19: 49407287-49407287 on Build GRCh38
rs57234750 Chr.19: 49407260-49407260 on Build GRCh38
rs562259101 Chr.19: 49407412-49407412 on Build GRCh38
rs371220692 Chr.19: 49407478-49407478 on Build GRCh38
rs769352429 Chr.19: 49407223-49407223 on Build GRCh38
rs752539755 Chr.19: 49407265-49407265 on Build GRCh38
rs201989078 Chr.19: 49407325-49407325 on Build GRCh38
rs754025481 Chr.19: 49407573-49407574 on Build GRCh38
rs191738825 Chr.19: 49407326-49407326 on Build GRCh38
rs527307252 Chr.19: 49407154-49407154 on Build GRCh38
rs772021543 Chr.19: 49407330-49407330 on Build GRCh38
rs974396185 Chr.19: 49407488-49407488 on Build GRCh38
rs1005217415 Chr.19: 49407272-49407272 on Build GRCh38
rs111577017 Chr.19: 49407216-49407216 on Build GRCh38
rs778491293 Chr.19: 49407601-49407601 on Build GRCh38
rs572388108 Chr.19: 49407319-49407319 on Build GRCh38
rs370996029 Chr.19: 49407264-49407264 on Build GRCh38
rs1028115614 Chr.19: 49407513-49407513 on Build GRCh38
rs570790266 Chr.19: 49407171-49407171 on Build GRCh38
rs568219666 Chr.19: 49407602-49407602 on Build GRCh38
rs534983676 Chr.19: 49407551-49407551 on Build GRCh38
rs749588732 Chr.19: 49407594-49407594 on Build GRCh38
rs888271698 Chr.19: 49407526-49407526 on Build GRCh38
rs1051492284 Chr.19: 49407578-49407578 on Build GRCh38
rs1005329578 Chr.19: 49407539-49407539 on Build GRCh38
rs755590313 Chr.19: 49407309-49407309 on Build GRCh38
rs576368270 Chr.19: 49407385-49407385 on Build GRCh38
rs377455499 Chr.19: 49407303-49407303 on Build GRCh38
rs886236285 Chr.19: 49407268-49407268 on Build GRCh38
rs543712878 Chr.19: 49407409-49407409 on Build GRCh38
rs921580578 Chr.19: 49407535-49407535 on Build GRCh38
rs753561234 Chr.19: 49407335-49407335 on Build GRCh38
rs1026079928 Chr.19: 49407162-49407171 on Build GRCh38
rs773833096 Chr.19: 49407241-49407241 on Build GRCh38
rs780430941 Chr.19: 49407199-49407199 on Build GRCh38
rs936367246 Chr.19: 49407479-49407479 on Build GRCh38
rs748551898 Chr.19: 49407318-49407318 on Build GRCh38
rs775369276 Chr.19: 49407332-49407332 on Build GRCh38
rs568170851 Chr.19: 49407240-49407240 on Build GRCh38
rs370261651 Chr.19: 49407305-49407305 on Build GRCh38
rs770669500 Chr.19: 49407600-49407600 on Build GRCh38
rs762371553 Chr.19: 49407233-49407233 on Build GRCh38
rs982125834 Chr.19: 49407450-49407450 on Build GRCh38
rs991372927 Chr.19: 49407380-49407380 on Build GRCh38
rs144457660 Chr.19: 49407418-49407418 on Build GRCh38
rs773894384 Chr.19: 49407327-49407327 on Build GRCh38
rs781745478 Chr.19: 49407311-49407311 on Build GRCh38
rs552486049 Chr.19: 49407167-49407167 on Build GRCh38
rs145306940 Chr.19: 49407444-49407445 on Build GRCh38

View Full Product Details