Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49474284-49474794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT3LG
Entrez Gene ID
2323
Gene Name
fms related tyrosine kinase 3 ligand
Gene Aliases
FL, FLT3L
UniGene
Hs.428
Gene Chromosome Location
Chr.19: 49474172-49487038 on Build GRCh38
Transcripts
NM_001204502.1 NM_001204503.1 NM_001278637.1 NM_001278638.1 NM_001459.3 XM_005258680.4 XM_005258681.4 XM_005258682.4 XM_005258683.4 XM_006723116.3 XM_011526675.2 XM_011526676.2 XM_011526677.2 XM_011526678.2 XM_011526680.2 XM_011526682.2 XM_017026532.1 XM_017026533.1 XM_017026534.1 XM_017026535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7368471 c.20C>A p.A7D Chr.19: 49474659-49474659 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs748399723 Chr.19: 49474653-49474653 on Build GRCh38
rs993554394 Chr.19: 49474493-49474493 on Build GRCh38
rs773713350 Chr.19: 49474682-49474682 on Build GRCh38
rs914021131 Chr.19: 49474604-49474604 on Build GRCh38
rs1032888118 Chr.19: 49474748-49474748 on Build GRCh38
rs772086727 Chr.19: 49474639-49474639 on Build GRCh38
rs984662005 Chr.19: 49474527-49474527 on Build GRCh38
rs922003721 Chr.19: 49474486-49474486 on Build GRCh38
rs75750998 Chr.19: 49474672-49474672 on Build GRCh38
rs566709354 Chr.19: 49474712-49474712 on Build GRCh38
rs552507865 Chr.19: 49474661-49474661 on Build GRCh38
rs907883240 Chr.19: 49474529-49474529 on Build GRCh38
rs1005983932 Chr.19: 49474742-49474742 on Build GRCh38
rs752419761 Chr.19: 49474627-49474627 on Build GRCh38
rs964507015 Chr.19: 49474535-49474535 on Build GRCh38
rs759035341 Chr.19: 49474636-49474636 on Build GRCh38
rs562390498 Chr.19: 49474520-49474520 on Build GRCh38
rs975085009 Chr.19: 49474536-49474536 on Build GRCh38
rs963847808 Chr.19: 49474471-49474471 on Build GRCh38
rs965399741 Chr.19: 49474533-49474533 on Build GRCh38
rs1014326804 Chr.19: 49474416-49474416 on Build GRCh38
rs937486189 Chr.19: 49474609-49474609 on Build GRCh38
rs183498523 Chr.19: 49474420-49474420 on Build GRCh38
rs923156872 Chr.19: 49474550-49474550 on Build GRCh38
rs895602936 Chr.19: 49474344-49474344 on Build GRCh38
rs892531290 Chr.19: 49474438-49474438 on Build GRCh38
rs967674468 Chr.19: 49474430-49474430 on Build GRCh38
rs377157097 Chr.19: 49474632-49474632 on Build GRCh38
rs762494994 Chr.19: 49474592-49474592 on Build GRCh38
rs560612184 Chr.19: 49474575-49474575 on Build GRCh38
rs548187028 Chr.19: 49474466-49474466 on Build GRCh38
rs1040655295 Chr.19: 49474364-49474364 on Build GRCh38
rs548425309 Chr.19: 49474557-49474557 on Build GRCh38
rs373840869 Chr.19: 49474637-49474637 on Build GRCh38
rs984611477 Chr.19: 49474329-49474329 on Build GRCh38
rs932220758 Chr.19: 49474544-49474544 on Build GRCh38
rs1017452368 Chr.19: 49474534-49474534 on Build GRCh38
rs538588013 Chr.19: 49474735-49474735 on Build GRCh38
rs1001246541 Chr.19: 49474319-49474319 on Build GRCh38
rs372444883 Chr.19: 49474542-49474542 on Build GRCh38
rs1043863119 Chr.19: 49474480-49474480 on Build GRCh38
rs762497507 Chr.19: 49474630-49474630 on Build GRCh38
rs16981277 Chr.19: 49474758-49474758 on Build GRCh38
rs540199440 Chr.19: 49474340-49474340 on Build GRCh38
rs767350748 Chr.19: 49474690-49474690 on Build GRCh38
rs759575286 Chr.19: 49474687-49474687 on Build GRCh38
rs772951923 Chr.19: 49474659-49474659 on Build GRCh38
rs1002460191 Chr.19: 49474485-49474485 on Build GRCh38
rs544004477 Chr.19: 49474492-49474492 on Build GRCh38
rs368120792 Chr.19: 49474593-49474593 on Build GRCh38
rs769588331 Chr.19: 49474654-49474654 on Build GRCh38
rs529828729 Chr.19: 49474524-49474524 on Build GRCh38
rs984960640 Chr.19: 49474573-49474573 on Build GRCh38
rs780567056 Chr.19: 49474643-49474643 on Build GRCh38
rs988533285 Chr.19: 49474576-49474576 on Build GRCh38
rs781087811 Chr.19: 49474617-49474617 on Build GRCh38
rs187456505 Chr.19: 49474483-49474483 on Build GRCh38
rs568707596 Chr.19: 49474751-49474751 on Build GRCh38
rs777122834 Chr.19: 49474651-49474651 on Build GRCh38
rs761730159 Chr.19: 49474698-49474699 on Build GRCh38
rs751440277 Chr.19: 49474698-49474698 on Build GRCh38
rs922193506 Chr.19: 49474539-49474539 on Build GRCh38
rs371233989 Chr.19: 49474633-49474633 on Build GRCh38
rs1019267261 Chr.19: 49474528-49474528 on Build GRCh38
rs1041746278 Chr.19: 49474670-49474670 on Build GRCh38
rs371642476 Chr.19: 49474606-49474606 on Build GRCh38
rs111741433 Chr.19: 49474737-49474737 on Build GRCh38
rs751476177 Chr.19: 49474607-49474607 on Build GRCh38
rs1032771997 Chr.19: 49474467-49474467 on Build GRCh38
rs747589247 Chr.19: 49474647-49474647 on Build GRCh38
rs922635913 Chr.19: 49474387-49474387 on Build GRCh38
rs754841194 Chr.19: 49474610-49474610 on Build GRCh38

View Full Product Details