Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49478813-49479187 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FLT3LG
Entrez Gene ID
2323
Gene Name
fms related tyrosine kinase 3 ligand
Gene Aliases
FL, FLT3L
UniGene
Hs.428
Gene Chromosome Location
Chr.19: 49474172-49487038 on Build GRCh38
Transcripts
NM_001204502.1 NM_001204503.1 NM_001278637.1 NM_001278638.1 NM_001459.3 XM_005258680.4 XM_005258681.4 XM_005258682.4 XM_005258683.4 XM_006723116.3 XM_011526675.2 XM_011526676.2 XM_011526677.2 XM_011526678.2 XM_011526680.2 XM_011526682.2 XM_017026532.1 XM_017026533.1 XM_017026534.1 XM_017026535.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6327630 c.385C>T p.R129C Chr.19: 49478951-49478951 on Build GRCh38
232354 c.401C>T p.T134I Chr.19: 49478967-49478967 on Build GRCh38
5438690 c.350C>T p.P117L Chr.19: 49478916-49478916 on Build GRCh38
1481322 c.343-1_343insC p.S118fs*>119 Chr.19: 49478908-49478909 on Build GRCh38
6085769 c.455G>T p.R152L Chr.19: 49479021-49479021 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs148441000 Chr.19: 49479004-49479004 on Build GRCh38
rs368426909 Chr.19: 49479134-49479134 on Build GRCh38
rs763704897 Chr.19: 49478911-49478911 on Build GRCh38
rs956458829 Chr.19: 49479100-49479100 on Build GRCh38
rs114380263 Chr.19: 49478863-49478863 on Build GRCh38
rs769957989 Chr.19: 49478862-49478862 on Build GRCh38
rs752351114 Chr.19: 49478910-49478910 on Build GRCh38
rs763649058 Chr.19: 49479031-49479031 on Build GRCh38
rs371655014 Chr.19: 49479159-49479159 on Build GRCh38
rs760433482 Chr.19: 49479021-49479021 on Build GRCh38
rs896713347 Chr.19: 49478855-49478855 on Build GRCh38
rs369589648 Chr.19: 49478947-49478947 on Build GRCh38
rs915094983 Chr.19: 49479151-49479151 on Build GRCh38
rs763513799 Chr.19: 49478866-49478866 on Build GRCh38
rs750427971 Chr.19: 49479053-49479053 on Build GRCh38
rs766254929 Chr.19: 49479064-49479064 on Build GRCh38
rs769482947 Chr.19: 49478908-49478909 on Build GRCh38
rs201554548 Chr.19: 49478898-49478898 on Build GRCh38
rs116114136 Chr.19: 49479127-49479127 on Build GRCh38
rs926682933 Chr.19: 49479113-49479113 on Build GRCh38
rs979987941 Chr.19: 49479094-49479094 on Build GRCh38
rs10420643 Chr.19: 49478966-49478966 on Build GRCh38
rs983372622 Chr.19: 49479006-49479006 on Build GRCh38
rs772522910 Chr.19: 49478996-49478996 on Build GRCh38
rs910896503 Chr.19: 49478853-49478853 on Build GRCh38
rs774922225 Chr.19: 49478909-49478909 on Build GRCh38
rs777979329 Chr.19: 49478960-49478960 on Build GRCh38
rs771747171 Chr.19: 49479106-49479106 on Build GRCh38
rs778930544 Chr.19: 49479092-49479092 on Build GRCh38
rs751311198 Chr.19: 49479065-49479065 on Build GRCh38
rs1018918392 Chr.19: 49479066-49479066 on Build GRCh38
rs747638047 Chr.19: 49478878-49478878 on Build GRCh38
rs765298748 Chr.19: 49479041-49479041 on Build GRCh38
rs779684637 Chr.19: 49478933-49478933 on Build GRCh38
rs373232707 Chr.19: 49479096-49479096 on Build GRCh38
rs774788539 Chr.19: 49478986-49478986 on Build GRCh38
rs775989477 Chr.19: 49479097-49479097 on Build GRCh38
rs372857705 Chr.19: 49478913-49478913 on Build GRCh38
rs753402069 Chr.19: 49479034-49479034 on Build GRCh38
rs374893974 Chr.19: 49478916-49478916 on Build GRCh38
rs540792169 Chr.19: 49479086-49479086 on Build GRCh38
rs146198329 Chr.19: 49478972-49478972 on Build GRCh38
rs538660761 Chr.19: 49478836-49478836 on Build GRCh38
rs771426751 Chr.19: 49478869-49478869 on Build GRCh38
rs758247403 Chr.19: 49479056-49479056 on Build GRCh38
rs904016135 Chr.19: 49478887-49478887 on Build GRCh38
rs374337774 Chr.19: 49478914-49478914 on Build GRCh38
rs757285563 Chr.19: 49479088-49479088 on Build GRCh38
rs142398567 Chr.19: 49479036-49479036 on Build GRCh38
rs750254491 Chr.19: 49478915-49478915 on Build GRCh38
rs767095501 Chr.19: 49478905-49478905 on Build GRCh38
rs1056415021 Chr.19: 49478846-49478846 on Build GRCh38
rs1056356644 Chr.19: 49479142-49479142 on Build GRCh38
rs749301992 Chr.19: 49478971-49478971 on Build GRCh38
rs746419008 Chr.19: 49479093-49479093 on Build GRCh38
rs759921995 Chr.19: 49478995-49478995 on Build GRCh38
rs577771720 Chr.19: 49479153-49479153 on Build GRCh38
rs1002854603 Chr.19: 49478917-49478917 on Build GRCh38
rs756367356 Chr.19: 49479069-49479069 on Build GRCh38
rs769895049 Chr.19: 49478951-49478951 on Build GRCh38
rs569231633 Chr.19: 49478893-49478893 on Build GRCh38
rs965044167 Chr.19: 49479075-49479075 on Build GRCh38
rs777803841 Chr.19: 49479080-49479080 on Build GRCh38

View Full Product Details