Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49513168-49513660 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGRT
Entrez Gene ID
2217
Gene Name
Fc fragment of IgG receptor and transporter
Gene Aliases
FCRN, alpha-chain
UniGene
Hs.111903
Gene Chromosome Location
Chr.19: 49512279-49526428 on Build GRCh38
Transcripts
NM_001136019.2 NM_004107.4 XM_005258657.1 XM_011526627.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4632231 c.55C>T p.P19S Chr.19: 49513455-49513455 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs933570468 Chr.19: 49513314-49513314 on Build GRCh38
rs915633311 Chr.19: 49513282-49513282 on Build GRCh38
rs1014602885 Chr.19: 49513429-49513429 on Build GRCh38
rs986126095 Chr.19: 49513251-49513251 on Build GRCh38
rs928394747 Chr.19: 49513539-49513539 on Build GRCh38
rs908047692 Chr.19: 49513331-49513331 on Build GRCh38
rs937095001 Chr.19: 49513574-49513574 on Build GRCh38
rs1001239971 Chr.19: 49513220-49513220 on Build GRCh38
rs898949490 Chr.19: 49513371-49513371 on Build GRCh38
rs962535908 Chr.19: 49513196-49513196 on Build GRCh38
rs59774409 Chr.19: 49513491-49513491 on Build GRCh38
rs554985144 Chr.19: 49513281-49513281 on Build GRCh38
rs936959203 Chr.19: 49513382-49513382 on Build GRCh38
rs966792187 Chr.19: 49513271-49513271 on Build GRCh38
rs895461246 Chr.19: 49513580-49513580 on Build GRCh38
rs940832086 Chr.19: 49513567-49513567 on Build GRCh38
rs973576740 Chr.19: 49513568-49513568 on Build GRCh38
rs748687030 Chr.19: 49513452-49513452 on Build GRCh38
rs925392520 Chr.19: 49513284-49513284 on Build GRCh38
rs922172950 Chr.19: 49513290-49513290 on Build GRCh38
rs908159163 Chr.19: 49513555-49513555 on Build GRCh38
rs1008101201 Chr.19: 49513618-49513618 on Build GRCh38
rs958122410 Chr.19: 49513500-49513500 on Build GRCh38
rs981598421 Chr.19: 49513538-49513538 on Build GRCh38
rs990750891 Chr.19: 49513277-49513277 on Build GRCh38
rs931226381 Chr.19: 49513584-49513584 on Build GRCh38
rs939709852 Chr.19: 49513326-49513326 on Build GRCh38
rs1014052114 Chr.19: 49513269-49513269 on Build GRCh38
rs1032809625 Chr.19: 49513612-49513612 on Build GRCh38
rs865842679 Chr.19: 49513413-49513413 on Build GRCh38
rs768835880 Chr.19: 49513398-49513398 on Build GRCh38
rs949769515 Chr.19: 49513581-49513581 on Build GRCh38
rs898444338 Chr.19: 49513343-49513343 on Build GRCh38
rs1037453411 Chr.19: 49513356-49513356 on Build GRCh38
rs953601260 Chr.19: 49513243-49513243 on Build GRCh38
rs996228501 Chr.19: 49513626-49513626 on Build GRCh38
rs551056200 Chr.19: 49513215-49513215 on Build GRCh38
rs903140188 Chr.19: 49513608-49513608 on Build GRCh38
rs966015368 Chr.19: 49513462-49513462 on Build GRCh38
rs1020557156 Chr.19: 49513270-49513270 on Build GRCh38
rs34972354 Chr.19: 49513294-49513294 on Build GRCh38
rs1036934411 Chr.19: 49513328-49513328 on Build GRCh38
rs889167833 Chr.19: 49513361-49513361 on Build GRCh38
rs62128085 Chr.19: 49513392-49513392 on Build GRCh38
rs924905490 Chr.19: 49513258-49513258 on Build GRCh38
rs1045371810 Chr.19: 49513308-49513308 on Build GRCh38
rs960238896 Chr.19: 49513257-49513257 on Build GRCh38
rs954954731 Chr.19: 49513528-49513528 on Build GRCh38
rs1028594266 Chr.19: 49513352-49513352 on Build GRCh38
rs1029425000 Chr.19: 49513508-49513508 on Build GRCh38
rs916073840 Chr.19: 49513510-49513510 on Build GRCh38
rs958027185 Chr.19: 49513285-49513285 on Build GRCh38
rs1037484734 Chr.19: 49513189-49513189 on Build GRCh38
rs987632337 Chr.19: 49513325-49513325 on Build GRCh38
rs868711387 Chr.19: 49513355-49513355 on Build GRCh38
rs1042021659 Chr.19: 49513598-49513598 on Build GRCh38
rs988110711 Chr.19: 49513529-49513529 on Build GRCh38
rs1055438298 Chr.19: 49513579-49513579 on Build GRCh38
rs1020227408 Chr.19: 49513443-49513443 on Build GRCh38
rs974747483 Chr.19: 49513287-49513287 on Build GRCh38
rs561767692 Chr.19: 49513354-49513354 on Build GRCh38
rs116498889 Chr.19: 49513195-49513195 on Build GRCh38
rs543281857 Chr.19: 49513280-49513280 on Build GRCh38
rs111981233 Chr.19: 49513222-49513222 on Build GRCh38
rs1000021938 Chr.19: 49513479-49513479 on Build GRCh38
rs920304928 Chr.19: 49513569-49513569 on Build GRCh38
rs376025216 Chr.19: 49513487-49513487 on Build GRCh38
rs977416239 Chr.19: 49513465-49513465 on Build GRCh38
rs201887538 Chr.19: 49513407-49513407 on Build GRCh38
rs939655268 Chr.19: 49513571-49513571 on Build GRCh38
rs978132538 Chr.19: 49513276-49513276 on Build GRCh38
rs779742308 Chr.19: 49513278-49513278 on Build GRCh38
rs893868054 Chr.19: 49513624-49513624 on Build GRCh38
rs142385484 Chr.19: 49513502-49513502 on Build GRCh38
rs895711984 Chr.19: 49513399-49513399 on Build GRCh38
rs1049544938 Chr.19: 49513600-49513600 on Build GRCh38
rs911883407 Chr.19: 49513255-49513255 on Build GRCh38
rs999949708 Chr.19: 49513610-49513610 on Build GRCh38
rs906862103 Chr.19: 49513311-49513311 on Build GRCh38
rs1008064323 Chr.19: 49513370-49513370 on Build GRCh38
rs995323460 Chr.19: 49513348-49513348 on Build GRCh38
rs577325226 Chr.19: 49513619-49513619 on Build GRCh38
rs1055873530 Chr.19: 49513395-49513395 on Build GRCh38

View Full Product Details