Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49689011-49689510 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM5
Entrez Gene ID
199800
Gene Name
adrenomedullin 5 (putative)
Gene Aliases
AM5, C19orf76
UniGene
Hs.311193
Gene Chromosome Location
Chr.19: 49688685-49690990 on Build GRCh38
Transcripts
NM_001101340.1

SNP Detail

SNP ID Chromosome Location
rs915836231 Chr.19: 49689179-49689179 on Build GRCh38
rs368265212 Chr.19: 49689371-49689371 on Build GRCh38
rs113467167 Chr.19: 49689244-49689244 on Build GRCh38
rs966754761 Chr.19: 49689102-49689113 on Build GRCh38
rs988571824 Chr.19: 49689453-49689453 on Build GRCh38
rs935271562 Chr.19: 49689399-49689399 on Build GRCh38
rs955910183 Chr.19: 49689443-49689443 on Build GRCh38
rs61201794 Chr.19: 49689210-49689210 on Build GRCh38
rs113947270 Chr.19: 49689422-49689422 on Build GRCh38
rs1026101969 Chr.19: 49689348-49689348 on Build GRCh38
rs982705649 Chr.19: 49689271-49689284 on Build GRCh38
rs1053615937 Chr.19: 49689400-49689400 on Build GRCh38
rs551277771 Chr.19: 49689448-49689448 on Build GRCh38
rs985220175 Chr.19: 49689322-49689322 on Build GRCh38
rs928554886 Chr.19: 49689438-49689438 on Build GRCh38
rs60730355 Chr.19: 49689209-49689209 on Build GRCh38
rs951851085 Chr.19: 49689351-49689351 on Build GRCh38
rs10425688 Chr.19: 49689383-49689383 on Build GRCh38
rs765805575 Chr.19: 49689247-49689247 on Build GRCh38
rs546155866 Chr.19: 49689365-49689365 on Build GRCh38
rs531552966 Chr.19: 49689444-49689444 on Build GRCh38
rs902035235 Chr.19: 49689330-49689330 on Build GRCh38
rs183879246 Chr.19: 49689277-49689277 on Build GRCh38
rs1023029153 Chr.19: 49689396-49689396 on Build GRCh38
rs576049441 Chr.19: 49689194-49689194 on Build GRCh38
rs984855224 Chr.19: 49689169-49689169 on Build GRCh38
rs386810161 Chr.19: 49689209-49689210 on Build GRCh38
rs981350546 Chr.19: 49689425-49689425 on Build GRCh38
rs939412207 Chr.19: 49689053-49689053 on Build GRCh38
rs993663096 Chr.19: 49689343-49689343 on Build GRCh38
rs902394403 Chr.19: 49689205-49689205 on Build GRCh38
rs976857998 Chr.19: 49689393-49689393 on Build GRCh38
rs981314082 Chr.19: 49689195-49689195 on Build GRCh38
rs951729980 Chr.19: 49689166-49689166 on Build GRCh38
rs187806688 Chr.19: 49689097-49689097 on Build GRCh38
rs533730833 Chr.19: 49689066-49689066 on Build GRCh38
rs1052397144 Chr.19: 49689057-49689057 on Build GRCh38
rs545222992 Chr.19: 49689112-49689112 on Build GRCh38
rs1035955802 Chr.19: 49689286-49689286 on Build GRCh38
rs1011644074 Chr.19: 49689240-49689240 on Build GRCh38
rs572063534 Chr.19: 49689188-49689188 on Build GRCh38
rs961966801 Chr.19: 49689300-49689300 on Build GRCh38
rs1044079746 Chr.19: 49689263-49689263 on Build GRCh38
rs906628248 Chr.19: 49689044-49689044 on Build GRCh38
rs1044491426 Chr.19: 49689467-49689467 on Build GRCh38
rs965799813 Chr.19: 49689133-49689133 on Build GRCh38
rs952844728 Chr.19: 49689313-49689313 on Build GRCh38
rs191541867 Chr.19: 49689167-49689167 on Build GRCh38
rs770993680 Chr.19: 49689188-49689190 on Build GRCh38
rs972978847 Chr.19: 49689302-49689302 on Build GRCh38
rs1008196555 Chr.19: 49689295-49689295 on Build GRCh38
rs985189205 Chr.19: 49689145-49689145 on Build GRCh38
rs559646195 Chr.19: 49689305-49689305 on Build GRCh38
rs971836230 Chr.19: 49689142-49689142 on Build GRCh38
rs528868068 Chr.19: 49689236-49689236 on Build GRCh38
rs1026050329 Chr.19: 49689156-49689156 on Build GRCh38
rs1022581545 Chr.19: 49689096-49689096 on Build GRCh38
rs145596942 Chr.19: 49689068-49689068 on Build GRCh38
rs910922170 Chr.19: 49689324-49689324 on Build GRCh38
rs1019100665 Chr.19: 49689126-49689126 on Build GRCh38
rs762610108 Chr.19: 49689269-49689273 on Build GRCh38
rs1045937707 Chr.19: 49689039-49689039 on Build GRCh38

View Full Product Details