Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49689407-49689907 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADM5
Entrez Gene ID
199800
Gene Name
adrenomedullin 5 (putative)
Gene Aliases
AM5, C19orf76
UniGene
Hs.311193
Gene Chromosome Location
Chr.19: 49688685-49690990 on Build GRCh38
Transcripts
NM_001101340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6595255 c.31C>T p.L11F Chr.19: 49689868-49689868 on Build GRCh38
2753103 c.31C>A p.L11I Chr.19: 49689868-49689868 on Build GRCh38
6595256 c.37G>T p.A13S Chr.19: 49689874-49689874 on Build GRCh38
6211924 c.4A>T p.T2S Chr.19: 49689841-49689841 on Build GRCh38
4132276 c.49C>T p.Q17* Chr.19: 49689886-49689886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778277957 Chr.19: 49689790-49689790 on Build GRCh38
rs887969361 Chr.19: 49689855-49689855 on Build GRCh38
rs753640028 Chr.19: 49689823-49689823 on Build GRCh38
rs771393198 Chr.19: 49689859-49689859 on Build GRCh38
rs763948661 Chr.19: 49689822-49689822 on Build GRCh38
rs968568767 Chr.19: 49689705-49689705 on Build GRCh38
rs913991204 Chr.19: 49689496-49689496 on Build GRCh38
rs943364789 Chr.19: 49689516-49689516 on Build GRCh38
rs746455414 Chr.19: 49689887-49689887 on Build GRCh38
rs988571824 Chr.19: 49689453-49689453 on Build GRCh38
rs955910183 Chr.19: 49689443-49689443 on Build GRCh38
rs189326953 Chr.19: 49689565-49689565 on Build GRCh38
rs1006352744 Chr.19: 49689581-49689581 on Build GRCh38
rs988624147 Chr.19: 49689685-49689685 on Build GRCh38
rs1040688420 Chr.19: 49689518-49689518 on Build GRCh38
rs781318186 Chr.19: 49689849-49689849 on Build GRCh38
rs780550954 Chr.19: 49689728-49689728 on Build GRCh38
rs551277771 Chr.19: 49689448-49689448 on Build GRCh38
rs775073277 Chr.19: 49689883-49689883 on Build GRCh38
rs910634904 Chr.19: 49689751-49689751 on Build GRCh38
rs1002488473 Chr.19: 49689618-49689618 on Build GRCh38
rs1027212362 Chr.19: 49689564-49689565 on Build GRCh38
rs764144400 Chr.19: 49689562-49689563 on Build GRCh38
rs943385044 Chr.19: 49689757-49689757 on Build GRCh38
rs928554886 Chr.19: 49689438-49689438 on Build GRCh38
rs1047885412 Chr.19: 49689568-49689568 on Build GRCh38
rs1018435920 Chr.19: 49689582-49689582 on Build GRCh38
rs531552966 Chr.19: 49689444-49689444 on Build GRCh38
rs375061308 Chr.19: 49689851-49689851 on Build GRCh38
rs887551464 Chr.19: 49689569-49689569 on Build GRCh38
rs368483666 Chr.19: 49689499-49689499 on Build GRCh38
rs543566261 Chr.19: 49689844-49689844 on Build GRCh38
rs902087557 Chr.19: 49689546-49689546 on Build GRCh38
rs547218388 Chr.19: 49689556-49689556 on Build GRCh38
rs567337039 Chr.19: 49689603-49689603 on Build GRCh38
rs76806983 Chr.19: 49689852-49689852 on Build GRCh38
rs751773007 Chr.19: 49689837-49689837 on Build GRCh38
rs989372770 Chr.19: 49689641-49689641 on Build GRCh38
rs927818891 Chr.19: 49689803-49689803 on Build GRCh38
rs62128126 Chr.19: 49689583-49689583 on Build GRCh38
rs771553653 Chr.19: 49689876-49689876 on Build GRCh38
rs1043746658 Chr.19: 49689500-49689500 on Build GRCh38
rs994451158 Chr.19: 49689522-49689522 on Build GRCh38
rs1040363645 Chr.19: 49689776-49689776 on Build GRCh38
rs201647005 Chr.19: 49689825-49689825 on Build GRCh38
rs543215123 Chr.19: 49689573-49689573 on Build GRCh38
rs774017321 Chr.19: 49689800-49689800 on Build GRCh38
rs563007451 Chr.19: 49689710-49689710 on Build GRCh38
rs771591212 Chr.19: 49689813-49689813 on Build GRCh38
rs770730925 Chr.19: 49689799-49689799 on Build GRCh38
rs749702648 Chr.19: 49689858-49689858 on Build GRCh38
rs749607522 Chr.19: 49689794-49689794 on Build GRCh38
rs1044491426 Chr.19: 49689467-49689467 on Build GRCh38
rs530886497 Chr.19: 49689575-49689575 on Build GRCh38
rs878890381 Chr.19: 49689756-49689756 on Build GRCh38
rs571846401 Chr.19: 49689734-49689734 on Build GRCh38
rs914362844 Chr.19: 49689699-49689699 on Build GRCh38
rs553252087 Chr.19: 49689678-49689678 on Build GRCh38
rs573688973 Chr.19: 49689872-49689872 on Build GRCh38
rs747874644 Chr.19: 49689745-49689745 on Build GRCh38
rs758806695 Chr.19: 49689501-49689501 on Build GRCh38
rs181948696 Chr.19: 49689622-49689622 on Build GRCh38
rs753210864 Chr.19: 49689850-49689850 on Build GRCh38
rs75083324 Chr.19: 49689590-49689590 on Build GRCh38
rs372067417 Chr.19: 49689814-49689814 on Build GRCh38
rs372201227 Chr.19: 49689875-49689875 on Build GRCh38
rs897137404 Chr.19: 49689511-49689511 on Build GRCh38
rs539264345 Chr.19: 49689808-49689808 on Build GRCh38

View Full Product Details