Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49706145-49706646 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPT1C
Entrez Gene ID
126129
Gene Name
carnitine palmitoyltransferase 1C
Gene Aliases
CATL1, CPT1-B, CPT1P, CPTI-B, CPTIC, SPG73
UniGene
Hs.112195
Gene Chromosome Location
Chr.19: 49690672-49713731 on Build GRCh38
Transcripts
NM_001136052.2 NM_001199752.2 NM_001199753.1 NM_152359.2 NR_108072.1 XM_005258505.2 XM_005258506.4 XM_006723009.3 XM_011526438.2 XM_011526439.2 XM_011526440.2 XM_017026265.1 XM_017026266.1 XM_017026267.1 XM_017026268.1 XM_017026269.1 XM_017026270.1 XM_017026271.1 XM_017026272.1 XM_017026273.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5594450 c.1198C>T p.Q400* Chr.19: 49706268-49706268 on Build GRCh38
3707297 c.1171G>A p.A391T Chr.19: 49706241-49706241 on Build GRCh38
5596533 c.1285C>T p.P429S Chr.19: 49706355-49706355 on Build GRCh38
5390323 c.1272C>T p.L424L Chr.19: 49706342-49706342 on Build GRCh38
4613498 c.1228delG p.A411fs*46 Chr.19: 49706298-49706298 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755372137 Chr.19: 49706421-49706421 on Build GRCh38
rs770799285 Chr.19: 49706336-49706336 on Build GRCh38
rs1040650134 Chr.19: 49706419-49706419 on Build GRCh38
rs562832000 Chr.19: 49706434-49706434 on Build GRCh38
rs927675700 Chr.19: 49706169-49706169 on Build GRCh38
rs570940671 Chr.19: 49706547-49706547 on Build GRCh38
rs116159823 Chr.19: 49706476-49706476 on Build GRCh38
rs370573255 Chr.19: 49706218-49706218 on Build GRCh38
rs1005527341 Chr.19: 49706467-49706467 on Build GRCh38
rs985822472 Chr.19: 49706397-49706397 on Build GRCh38
rs750400954 Chr.19: 49706563-49706563 on Build GRCh38
rs746293416 Chr.19: 49706189-49706189 on Build GRCh38
rs1047103188 Chr.19: 49706454-49706454 on Build GRCh38
rs776700182 Chr.19: 49706362-49706362 on Build GRCh38
rs143137118 Chr.19: 49706608-49706608 on Build GRCh38
rs371844232 Chr.19: 49706366-49706366 on Build GRCh38
rs749232107 Chr.19: 49706335-49706335 on Build GRCh38
rs1028204630 Chr.19: 49706555-49706555 on Build GRCh38
rs769810756 Chr.19: 49706236-49706236 on Build GRCh38
rs960873215 Chr.19: 49706485-49706485 on Build GRCh38
rs988311925 Chr.19: 49706514-49706514 on Build GRCh38
rs751926613 Chr.19: 49706303-49706303 on Build GRCh38
rs913623562 Chr.19: 49706526-49706526 on Build GRCh38
rs906518710 Chr.19: 49706491-49706491 on Build GRCh38
rs145807606 Chr.19: 49706237-49706237 on Build GRCh38
rs780960666 Chr.19: 49706428-49706428 on Build GRCh38
rs866635540 Chr.19: 49706398-49706398 on Build GRCh38
rs368217937 Chr.19: 49706220-49706220 on Build GRCh38
rs780678339 Chr.19: 49706320-49706320 on Build GRCh38
rs552742234 Chr.19: 49706506-49706506 on Build GRCh38
rs150328622 Chr.19: 49706363-49706363 on Build GRCh38
rs969558222 Chr.19: 49706349-49706349 on Build GRCh38
rs943323421 Chr.19: 49706577-49706577 on Build GRCh38
rs12980899 Chr.19: 49706482-49706482 on Build GRCh38
rs547155396 Chr.19: 49706177-49706177 on Build GRCh38
rs980964959 Chr.19: 49706351-49706351 on Build GRCh38
rs777837083 Chr.19: 49706334-49706334 on Build GRCh38
rs951325541 Chr.19: 49706304-49706304 on Build GRCh38
rs1027447826 Chr.19: 49706379-49706379 on Build GRCh38
rs1018765350 Chr.19: 49706266-49706266 on Build GRCh38
rs747744144 Chr.19: 49706324-49706324 on Build GRCh38
rs138872867 Chr.19: 49706378-49706378 on Build GRCh38
rs967755496 Chr.19: 49706532-49706532 on Build GRCh38
rs998684440 Chr.19: 49706453-49706453 on Build GRCh38
rs764179585 Chr.19: 49706275-49706275 on Build GRCh38
rs780577623 Chr.19: 49706301-49706301 on Build GRCh38
rs943269432 Chr.19: 49706413-49706413 on Build GRCh38
rs141256712 Chr.19: 49706352-49706352 on Build GRCh38
rs758822585 Chr.19: 49706190-49706190 on Build GRCh38
rs548432960 Chr.19: 49706472-49706472 on Build GRCh38
rs371325260 Chr.19: 49706228-49706228 on Build GRCh38
rs935872883 Chr.19: 49706461-49706461 on Build GRCh38
rs12978911 Chr.19: 49706396-49706396 on Build GRCh38
rs549330482 Chr.19: 49706621-49706621 on Build GRCh38
rs898071604 Chr.19: 49706540-49706540 on Build GRCh38
rs939103812 Chr.19: 49706171-49706171 on Build GRCh38
rs979088760 Chr.19: 49706537-49706537 on Build GRCh38
rs147455783 Chr.19: 49706618-49706618 on Build GRCh38
rs1053903749 Chr.19: 49706245-49706245 on Build GRCh38
rs752011825 Chr.19: 49706401-49706401 on Build GRCh38
rs910215951 Chr.19: 49706558-49706558 on Build GRCh38
rs770711301 Chr.19: 49706247-49706247 on Build GRCh38
rs774159669 Chr.19: 49706340-49706340 on Build GRCh38
rs374293545 Chr.19: 49706219-49706219 on Build GRCh38
rs368513098 Chr.19: 49706356-49706356 on Build GRCh38
rs778795927 Chr.19: 49706462-49706462 on Build GRCh38
rs61748956 Chr.19: 49706290-49706290 on Build GRCh38
rs760907133 Chr.19: 49706269-49706269 on Build GRCh38
rs754710743 Chr.19: 49706306-49706306 on Build GRCh38
rs375722992 Chr.19: 49706456-49706456 on Build GRCh38
rs376287276 Chr.19: 49706272-49706272 on Build GRCh38
rs892129620 Chr.19: 49706179-49706179 on Build GRCh38
rs774069280 Chr.19: 49706250-49706250 on Build GRCh38
rs969654739 Chr.19: 49706474-49706474 on Build GRCh38
rs758907193 Chr.19: 49706291-49706291 on Build GRCh38
rs201085193 Chr.19: 49706196-49706196 on Build GRCh38
rs181596690 Chr.19: 49706285-49706285 on Build GRCh38
rs960780559 Chr.19: 49706354-49706354 on Build GRCh38
rs769178605 Chr.19: 49706333-49706333 on Build GRCh38
rs777164545 Chr.19: 49706449-49706449 on Build GRCh38
rs761772845 Chr.19: 49706279-49706279 on Build GRCh38
rs376625740 Chr.19: 49706186-49706186 on Build GRCh38
rs572265604 Chr.19: 49706276-49706276 on Build GRCh38

View Full Product Details