Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49812917-49813425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FUZ
Entrez Gene ID
80199
Gene Name
fuzzy planar cell polarity protein
Gene Aliases
FY, NTD
UniGene
Hs.288800
Gene Chromosome Location
Chr.19: 49806866-49817376 on Build GRCh38
Transcripts
NM_001171937.1 NM_025129.4 NR_033269.1 XM_006723399.3 XM_011527339.1 XM_011527340.1 XM_011527341.2 XM_011527342.1 XM_011527343.1 XM_011527345.1 XM_011527346.1 XM_011527347.1 XM_017027319.1 XM_017027320.1 XM_017027321.1 XM_017027322.1 XM_017027323.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6296009 c.45C>A p.L15L Chr.19: 49813062-49813062 on Build GRCh38
5553515 c.11A>C p.E4A Chr.19: 49813096-49813096 on Build GRCh38
7322728 c.72C>G p.F24L Chr.19: 49813035-49813035 on Build GRCh38
1395334 c.87C>T p.R29R Chr.19: 49813020-49813020 on Build GRCh38
7322730 c.34C>T p.L12L Chr.19: 49813073-49813073 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779959041 Chr.19: 49813115-49813115 on Build GRCh38
rs890239094 Chr.19: 49813245-49813245 on Build GRCh38
rs929514300 Chr.19: 49813216-49813216 on Build GRCh38
rs941808220 Chr.19: 49813248-49813248 on Build GRCh38
rs1000978206 Chr.19: 49813312-49813312 on Build GRCh38
rs772606201 Chr.19: 49813150-49813150 on Build GRCh38
rs750242672 Chr.19: 49813097-49813097 on Build GRCh38
rs1050600061 Chr.19: 49812939-49812939 on Build GRCh38
rs762129315 Chr.19: 49813049-49813049 on Build GRCh38
rs764877879 Chr.19: 49813094-49813094 on Build GRCh38
rs35562710 Chr.19: 49813296-49813297 on Build GRCh38
rs116006228 Chr.19: 49813211-49813211 on Build GRCh38
rs887531566 Chr.19: 49813052-49813052 on Build GRCh38
rs903125223 Chr.19: 49813005-49813005 on Build GRCh38
rs761879368 Chr.19: 49813044-49813044 on Build GRCh38
rs552157094 Chr.19: 49813378-49813378 on Build GRCh38
rs765480197 Chr.19: 49813242-49813242 on Build GRCh38
rs376521642 Chr.19: 49813341-49813341 on Build GRCh38
rs1002533642 Chr.19: 49813285-49813285 on Build GRCh38
rs751448820 Chr.19: 49813316-49813316 on Build GRCh38
rs569464121 Chr.19: 49812946-49812946 on Build GRCh38
rs1013179957 Chr.19: 49813315-49813315 on Build GRCh38
rs909344036 Chr.19: 49813230-49813230 on Build GRCh38
rs540685473 Chr.19: 49813014-49813014 on Build GRCh38
rs1010070410 Chr.19: 49813128-49813128 on Build GRCh38
rs35138412 Chr.19: 49813007-49813007 on Build GRCh38
rs751534804 Chr.19: 49813116-49813116 on Build GRCh38
rs752240337 Chr.19: 49813346-49813346 on Build GRCh38
rs1030248986 Chr.19: 49813395-49813395 on Build GRCh38
rs773369720 Chr.19: 49813272-49813272 on Build GRCh38
rs1024082719 Chr.19: 49813319-49813319 on Build GRCh38
rs765257649 Chr.19: 49813081-49813081 on Build GRCh38
rs571655708 Chr.19: 49813401-49813401 on Build GRCh38
rs555614456 Chr.19: 49813294-49813294 on Build GRCh38
rs1035593685 Chr.19: 49813104-49813104 on Build GRCh38
rs1047970332 Chr.19: 49813026-49813026 on Build GRCh38
rs905504774 Chr.19: 49813085-49813085 on Build GRCh38
rs979894613 Chr.19: 49813149-49813149 on Build GRCh38
rs754939669 Chr.19: 49813337-49813337 on Build GRCh38
rs970823306 Chr.19: 49813332-49813332 on Build GRCh38
rs1017628387 Chr.19: 49813349-49813349 on Build GRCh38
rs375562420 Chr.19: 49813133-49813133 on Build GRCh38
rs777348801 Chr.19: 49813136-49813136 on Build GRCh38
rs905557061 Chr.19: 49813276-49813276 on Build GRCh38
rs964715532 Chr.19: 49813381-49813381 on Build GRCh38
rs866230700 Chr.19: 49813163-49813163 on Build GRCh38
rs902840683 Chr.19: 49813299-49813299 on Build GRCh38
rs566599341 Chr.19: 49812940-49812940 on Build GRCh38
rs750299203 Chr.19: 49813297-49813297 on Build GRCh38
rs1047892196 Chr.19: 49813227-49813227 on Build GRCh38
rs776119260 Chr.19: 49813155-49813155 on Build GRCh38
rs746240237 Chr.19: 49813138-49813138 on Build GRCh38
rs964662391 Chr.19: 49813167-49813167 on Build GRCh38
rs544884496 Chr.19: 49813132-49813132 on Build GRCh38
rs548706733 Chr.19: 49812987-49813009 on Build GRCh38
rs111464194 Chr.19: 49812993-49812993 on Build GRCh38
rs555174349 Chr.19: 49813004-49813004 on Build GRCh38
rs199809983 Chr.19: 49813123-49813123 on Build GRCh38
rs763833574 Chr.19: 49813237-49813237 on Build GRCh38
rs528559533 Chr.19: 49813306-49813306 on Build GRCh38
rs1057113927 Chr.19: 49813289-49813289 on Build GRCh38
rs552904239 Chr.19: 49813108-49813108 on Build GRCh38
rs1010537286 Chr.19: 49813300-49813300 on Build GRCh38
rs1044520267 Chr.19: 49813261-49813261 on Build GRCh38
rs768819337 Chr.19: 49813198-49813198 on Build GRCh38
rs1006344377 Chr.19: 49813060-49813060 on Build GRCh38
rs753379722 Chr.19: 49812979-49812979 on Build GRCh38
rs968931051 Chr.19: 49813307-49813307 on Build GRCh38
rs754068870 Chr.19: 49813021-49813021 on Build GRCh38
rs138010676 Chr.19: 49813388-49813388 on Build GRCh38
rs11882463 Chr.19: 49813292-49813292 on Build GRCh38
rs776858781 Chr.19: 49813203-49813203 on Build GRCh38
rs774809194 Chr.19: 49812955-49812955 on Build GRCh38
rs762871540 Chr.19: 49813128-49813129 on Build GRCh38
rs563180874 Chr.19: 49813186-49813186 on Build GRCh38
rs923463418 Chr.19: 49813008-49813008 on Build GRCh38
rs762394973 Chr.19: 49813290-49813290 on Build GRCh38
rs759904832 Chr.19: 49812960-49812960 on Build GRCh38
rs940984650 Chr.19: 49813185-49813185 on Build GRCh38
rs535629427 Chr.19: 49813208-49813208 on Build GRCh38
rs751345148 Chr.19: 49813305-49813305 on Build GRCh38

View Full Product Details