Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 49876501-49876996 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT1S1
Entrez Gene ID
84335
Gene Name
AKT1 substrate 1
Gene Aliases
Lobe, PRAS40
UniGene
Hs.515542
Gene Chromosome Location
Chr.19: 49869033-49878356 on Build GRCh38
Transcripts
NM_001098632.2 NM_001098633.3 NM_001278160.1 NM_032375.5

SNP Detail

SNP ID Chromosome Location
rs905946458 Chr.19: 49876841-49876841 on Build GRCh38
rs1035487033 Chr.19: 49876865-49876865 on Build GRCh38
rs961566311 Chr.19: 49876772-49876772 on Build GRCh38
rs545016565 Chr.19: 49876756-49876756 on Build GRCh38
rs921726692 Chr.19: 49876592-49876592 on Build GRCh38
rs1026916732 Chr.19: 49876908-49876908 on Build GRCh38
rs1031988727 Chr.19: 49876553-49876553 on Build GRCh38
rs754536239 Chr.19: 49876691-49876691 on Build GRCh38
rs920227745 Chr.19: 49876781-49876781 on Build GRCh38
rs972929481 Chr.19: 49876964-49876964 on Build GRCh38
rs539469413 Chr.19: 49876653-49876653 on Build GRCh38
rs113664904 Chr.19: 49876549-49876549 on Build GRCh38
rs1002990447 Chr.19: 49876857-49876857 on Build GRCh38
rs996211300 Chr.19: 49876858-49876858 on Build GRCh38
rs577585015 Chr.19: 49876735-49876735 on Build GRCh38
rs183401428 Chr.19: 49876962-49876962 on Build GRCh38
rs961580265 Chr.19: 49876709-49876709 on Build GRCh38
rs1008638876 Chr.19: 49876897-49876897 on Build GRCh38
rs569669397 Chr.19: 49876698-49876698 on Build GRCh38
rs985073076 Chr.19: 49876955-49876955 on Build GRCh38
rs747800215 Chr.19: 49876700-49876700 on Build GRCh38
rs770494508 Chr.19: 49876659-49876659 on Build GRCh38
rs761636229 Chr.19: 49876667-49876667 on Build GRCh38
rs939978236 Chr.19: 49876537-49876537 on Build GRCh38
rs893759102 Chr.19: 49876843-49876843 on Build GRCh38
rs780079786 Chr.19: 49876682-49876682 on Build GRCh38
rs949487178 Chr.19: 49876823-49876823 on Build GRCh38
rs368073989 Chr.19: 49876734-49876734 on Build GRCh38
rs750324380 Chr.19: 49876678-49876678 on Build GRCh38
rs1054111131 Chr.19: 49876842-49876842 on Build GRCh38
rs893100860 Chr.19: 49876809-49876809 on Build GRCh38
rs902085371 Chr.19: 49876616-49876616 on Build GRCh38
rs374755927 Chr.19: 49876789-49876789 on Build GRCh38
rs562509218 Chr.19: 49876835-49876835 on Build GRCh38
rs931020604 Chr.19: 49876564-49876564 on Build GRCh38
rs1046215986 Chr.19: 49876701-49876701 on Build GRCh38
rs192507165 Chr.19: 49876726-49876726 on Build GRCh38
rs970160370 Chr.19: 49876673-49876673 on Build GRCh38
rs952766770 Chr.19: 49876912-49876912 on Build GRCh38
rs889258262 Chr.19: 49876567-49876567 on Build GRCh38
rs537614295 Chr.19: 49876711-49876711 on Build GRCh38
rs773855174 Chr.19: 49876722-49876722 on Build GRCh38
rs114481180 Chr.19: 49876525-49876525 on Build GRCh38
rs999152903 Chr.19: 49876634-49876634 on Build GRCh38
rs890172626 Chr.19: 49876876-49876876 on Build GRCh38
rs919635346 Chr.19: 49876563-49876563 on Build GRCh38
rs547752182 Chr.19: 49876600-49876600 on Build GRCh38
rs1055613639 Chr.19: 49876689-49876689 on Build GRCh38
rs774852190 Chr.19: 49876557-49876557 on Build GRCh38
rs751010100 Chr.19: 49876686-49876686 on Build GRCh38
rs8105025 Chr.19: 49876828-49876828 on Build GRCh38
rs1037100843 Chr.19: 49876538-49876538 on Build GRCh38
rs865922611 Chr.19: 49876692-49876692 on Build GRCh38
rs143653837 Chr.19: 49876779-49876779 on Build GRCh38
rs529169191 Chr.19: 49876595-49876595 on Build GRCh38
rs574222760 Chr.19: 49876719-49876719 on Build GRCh38
rs902416590 Chr.19: 49876707-49876707 on Build GRCh38
rs186988431 Chr.19: 49876651-49876651 on Build GRCh38
rs973036236 Chr.19: 49876905-49876905 on Build GRCh38
rs561721681 Chr.19: 49876887-49876887 on Build GRCh38
rs1023071384 Chr.19: 49876666-49876666 on Build GRCh38
rs1040546926 Chr.19: 49876593-49876593 on Build GRCh38
rs935190552 Chr.19: 49876838-49876838 on Build GRCh38
rs765139198 Chr.19: 49876669-49876669 on Build GRCh38
rs529118014 Chr.19: 49876919-49876919 on Build GRCh38
rs753447833 Chr.19: 49876567-49876567 on Build GRCh38
rs1032250704 Chr.19: 49876714-49876714 on Build GRCh38
rs542233340 Chr.19: 49876699-49876699 on Build GRCh38
rs534998891 Chr.19: 49876723-49876723 on Build GRCh38
rs555686149 Chr.19: 49876716-49876716 on Build GRCh38
rs990843255 Chr.19: 49876795-49876795 on Build GRCh38
rs916610444 Chr.19: 49876817-49876817 on Build GRCh38
rs893446247 Chr.19: 49876715-49876715 on Build GRCh38
rs957784066 Chr.19: 49876556-49876556 on Build GRCh38
rs866830970 Chr.19: 49876649-49876649 on Build GRCh38
rs746769871 Chr.19: 49876570-49876571 on Build GRCh38
rs1053392526 Chr.19: 49876793-49876793 on Build GRCh38
rs1012053703 Chr.19: 49876664-49876664 on Build GRCh38
rs1027614974 Chr.19: 49876967-49876967 on Build GRCh38
rs974886829 Chr.19: 49876591-49876591 on Build GRCh38
rs551475314 Chr.19: 49876662-49876662 on Build GRCh38
rs966605157 Chr.19: 49876520-49876520 on Build GRCh38
rs1028660985 Chr.19: 49876874-49876874 on Build GRCh38
rs961534563 Chr.19: 49876957-49876957 on Build GRCh38
rs1014705173 Chr.19: 49876941-49876941 on Build GRCh38

View Full Product Details