Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55593252-55593754 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FIZ1
Entrez Gene ID
84922
Gene Name
FLT3 interacting zinc finger 1
Gene Aliases
ZNF798
UniGene
Hs.515617
Gene Chromosome Location
Chr.19: 55591371-55599703 on Build GRCh38
Transcripts
NM_032836.2 XM_005259352.4 XM_011527426.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7219275 c.640G>C p.E214Q Chr.19: 55593301-55593301 on Build GRCh38
6374476 c.497C>T p.S166L Chr.19: 55593444-55593444 on Build GRCh38
1396628 c.351_353delCTC p.S118delS Chr.19: 55593588-55593590 on Build GRCh38
4914328 c.331G>A p.V111I Chr.19: 55593610-55593610 on Build GRCh38
4916585 c.536C>T p.A179V Chr.19: 55593405-55593405 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758460709 Chr.19: 55593393-55593393 on Build GRCh38
rs917399417 Chr.19: 55593371-55593371 on Build GRCh38
rs376511395 Chr.19: 55593522-55593522 on Build GRCh38
rs2287792 Chr.19: 55593554-55593554 on Build GRCh38
rs1040330736 Chr.19: 55593299-55593299 on Build GRCh38
rs760668931 Chr.19: 55593325-55593325 on Build GRCh38
rs773996729 Chr.19: 55593308-55593308 on Build GRCh38
rs1005976017 Chr.19: 55593323-55593323 on Build GRCh38
rs779635479 Chr.19: 55593697-55593697 on Build GRCh38
rs1028704001 Chr.19: 55593313-55593313 on Build GRCh38
rs79017610 Chr.19: 55593495-55593495 on Build GRCh38
rs771521229 Chr.19: 55593578-55593578 on Build GRCh38
rs568861567 Chr.19: 55593403-55593403 on Build GRCh38
rs555218367 Chr.19: 55593652-55593652 on Build GRCh38
rs927349406 Chr.19: 55593415-55593415 on Build GRCh38
rs1014687571 Chr.19: 55593681-55593681 on Build GRCh38
rs772755613 Chr.19: 55593612-55593612 on Build GRCh38
rs773601056 Chr.19: 55593281-55593281 on Build GRCh38
rs747522877 Chr.19: 55593425-55593425 on Build GRCh38
rs766448332 Chr.19: 55593649-55593649 on Build GRCh38
rs748201808 Chr.19: 55593278-55593278 on Build GRCh38
rs944115050 Chr.19: 55593550-55593550 on Build GRCh38
rs982710039 Chr.19: 55593537-55593537 on Build GRCh38
rs913280432 Chr.19: 55593449-55593449 on Build GRCh38
rs1054566421 Chr.19: 55593434-55593434 on Build GRCh38
rs1033921357 Chr.19: 55593451-55593451 on Build GRCh38
rs750386176 Chr.19: 55593318-55593318 on Build GRCh38
rs752906489 Chr.19: 55593694-55593694 on Build GRCh38
rs976903247 Chr.19: 55593653-55593653 on Build GRCh38
rs766319486 Chr.19: 55593326-55593326 on Build GRCh38
rs754774124 Chr.19: 55593347-55593347 on Build GRCh38
rs773107433 Chr.19: 55593547-55593547 on Build GRCh38
rs538399783 Chr.19: 55593429-55593429 on Build GRCh38
rs753719897 Chr.19: 55593335-55593335 on Build GRCh38
rs189761223 Chr.19: 55593383-55593383 on Build GRCh38
rs538296469 Chr.19: 55593362-55593362 on Build GRCh38
rs759415322 Chr.19: 55593674-55593674 on Build GRCh38
rs776947272 Chr.19: 55593291-55593291 on Build GRCh38
rs950647216 Chr.19: 55593536-55593536 on Build GRCh38
rs1040256213 Chr.19: 55593459-55593459 on Build GRCh38
rs575158270 Chr.19: 55593698-55593698 on Build GRCh38
rs771330574 Chr.19: 55593283-55593283 on Build GRCh38
rs961585581 Chr.19: 55593349-55593349 on Build GRCh38
rs973707762 Chr.19: 55593521-55593521 on Build GRCh38
rs761233202 Chr.19: 55593310-55593310 on Build GRCh38
rs371860981 Chr.19: 55593293-55593293 on Build GRCh38
rs746237008 Chr.19: 55593493-55593493 on Build GRCh38
rs921415023 Chr.19: 55593656-55593656 on Build GRCh38
rs747534082 Chr.19: 55593282-55593282 on Build GRCh38
rs894039911 Chr.19: 55593705-55593705 on Build GRCh38
rs1050299075 Chr.19: 55593586-55593586 on Build GRCh38
rs760211380 Chr.19: 55593626-55593626 on Build GRCh38
rs777883921 Chr.19: 55593405-55593405 on Build GRCh38
rs367954318 Chr.19: 55593530-55593530 on Build GRCh38
rs945612739 Chr.19: 55593720-55593720 on Build GRCh38
rs1003300949 Chr.19: 55593437-55593437 on Build GRCh38
rs372128345 Chr.19: 55593695-55593695 on Build GRCh38
rs1054915578 Chr.19: 55593284-55593284 on Build GRCh38
rs546515126 Chr.19: 55593302-55593302 on Build GRCh38
rs901676871 Chr.19: 55593500-55593500 on Build GRCh38
rs1006275632 Chr.19: 55593671-55593671 on Build GRCh38
rs566444859 Chr.19: 55593317-55593317 on Build GRCh38
rs765063254 Chr.19: 55593688-55593688 on Build GRCh38
rs772136806 Chr.19: 55593279-55593279 on Build GRCh38
rs779240583 Chr.19: 55593274-55593274 on Build GRCh38
rs781636082 Chr.19: 55593490-55593490 on Build GRCh38
rs759852286 Chr.19: 55593294-55593294 on Build GRCh38
rs908559646 Chr.19: 55593545-55593545 on Build GRCh38
rs770087686 Chr.19: 55593506-55593506 on Build GRCh38

View Full Product Details