Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55677369-55677884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN1
Entrez Gene ID
29924
Gene Name
epsin 1
Gene Aliases
-
UniGene
Hs.279953
Gene Chromosome Location
Chr.19: 55675195-55695767 on Build GRCh38
Transcripts
NM_001130071.1 NM_001130072.1 NM_013333.3 XM_005258829.2 XM_005258830.2 XM_011526880.1 XM_011526881.1 XM_017026722.1 XM_017026723.1 XM_017026724.1 XM_017026725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6406910 c.191C>A p.T64K Chr.19: 55677693-55677693 on Build GRCh38
5655870 c.57C>G p.V19V Chr.19: 55677559-55677559 on Build GRCh38
6421451 c.178G>T p.A60S Chr.19: 55677680-55677680 on Build GRCh38
5458787 c.66C>T p.C22C Chr.19: 55677568-55677568 on Build GRCh38
5105106 c.100A>C p.S34R Chr.19: 55677602-55677602 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766600178 Chr.19: 55677615-55677616 on Build GRCh38
rs367940444 Chr.19: 55677604-55677604 on Build GRCh38
rs1045032663 Chr.19: 55677403-55677403 on Build GRCh38
rs747647038 Chr.19: 55677754-55677754 on Build GRCh38
rs560770102 Chr.19: 55677470-55677470 on Build GRCh38
rs771788748 Chr.19: 55677426-55677426 on Build GRCh38
rs748376690 Chr.19: 55677490-55677490 on Build GRCh38
rs776944633 Chr.19: 55677657-55677657 on Build GRCh38
rs774750574 Chr.19: 55677679-55677679 on Build GRCh38
rs1036514354 Chr.19: 55677730-55677730 on Build GRCh38
rs1019301347 Chr.19: 55677486-55677486 on Build GRCh38
rs531746730 Chr.19: 55677585-55677585 on Build GRCh38
rs985229252 Chr.19: 55677597-55677597 on Build GRCh38
rs759630951 Chr.19: 55677663-55677663 on Build GRCh38
rs1044918979 Chr.19: 55677626-55677626 on Build GRCh38
rs10415521 Chr.19: 55677406-55677406 on Build GRCh38
rs531866551 Chr.19: 55677611-55677611 on Build GRCh38
rs1030660698 Chr.19: 55677427-55677427 on Build GRCh38
rs757642644 Chr.19: 55677711-55677711 on Build GRCh38
rs183276241 Chr.19: 55677555-55677555 on Build GRCh38
rs746881434 Chr.19: 55677768-55677768 on Build GRCh38
rs779769951 Chr.19: 55677589-55677589 on Build GRCh38
rs997741902 Chr.19: 55677753-55677753 on Build GRCh38
rs777769354 Chr.19: 55677734-55677734 on Build GRCh38
rs976060242 Chr.19: 55677489-55677489 on Build GRCh38
rs754342627 Chr.19: 55677488-55677488 on Build GRCh38
rs546451407 Chr.19: 55677641-55677641 on Build GRCh38
rs375326620 Chr.19: 55677732-55677732 on Build GRCh38
rs574990870 Chr.19: 55677414-55677414 on Build GRCh38
rs770781947 Chr.19: 55677782-55677782 on Build GRCh38
rs537506253 Chr.19: 55677838-55677838 on Build GRCh38
rs892094345 Chr.19: 55677449-55677449 on Build GRCh38
rs1018324891 Chr.19: 55677789-55677789 on Build GRCh38
rs190641857 Chr.19: 55677518-55677518 on Build GRCh38
rs775209668 Chr.19: 55677456-55677456 on Build GRCh38
rs1015567996 Chr.19: 55677746-55677746 on Build GRCh38
rs778342121 Chr.19: 55677603-55677603 on Build GRCh38
rs1007936526 Chr.19: 55677454-55677454 on Build GRCh38
rs116764574 Chr.19: 55677848-55677848 on Build GRCh38
rs796405646 Chr.19: 55677605-55677605 on Build GRCh38
rs748821014 Chr.19: 55677595-55677595 on Build GRCh38
rs767440458 Chr.19: 55677852-55677852 on Build GRCh38
rs752791573 Chr.19: 55677678-55677678 on Build GRCh38
rs375465578 Chr.19: 55677728-55677728 on Build GRCh38
rs766949554 Chr.19: 55677674-55677674 on Build GRCh38
rs752053651 Chr.19: 55677693-55677693 on Build GRCh38
rs771516776 Chr.19: 55677756-55677756 on Build GRCh38
rs568554895 Chr.19: 55677715-55677715 on Build GRCh38
rs925141587 Chr.19: 55677820-55677820 on Build GRCh38
rs1014672057 Chr.19: 55677467-55677467 on Build GRCh38
rs764683689 Chr.19: 55677683-55677683 on Build GRCh38
rs925226836 Chr.19: 55677627-55677627 on Build GRCh38
rs577269291 Chr.19: 55677504-55677504 on Build GRCh38
rs917993753 Chr.19: 55677615-55677615 on Build GRCh38
rs773053554 Chr.19: 55677610-55677610 on Build GRCh38
rs551786479 Chr.19: 55677729-55677729 on Build GRCh38
rs974866779 Chr.19: 55677538-55677538 on Build GRCh38
rs996338415 Chr.19: 55677714-55677714 on Build GRCh38
rs962183889 Chr.19: 55677483-55677483 on Build GRCh38
rs548679846 Chr.19: 55677586-55677586 on Build GRCh38
rs187943456 Chr.19: 55677560-55677560 on Build GRCh38
rs568593622 Chr.19: 55677796-55677796 on Build GRCh38
rs77635294 Chr.19: 55677602-55677602 on Build GRCh38
rs1029681574 Chr.19: 55677583-55677584 on Build GRCh38
rs138155404 Chr.19: 55677419-55677419 on Build GRCh38
rs192665194 Chr.19: 55677847-55677847 on Build GRCh38
rs566472226 Chr.19: 55677652-55677652 on Build GRCh38
rs1040798833 Chr.19: 55677680-55677680 on Build GRCh38
rs760037138 Chr.19: 55677834-55677834 on Build GRCh38
rs953249020 Chr.19: 55677781-55677781 on Build GRCh38
rs754561257 Chr.19: 55677727-55677727 on Build GRCh38
rs936611243 Chr.19: 55677654-55677654 on Build GRCh38
rs74369843 Chr.19: 55677619-55677619 on Build GRCh38
rs981228678 Chr.19: 55677786-55677786 on Build GRCh38
rs1006905470 Chr.19: 55677787-55677787 on Build GRCh38
rs962067706 Chr.19: 55677747-55677747 on Build GRCh38
rs777695285 Chr.19: 55677765-55677765 on Build GRCh38
rs762919490 Chr.19: 55677653-55677653 on Build GRCh38
rs952325665 Chr.19: 55677591-55677591 on Build GRCh38
rs985436169 Chr.19: 55677545-55677545 on Build GRCh38

View Full Product Details