Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55689746-55690251 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EPN1
Entrez Gene ID
29924
Gene Name
epsin 1
Gene Aliases
-
UniGene
Hs.279953
Gene Chromosome Location
Chr.19: 55675195-55695767 on Build GRCh38
Transcripts
NM_001130071.1 NM_001130072.1 NM_013333.3 XM_005258829.2 XM_005258830.2 XM_011526880.1 XM_011526881.1 XM_017026722.1 XM_017026723.1 XM_017026724.1 XM_017026725.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6035797 c.985G>A p.E329K Chr.19: 55689915-55689915 on Build GRCh38
6858509 c.955G>C p.G319R Chr.19: 55689885-55689885 on Build GRCh38
6858510 c.697G>C p.G233R Chr.19: 55689885-55689885 on Build GRCh38
6035798 c.727G>A p.E243K Chr.19: 55689915-55689915 on Build GRCh38
7297530 c.690C>T p.I230I Chr.19: 55689878-55689878 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757184530 Chr.19: 55689829-55689829 on Build GRCh38
rs112928863 Chr.19: 55689770-55689770 on Build GRCh38
rs756004190 Chr.19: 55689917-55689917 on Build GRCh38
rs900984579 Chr.19: 55690161-55690161 on Build GRCh38
rs540614654 Chr.19: 55689978-55689978 on Build GRCh38
rs150783945 Chr.19: 55690051-55690051 on Build GRCh38
rs535468920 Chr.19: 55689768-55689768 on Build GRCh38
rs748957707 Chr.19: 55689989-55689989 on Build GRCh38
rs1007591173 Chr.19: 55690189-55690189 on Build GRCh38
rs369530032 Chr.19: 55689905-55689905 on Build GRCh38
rs779027488 Chr.19: 55689860-55689860 on Build GRCh38
rs960173812 Chr.19: 55690208-55690208 on Build GRCh38
rs767572552 Chr.19: 55689914-55689914 on Build GRCh38
rs1030271689 Chr.19: 55689821-55689821 on Build GRCh38
rs968856352 Chr.19: 55690195-55690195 on Build GRCh38
rs373848848 Chr.19: 55689894-55689894 on Build GRCh38
rs772756148 Chr.19: 55690201-55690201 on Build GRCh38
rs1005849736 Chr.19: 55690152-55690152 on Build GRCh38
rs752283913 Chr.19: 55689819-55689819 on Build GRCh38
rs778920721 Chr.19: 55689956-55689956 on Build GRCh38
rs569998903 Chr.19: 55690090-55690090 on Build GRCh38
rs745923943 Chr.19: 55689846-55689846 on Build GRCh38
rs139148311 Chr.19: 55690175-55690175 on Build GRCh38
rs971963536 Chr.19: 55690179-55690179 on Build GRCh38
rs914741975 Chr.19: 55689842-55689842 on Build GRCh38
rs201054079 Chr.19: 55689969-55689969 on Build GRCh38
rs772556000 Chr.19: 55689874-55689874 on Build GRCh38
rs528550276 Chr.19: 55690018-55690018 on Build GRCh38
rs747469500 Chr.19: 55689967-55689967 on Build GRCh38
rs773651149 Chr.19: 55689879-55689879 on Build GRCh38
rs73933309 Chr.19: 55689999-55689999 on Build GRCh38
rs1027264012 Chr.19: 55689769-55689769 on Build GRCh38
rs888279513 Chr.19: 55689943-55689943 on Build GRCh38
rs887706727 Chr.19: 55690221-55690221 on Build GRCh38
rs909667214 Chr.19: 55690113-55690113 on Build GRCh38
rs866007670 Chr.19: 55690172-55690172 on Build GRCh38
rs758406549 Chr.19: 55689960-55689960 on Build GRCh38
rs998044247 Chr.19: 55689983-55689983 on Build GRCh38
rs754055979 Chr.19: 55689935-55689935 on Build GRCh38
rs762648488 Chr.19: 55689891-55689891 on Build GRCh38
rs543696962 Chr.19: 55689873-55689873 on Build GRCh38
rs369836746 Chr.19: 55689848-55689848 on Build GRCh38
rs535761633 Chr.19: 55690146-55690146 on Build GRCh38
rs370062190 Chr.19: 55689820-55689820 on Build GRCh38
rs927540438 Chr.19: 55690185-55690185 on Build GRCh38
rs776794363 Chr.19: 55690060-55690060 on Build GRCh38
rs749485844 Chr.19: 55689854-55689854 on Build GRCh38
rs763613050 Chr.19: 55689823-55689823 on Build GRCh38
rs962026977 Chr.19: 55689798-55689798 on Build GRCh38
rs145192073 Chr.19: 55690224-55690224 on Build GRCh38
rs539687094 Chr.19: 55690054-55690054 on Build GRCh38
rs1044419401 Chr.19: 55689812-55689812 on Build GRCh38
rs763549998 Chr.19: 55689893-55689893 on Build GRCh38
rs950317579 Chr.19: 55689795-55689795 on Build GRCh38
rs755938983 Chr.19: 55689802-55689802 on Build GRCh38
rs114589249 Chr.19: 55690216-55690216 on Build GRCh38
rs779943680 Chr.19: 55689923-55689923 on Build GRCh38
rs376690721 Chr.19: 55689843-55689843 on Build GRCh38
rs141887410 Chr.19: 55690023-55690023 on Build GRCh38
rs948375378 Chr.19: 55690069-55690069 on Build GRCh38
rs375981361 Chr.19: 55689883-55689883 on Build GRCh38
rs1021675976 Chr.19: 55690008-55690008 on Build GRCh38
rs530872693 Chr.19: 55689780-55689780 on Build GRCh38
rs1006068441 Chr.19: 55690222-55690222 on Build GRCh38
rs559712752 Chr.19: 55690007-55690007 on Build GRCh38
rs748274043 Chr.19: 55689959-55689959 on Build GRCh38
rs777026436 Chr.19: 55689980-55689980 on Build GRCh38
rs769424973 Chr.19: 55690183-55690183 on Build GRCh38
rs534379415 Chr.19: 55690190-55690190 on Build GRCh38
rs780034411 Chr.19: 55689998-55689998 on Build GRCh38
rs1048627228 Chr.19: 55690091-55690091 on Build GRCh38
rs2303760 Chr.19: 55689772-55689772 on Build GRCh38
rs371100606 Chr.19: 55689994-55689994 on Build GRCh38
rs1028863203 Chr.19: 55690144-55690144 on Build GRCh38
rs901716655 Chr.19: 55690206-55690206 on Build GRCh38
rs761033687 Chr.19: 55689882-55689882 on Build GRCh38
rs758331662 Chr.19: 55689974-55689974 on Build GRCh38
rs373584560 Chr.19: 55689828-55689828 on Build GRCh38
rs112909591 Chr.19: 55689952-55689952 on Build GRCh38
rs563684826 Chr.19: 55689936-55689936 on Build GRCh38
rs953379853 Chr.19: 55690006-55690006 on Build GRCh38
rs370140669 Chr.19: 55689884-55689884 on Build GRCh38
rs564980731 Chr.19: 55690053-55690053 on Build GRCh38
rs113109595 Chr.19: 55689951-55689951 on Build GRCh38
rs1031858879 Chr.19: 55690202-55690202 on Build GRCh38
rs977628158 Chr.19: 55690019-55690019 on Build GRCh38
rs993053178 Chr.19: 55690057-55690057 on Build GRCh38
rs762417950 Chr.19: 55690220-55690220 on Build GRCh38
rs972937271 Chr.19: 55689975-55689975 on Build GRCh38
rs780426545 Chr.19: 55689851-55689851 on Build GRCh38
rs747903554 Chr.19: 55690106-55690106 on Build GRCh38
rs372757543 Chr.19: 55689855-55689855 on Build GRCh38
rs550097728 Chr.19: 55690062-55690062 on Build GRCh38
rs996143620 Chr.19: 55690072-55690072 on Build GRCh38
rs555032815 Chr.19: 55690157-55690157 on Build GRCh38
rs939746688 Chr.19: 55690149-55690149 on Build GRCh38

View Full Product Details