Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 56183144-56183586 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALP
Entrez Gene ID
85569
Gene Name
galanin like peptide
Gene Aliases
-
UniGene
Hs.283915
Gene Chromosome Location
Chr.19: 56176020-56185775 on Build GRCh38
Transcripts
NM_001145546.1 NM_033106.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5568903 c.250T>C p.S84P Chr.19: 56183167-56183167 on Build GRCh38
714459 c.255G>T p.K85N Chr.19: 56183172-56183172 on Build GRCh38
5856854 c.295+1G>A p.? Chr.19: 56183213-56183213 on Build GRCh38
6997121 c.246G>A p.Q82Q Chr.19: 56183163-56183163 on Build GRCh38
3539924 c.288G>A p.E96E Chr.19: 56183205-56183205 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1019404118 Chr.19: 56183247-56183247 on Build GRCh38
rs558989766 Chr.19: 56183495-56183495 on Build GRCh38
rs767792935 Chr.19: 56183490-56183490 on Build GRCh38
rs991645700 Chr.19: 56183501-56183501 on Build GRCh38
rs542828152 Chr.19: 56183432-56183432 on Build GRCh38
rs928778723 Chr.19: 56183358-56183358 on Build GRCh38
rs981821374 Chr.19: 56183486-56183486 on Build GRCh38
rs908506309 Chr.19: 56183559-56183559 on Build GRCh38
rs759723558 Chr.19: 56183436-56183436 on Build GRCh38
rs543349847 Chr.19: 56183372-56183372 on Build GRCh38
rs199898051 Chr.19: 56183344-56183344 on Build GRCh38
rs931556428 Chr.19: 56183549-56183549 on Build GRCh38
rs892584685 Chr.19: 56183406-56183406 on Build GRCh38
rs137957133 Chr.19: 56183558-56183558 on Build GRCh38
rs780273695 Chr.19: 56183240-56183240 on Build GRCh38
rs970451311 Chr.19: 56183481-56183481 on Build GRCh38
rs200987565 Chr.19: 56183237-56183237 on Build GRCh38
rs773785154 Chr.19: 56183271-56183271 on Build GRCh38
rs371443019 Chr.19: 56183207-56183207 on Build GRCh38
rs745308267 Chr.19: 56183232-56183235 on Build GRCh38
rs1056036469 Chr.19: 56183361-56183361 on Build GRCh38
rs899636905 Chr.19: 56183415-56183415 on Build GRCh38
rs866110245 Chr.19: 56183357-56183357 on Build GRCh38
rs961785631 Chr.19: 56183519-56183519 on Build GRCh38
rs903983267 Chr.19: 56183428-56183428 on Build GRCh38
rs961754534 Chr.19: 56183334-56183334 on Build GRCh38
rs750069427 Chr.19: 56183230-56183230 on Build GRCh38
rs112153136 Chr.19: 56183264-56183264 on Build GRCh38
rs1018534412 Chr.19: 56183497-56183497 on Build GRCh38
rs1009509000 Chr.19: 56183441-56183441 on Build GRCh38
rs1021035207 Chr.19: 56183450-56183450 on Build GRCh38
rs553459016 Chr.19: 56183213-56183213 on Build GRCh38
rs752577921 Chr.19: 56183189-56183189 on Build GRCh38
rs545390063 Chr.19: 56183253-56183253 on Build GRCh38
rs764919010 Chr.19: 56183183-56183183 on Build GRCh38
rs772300764 Chr.19: 56183256-56183256 on Build GRCh38
rs1032190254 Chr.19: 56183433-56183433 on Build GRCh38
rs1008454094 Chr.19: 56183289-56183289 on Build GRCh38
rs997117984 Chr.19: 56183438-56183438 on Build GRCh38
rs567747236 Chr.19: 56183172-56183172 on Build GRCh38
rs920132532 Chr.19: 56183517-56183517 on Build GRCh38
rs949136657 Chr.19: 56183323-56183323 on Build GRCh38
rs767098292 Chr.19: 56183309-56183309 on Build GRCh38
rs57532371 Chr.19: 56183382-56183382 on Build GRCh38
rs1008722082 Chr.19: 56183396-56183396 on Build GRCh38
rs752474196 Chr.19: 56183553-56183553 on Build GRCh38
rs996653560 Chr.19: 56183416-56183416 on Build GRCh38
rs762876372 Chr.19: 56183287-56183287 on Build GRCh38
rs1047489020 Chr.19: 56183375-56183375 on Build GRCh38
rs957703846 Chr.19: 56183268-56183268 on Build GRCh38
rs971882666 Chr.19: 56183356-56183356 on Build GRCh38
rs373719152 Chr.19: 56183219-56183219 on Build GRCh38
rs770414156 Chr.19: 56183179-56183179 on Build GRCh38
rs200384723 Chr.19: 56183244-56183244 on Build GRCh38
rs150355226 Chr.19: 56183409-56183409 on Build GRCh38
rs963881024 Chr.19: 56183250-56183250 on Build GRCh38
rs1056461667 Chr.19: 56183397-56183397 on Build GRCh38
rs1029027880 Chr.19: 56183485-56183485 on Build GRCh38
rs762796454 Chr.19: 56183190-56183190 on Build GRCh38
rs898758351 Chr.19: 56183371-56183371 on Build GRCh38
rs997483920 Chr.19: 56183226-56183227 on Build GRCh38
rs575736751 Chr.19: 56183288-56183288 on Build GRCh38
rs937497732 Chr.19: 56183359-56183359 on Build GRCh38
rs749717587 Chr.19: 56183243-56183243 on Build GRCh38
rs764079273 Chr.19: 56183195-56183195 on Build GRCh38

View Full Product Details