Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 57972363-57972865 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf18
Entrez Gene ID
147685
Gene Name
chromosome 19 open reading frame 18
Gene Aliases
-
UniGene
Hs.134209
Gene Chromosome Location
Chr.19: 57958437-57974534 on Build GRCh38
Transcripts
NM_152474.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2973539 c.245C>G p.P82R Chr.19: 57972486-57972486 on Build GRCh38
1198299 c.233C>T p.S78F Chr.19: 57972498-57972498 on Build GRCh38
4082254 c.257T>C p.L86P Chr.19: 57972474-57972474 on Build GRCh38
3893761 c.249G>A p.M83I Chr.19: 57972482-57972482 on Build GRCh38
5963572 c.227-5T>C p.? Chr.19: 57972509-57972509 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs11668548 Chr.19: 57972586-57972586 on Build GRCh38
rs564972784 Chr.19: 57972601-57972601 on Build GRCh38
rs140552936 Chr.19: 57972419-57972419 on Build GRCh38
rs373597607 Chr.19: 57972486-57972486 on Build GRCh38
rs139303329 Chr.19: 57972492-57972492 on Build GRCh38
rs745795542 Chr.19: 57972674-57972674 on Build GRCh38
rs781449173 Chr.19: 57972538-57972538 on Build GRCh38
rs778029673 Chr.19: 57972505-57972505 on Build GRCh38
rs181615082 Chr.19: 57972497-57972497 on Build GRCh38
rs192110064 Chr.19: 57972648-57972648 on Build GRCh38
rs1053366754 Chr.19: 57972725-57972725 on Build GRCh38
rs151312833 Chr.19: 57972409-57972409 on Build GRCh38
rs538763620 Chr.19: 57972776-57972776 on Build GRCh38
rs779532154 Chr.19: 57972496-57972496 on Build GRCh38
rs920004862 Chr.19: 57972644-57972644 on Build GRCh38
rs755550384 Chr.19: 57972403-57972403 on Build GRCh38
rs779152605 Chr.19: 57972800-57972800 on Build GRCh38
rs370483026 Chr.19: 57972452-57972452 on Build GRCh38
rs1011730889 Chr.19: 57972826-57972826 on Build GRCh38
rs1030219691 Chr.19: 57972605-57972605 on Build GRCh38
rs757605935 Chr.19: 57972651-57972651 on Build GRCh38
rs767242543 Chr.19: 57972465-57972465 on Build GRCh38
rs967710378 Chr.19: 57972447-57972447 on Build GRCh38
rs761691057 Chr.19: 57972550-57972550 on Build GRCh38
rs772176063 Chr.19: 57972755-57972755 on Build GRCh38
rs1044387817 Chr.19: 57972686-57972686 on Build GRCh38
rs891961542 Chr.19: 57972575-57972575 on Build GRCh38
rs938955344 Chr.19: 57972703-57972703 on Build GRCh38
rs367650205 Chr.19: 57972503-57972503 on Build GRCh38
rs376354741 Chr.19: 57972438-57972438 on Build GRCh38
rs776695354 Chr.19: 57972430-57972430 on Build GRCh38
rs745940805 Chr.19: 57972542-57972542 on Build GRCh38
rs1012553625 Chr.19: 57972842-57972842 on Build GRCh38
rs747187026 Chr.19: 57972512-57972512 on Build GRCh38
rs999085165 Chr.19: 57972765-57972765 on Build GRCh38
rs762817612 Chr.19: 57972437-57972437 on Build GRCh38
rs758388241 Chr.19: 57972541-57972550 on Build GRCh38
rs772363290 Chr.19: 57972426-57972426 on Build GRCh38
rs1031806652 Chr.19: 57972574-57972574 on Build GRCh38
rs1032957305 Chr.19: 57972838-57972838 on Build GRCh38
rs572982358 Chr.19: 57972408-57972408 on Build GRCh38
rs971096656 Chr.19: 57972517-57972517 on Build GRCh38
rs371358991 Chr.19: 57972549-57972549 on Build GRCh38
rs183953039 Chr.19: 57972668-57972668 on Build GRCh38
rs201797138 Chr.19: 57972439-57972439 on Build GRCh38
rs974358671 Chr.19: 57972604-57972604 on Build GRCh38
rs928875477 Chr.19: 57972691-57972691 on Build GRCh38
rs755557145 Chr.19: 57972418-57972418 on Build GRCh38
rs940181157 Chr.19: 57972584-57972584 on Build GRCh38
rs373387103 Chr.19: 57972428-57972428 on Build GRCh38
rs752631857 Chr.19: 57972532-57972536 on Build GRCh38
rs187704590 Chr.19: 57972787-57972787 on Build GRCh38
rs769897740 Chr.19: 57972548-57972548 on Build GRCh38
rs201033494 Chr.19: 57972491-57972491 on Build GRCh38
rs112541740 Chr.19: 57972623-57972623 on Build GRCh38
rs758741294 Chr.19: 57972498-57972498 on Build GRCh38
rs771152908 Chr.19: 57972535-57972535 on Build GRCh38
rs200941854 Chr.19: 57972551-57972551 on Build GRCh38
rs185767847 Chr.19: 57972413-57972413 on Build GRCh38
rs779105483 Chr.19: 57972639-57972639 on Build GRCh38
rs761633348 Chr.19: 57972451-57972451 on Build GRCh38
rs147698410 Chr.19: 57972694-57972694 on Build GRCh38
rs747218238 Chr.19: 57972429-57972429 on Build GRCh38
rs778881137 Chr.19: 57972515-57972524 on Build GRCh38
rs257675 Chr.19: 57972841-57972841 on Build GRCh38
rs185945713 Chr.19: 57972640-57972640 on Build GRCh38
rs760252473 Chr.19: 57972489-57972489 on Build GRCh38
rs200016229 Chr.19: 57972425-57972425 on Build GRCh38
rs12151075 Chr.19: 57972728-57972728 on Build GRCh38
rs1049735731 Chr.19: 57972622-57972622 on Build GRCh38
rs548083353 Chr.19: 57972617-57972617 on Build GRCh38
rs568011410 Chr.19: 57972633-57972633 on Build GRCh38
rs149999005 Chr.19: 57972602-57972602 on Build GRCh38
rs948686785 Chr.19: 57972524-57972524 on Build GRCh38

View Full Product Details