Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 279838-280368 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM150B
Entrez Gene ID
285016
Gene Name
family with sequence similarity 150 member B
Gene Aliases
PRO1097, RGPG542
UniGene
Hs.355207
Gene Chromosome Location
Chr.2: 279558-288873 on Build GRCh38
Transcripts
NM_001002919.2 XM_005264689.4 XM_006711879.3 XM_011510342.2

SNP Detail

SNP ID Chromosome Location
rs569316765 Chr.2: 280164-280165 on Build GRCh38
rs990512254 Chr.2: 279925-279925 on Build GRCh38
rs372528659 Chr.2: 279873-279874 on Build GRCh38
rs753276246 Chr.2: 280029-280029 on Build GRCh38
rs528850099 Chr.2: 279893-279893 on Build GRCh38
rs887974366 Chr.2: 280049-280049 on Build GRCh38
rs186654944 Chr.2: 280145-280145 on Build GRCh38
rs955062315 Chr.2: 280226-280226 on Build GRCh38
rs551918533 Chr.2: 279993-279993 on Build GRCh38
rs749818760 Chr.2: 280162-280162 on Build GRCh38
rs772698560 Chr.2: 280133-280133 on Build GRCh38
rs988953082 Chr.2: 280238-280238 on Build GRCh38
rs184724641 Chr.2: 280338-280338 on Build GRCh38
rs750317044 Chr.2: 280180-280180 on Build GRCh38
rs762242140 Chr.2: 279976-279976 on Build GRCh38
rs538236489 Chr.2: 280272-280272 on Build GRCh38
rs200480474 Chr.2: 280168-280168 on Build GRCh38
rs375783659 Chr.2: 280177-280177 on Build GRCh38
rs968936893 Chr.2: 280300-280300 on Build GRCh38
rs772310409 Chr.2: 280172-280172 on Build GRCh38
rs977299518 Chr.2: 279892-279892 on Build GRCh38
rs765983695 Chr.2: 280025-280025 on Build GRCh38
rs1020504114 Chr.2: 280256-280256 on Build GRCh38
rs764767937 Chr.2: 280069-280069 on Build GRCh38
rs762898026 Chr.2: 280181-280181 on Build GRCh38
rs942292251 Chr.2: 280068-280069 on Build GRCh38
rs781387569 Chr.2: 280284-280284 on Build GRCh38
rs62114550 Chr.2: 279931-279931 on Build GRCh38
rs867103687 Chr.2: 280046-280046 on Build GRCh38
rs192577707 Chr.2: 280155-280155 on Build GRCh38
rs368858369 Chr.2: 280190-280190 on Build GRCh38
rs955014190 Chr.2: 279924-279924 on Build GRCh38
rs747050332 Chr.2: 280176-280177 on Build GRCh38
rs1006202900 Chr.2: 279865-279865 on Build GRCh38
rs556409330 Chr.2: 280320-280320 on Build GRCh38
rs567075589 Chr.2: 280112-280112 on Build GRCh38
rs895624756 Chr.2: 280075-280075 on Build GRCh38
rs182206342 Chr.2: 280076-280076 on Build GRCh38
rs906964145 Chr.2: 280130-280130 on Build GRCh38
rs760713365 Chr.2: 280121-280121 on Build GRCh38
rs943380254 Chr.2: 279951-279951 on Build GRCh38
rs765218522 Chr.2: 280134-280134 on Build GRCh38
rs190887274 Chr.2: 279902-279902 on Build GRCh38
rs760800165 Chr.2: 280176-280176 on Build GRCh38
rs113471627 Chr.2: 280171-280171 on Build GRCh38
rs978439505 Chr.2: 280302-280302 on Build GRCh38
rs774853814 Chr.2: 279969-279969 on Build GRCh38
rs937088106 Chr.2: 280019-280019 on Build GRCh38
rs368472956 Chr.2: 280140-280140 on Build GRCh38
rs750421460 Chr.2: 280136-280136 on Build GRCh38
rs926944906 Chr.2: 280308-280308 on Build GRCh38
rs758119660 Chr.2: 280139-280139 on Build GRCh38
rs533396580 Chr.2: 279981-279981 on Build GRCh38
rs1016365894 Chr.2: 279880-279880 on Build GRCh38
rs373955412 Chr.2: 279918-279919 on Build GRCh38
rs996650704 Chr.2: 280200-280200 on Build GRCh38
rs147466579 Chr.2: 279900-279900 on Build GRCh38
rs983978154 Chr.2: 280310-280310 on Build GRCh38
rs754599341 Chr.2: 280146-280146 on Build GRCh38
rs536083677 Chr.2: 280227-280227 on Build GRCh38
rs894130445 Chr.2: 280084-280084 on Build GRCh38
rs759285302 Chr.2: 280043-280043 on Build GRCh38
rs766349513 Chr.2: 280185-280185 on Build GRCh38
rs771383003 Chr.2: 280166-280166 on Build GRCh38
rs1047901633 Chr.2: 280030-280030 on Build GRCh38
rs3841091 Chr.2: 279876-279877 on Build GRCh38
rs371194730 Chr.2: 280217-280217 on Build GRCh38
rs964783201 Chr.2: 279882-279882 on Build GRCh38
rs758294010 Chr.2: 280161-280161 on Build GRCh38
rs780747627 Chr.2: 280154-280154 on Build GRCh38
rs913464802 Chr.2: 279964-279964 on Build GRCh38
rs1012548736 Chr.2: 280092-280092 on Build GRCh38
rs763343598 Chr.2: 280296-280296 on Build GRCh38

View Full Product Details