Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3678341-3678847 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALLC
Entrez Gene ID
55821
Gene Name
allantoicase
Gene Aliases
ALC
UniGene
Hs.97899
Gene Chromosome Location
Chr.2: 3645368-3702671 on Build GRCh38
Transcripts
NM_018436.3 XM_011510369.2 XM_011510370.2 XM_017004495.1 XM_017004496.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6890773 c.169C>T p.P57S Chr.2: 3678552-3678552 on Build GRCh38
6999223 c.146G>T p.W49L Chr.2: 3678529-3678529 on Build GRCh38
4649995 c.99C>T p.C33C Chr.2: 3678482-3678482 on Build GRCh38
1020660 c.106G>T p.E36* Chr.2: 3678489-3678489 on Build GRCh38
2943655 c.97T>C p.C33R Chr.2: 3678480-3678480 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs115017990 Chr.2: 3678493-3678493 on Build GRCh38
rs764315712 Chr.2: 3678480-3678480 on Build GRCh38
rs1041892650 Chr.2: 3678639-3678639 on Build GRCh38
rs538862556 Chr.2: 3678406-3678406 on Build GRCh38
rs111778437 Chr.2: 3678782-3678782 on Build GRCh38
rs780785555 Chr.2: 3678502-3678502 on Build GRCh38
rs754755079 Chr.2: 3678501-3678501 on Build GRCh38
rs772288129 Chr.2: 3678474-3678474 on Build GRCh38
rs895590915 Chr.2: 3678609-3678609 on Build GRCh38
rs746201781 Chr.2: 3678554-3678554 on Build GRCh38
rs1056656128 Chr.2: 3678670-3678670 on Build GRCh38
rs895286584 Chr.2: 3678465-3678465 on Build GRCh38
rs1046574660 Chr.2: 3678629-3678629 on Build GRCh38
rs530106668 Chr.2: 3678785-3678785 on Build GRCh38
rs751702034 Chr.2: 3678736-3678736 on Build GRCh38
rs372808117 Chr.2: 3678551-3678551 on Build GRCh38
rs72769388 Chr.2: 3678374-3678374 on Build GRCh38
rs545982011 Chr.2: 3678404-3678404 on Build GRCh38
rs922017484 Chr.2: 3678494-3678494 on Build GRCh38
rs879024979 Chr.2: 3678365-3678365 on Build GRCh38
rs114821937 Chr.2: 3678811-3678811 on Build GRCh38
rs2043078 Chr.2: 3678462-3678462 on Build GRCh38
rs372653980 Chr.2: 3678590-3678590 on Build GRCh38
rs945707951 Chr.2: 3678401-3678401 on Build GRCh38
rs150700575 Chr.2: 3678684-3678684 on Build GRCh38
rs979053754 Chr.2: 3678777-3678777 on Build GRCh38
rs375401460 Chr.2: 3678533-3678533 on Build GRCh38
rs200158588 Chr.2: 3678546-3678546 on Build GRCh38
rs1042728721 Chr.2: 3678550-3678550 on Build GRCh38
rs55782533 Chr.2: 3678722-3678722 on Build GRCh38
rs371999138 Chr.2: 3678517-3678517 on Build GRCh38
rs370586019 Chr.2: 3678578-3678578 on Build GRCh38
rs142002172 Chr.2: 3678572-3678572 on Build GRCh38
rs772361133 Chr.2: 3678567-3678567 on Build GRCh38
rs1016700093 Chr.2: 3678481-3678481 on Build GRCh38
rs201623214 Chr.2: 3678503-3678503 on Build GRCh38
rs946299863 Chr.2: 3678630-3678630 on Build GRCh38
rs749845793 Chr.2: 3678440-3678440 on Build GRCh38
rs540280415 Chr.2: 3678478-3678478 on Build GRCh38
rs1001731428 Chr.2: 3678446-3678446 on Build GRCh38
rs780735056 Chr.2: 3678447-3678449 on Build GRCh38
rs759831472 Chr.2: 3678467-3678467 on Build GRCh38
rs963673921 Chr.2: 3678672-3678672 on Build GRCh38
rs764556912 Chr.2: 3678571-3678571 on Build GRCh38
rs556671557 Chr.2: 3678746-3678746 on Build GRCh38
rs967313669 Chr.2: 3678776-3678776 on Build GRCh38
rs549064425 Chr.2: 3678646-3678646 on Build GRCh38
rs2043080 Chr.2: 3678573-3678573 on Build GRCh38
rs1025449842 Chr.2: 3678489-3678489 on Build GRCh38
rs750368846 Chr.2: 3678487-3678487 on Build GRCh38
rs2043079 Chr.2: 3678500-3678500 on Build GRCh38
rs949824727 Chr.2: 3678793-3678793 on Build GRCh38
rs561864863 Chr.2: 3678479-3678479 on Build GRCh38
rs185756107 Chr.2: 3678361-3678361 on Build GRCh38
rs904255258 Chr.2: 3678445-3678445 on Build GRCh38
rs761815778 Chr.2: 3678579-3678579 on Build GRCh38
rs766295568 Chr.2: 3678589-3678589 on Build GRCh38
rs368293033 Chr.2: 3678516-3678516 on Build GRCh38
rs538562102 Chr.2: 3678697-3678697 on Build GRCh38
rs1020577113 Chr.2: 3678735-3678735 on Build GRCh38
rs767875473 Chr.2: 3678807-3678807 on Build GRCh38
rs955416603 Chr.2: 3678747-3678747 on Build GRCh38
rs1016256722 Chr.2: 3678659-3678659 on Build GRCh38
rs777323507 Chr.2: 3678532-3678532 on Build GRCh38
rs374848881 Chr.2: 3678552-3678552 on Build GRCh38
rs778223636 Chr.2: 3678420-3678420 on Build GRCh38
rs13420871 Chr.2: 3678654-3678654 on Build GRCh38
rs1025159887 Chr.2: 3678477-3678477 on Build GRCh38
rs771411217 Chr.2: 3678547-3678547 on Build GRCh38
rs373576327 Chr.2: 3678428-3678428 on Build GRCh38
rs1051645871 Chr.2: 3678375-3678375 on Build GRCh38
rs771187886 Chr.2: 3678460-3678460 on Build GRCh38
rs575920105 Chr.2: 3678392-3678392 on Build GRCh38
rs73140830 Chr.2: 3678783-3678783 on Build GRCh38
rs564378445 Chr.2: 3678409-3678409 on Build GRCh38
rs1011835167 Chr.2: 3678696-3678696 on Build GRCh38
rs539701911 Chr.2: 3678727-3678727 on Build GRCh38
rs763009149 Chr.2: 3678491-3678491 on Build GRCh38
rs963490270 Chr.2: 3678482-3678482 on Build GRCh38
rs1013680770 Chr.2: 3678624-3678624 on Build GRCh38
rs556169534 Chr.2: 3678695-3678695 on Build GRCh38
rs925897034 Chr.2: 3678779-3678779 on Build GRCh38
rs73910326 Chr.2: 3678728-3678728 on Build GRCh38

View Full Product Details