Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 3682862-3683374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ALLC
Entrez Gene ID
55821
Gene Name
allantoicase
Gene Aliases
ALC
UniGene
Hs.97899
Gene Chromosome Location
Chr.2: 3645368-3702671 on Build GRCh38
Transcripts
NM_018436.3 XM_011510369.2 XM_011510370.2 XM_017004495.1 XM_017004496.1 XM_017004497.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4094077 c.388G>C p.D130H Chr.2: 3682951-3682951 on Build GRCh38
5862488 c.508C>T p.P170S Chr.2: 3683071-3683071 on Build GRCh38
287759 c.390C>T p.D130D Chr.2: 3682953-3682953 on Build GRCh38
3962818 c.389A>T p.D130V Chr.2: 3682952-3682952 on Build GRCh38
3407850 c.446G>A p.G149D Chr.2: 3683009-3683009 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs945839207 Chr.2: 3683230-3683230 on Build GRCh38
rs946511339 Chr.2: 3683304-3683304 on Build GRCh38
rs766127038 Chr.2: 3683186-3683186 on Build GRCh38
rs6732295 Chr.2: 3683287-3683287 on Build GRCh38
rs370190116 Chr.2: 3682898-3682898 on Build GRCh38
rs201690293 Chr.2: 3682953-3682953 on Build GRCh38
rs780964237 Chr.2: 3682904-3682904 on Build GRCh38
rs975029046 Chr.2: 3683106-3683106 on Build GRCh38
rs781206931 Chr.2: 3683080-3683080 on Build GRCh38
rs375710666 Chr.2: 3682951-3682951 on Build GRCh38
rs760439229 Chr.2: 3683131-3683131 on Build GRCh38
rs1025671229 Chr.2: 3683128-3683128 on Build GRCh38
rs762923370 Chr.2: 3683013-3683013 on Build GRCh38
rs773200935 Chr.2: 3683094-3683094 on Build GRCh38
rs1042072854 Chr.2: 3682989-3682989 on Build GRCh38
rs946345614 Chr.2: 3682986-3682986 on Build GRCh38
rs778684877 Chr.2: 3683048-3683048 on Build GRCh38
rs540254607 Chr.2: 3683008-3683008 on Build GRCh38
rs779548984 Chr.2: 3683065-3683065 on Build GRCh38
rs942811124 Chr.2: 3683276-3683276 on Build GRCh38
rs761310489 Chr.2: 3682988-3682988 on Build GRCh38
rs1025464231 Chr.2: 3683047-3683047 on Build GRCh38
rs149774631 Chr.2: 3683054-3683054 on Build GRCh38
rs758534409 Chr.2: 3682899-3682899 on Build GRCh38
rs912954842 Chr.2: 3683208-3683208 on Build GRCh38
rs180717306 Chr.2: 3683297-3683297 on Build GRCh38
rs753639446 Chr.2: 3683039-3683039 on Build GRCh38
rs987183305 Chr.2: 3683199-3683199 on Build GRCh38
rs932952059 Chr.2: 3683172-3683172 on Build GRCh38
rs1042795056 Chr.2: 3683283-3683283 on Build GRCh38
rs777151820 Chr.2: 3683109-3683109 on Build GRCh38
rs752577138 Chr.2: 3683027-3683027 on Build GRCh38
rs750694861 Chr.2: 3683012-3683012 on Build GRCh38
rs77130098 Chr.2: 3683330-3683330 on Build GRCh38
rs200104628 Chr.2: 3683045-3683045 on Build GRCh38
rs562807429 Chr.2: 3683274-3683274 on Build GRCh38
rs770103349 Chr.2: 3682895-3682897 on Build GRCh38
rs757890281 Chr.2: 3683030-3683030 on Build GRCh38
rs748168086 Chr.2: 3682911-3682911 on Build GRCh38
rs745331022 Chr.2: 3682958-3682958 on Build GRCh38
rs983279523 Chr.2: 3683079-3683079 on Build GRCh38
rs113101825 Chr.2: 3683004-3683004 on Build GRCh38
rs955598051 Chr.2: 3683132-3683132 on Build GRCh38
rs374055311 Chr.2: 3683007-3683007 on Build GRCh38
rs374582443 Chr.2: 3683063-3683063 on Build GRCh38
rs771660512 Chr.2: 3682977-3682977 on Build GRCh38
rs527340401 Chr.2: 3683348-3683348 on Build GRCh38
rs911477560 Chr.2: 3683228-3683228 on Build GRCh38
rs756013424 Chr.2: 3683028-3683028 on Build GRCh38
rs759407947 Chr.2: 3683084-3683085 on Build GRCh38
rs774170405 Chr.2: 3682954-3682954 on Build GRCh38
rs764588725 Chr.2: 3682999-3682999 on Build GRCh38
rs769098148 Chr.2: 3683105-3683105 on Build GRCh38
rs775844908 Chr.2: 3682985-3682985 on Build GRCh38
rs544464131 Chr.2: 3683143-3683143 on Build GRCh38
rs770649554 Chr.2: 3682952-3682952 on Build GRCh38
rs139677162 Chr.2: 3682928-3682928 on Build GRCh38
rs773276006 Chr.2: 3682935-3682935 on Build GRCh38
rs868458289 Chr.2: 3683051-3683051 on Build GRCh38
rs987088665 Chr.2: 3683181-3683181 on Build GRCh38
rs936859564 Chr.2: 3683299-3683299 on Build GRCh38
rs963217399 Chr.2: 3683098-3683098 on Build GRCh38
rs372636443 Chr.2: 3682950-3682950 on Build GRCh38
rs766542977 Chr.2: 3683018-3683018 on Build GRCh38

View Full Product Details