Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9432415-9432964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3
Entrez Gene ID
51692
Gene Name
cleavage and polyadenylation specific factor 3
Gene Aliases
CPSF-73, CPSF73
UniGene
Hs.515972
Gene Chromosome Location
Chr.2: 9423537-9473110 on Build GRCh38
Transcripts
NM_001321833.1 NM_001321834.1 NM_001321835.1 NM_001321836.1 NM_016207.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5476406 c.508G>A p.A170T Chr.2: 9432677-9432677 on Build GRCh38
1495003 c.473A>T p.H158L Chr.2: 9432642-9432642 on Build GRCh38
6158943 c.516G>A p.V172V Chr.2: 9432685-9432685 on Build GRCh38
7238577 c.342-10C>T p.? Chr.2: 9432501-9432501 on Build GRCh38
243829 c.486C>T p.A162A Chr.2: 9432655-9432655 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768261132 Chr.2: 9432725-9432725 on Build GRCh38
rs899165730 Chr.2: 9432891-9432891 on Build GRCh38
rs768671041 Chr.2: 9432608-9432608 on Build GRCh38
rs988059694 Chr.2: 9432836-9432836 on Build GRCh38
rs745655064 Chr.2: 9432491-9432491 on Build GRCh38
rs763397155 Chr.2: 9432558-9432558 on Build GRCh38
rs766617835 Chr.2: 9432689-9432689 on Build GRCh38
rs17850770 Chr.2: 9432594-9432594 on Build GRCh38
rs546091552 Chr.2: 9432769-9432769 on Build GRCh38
rs143858016 Chr.2: 9432646-9432646 on Build GRCh38
rs767835032 Chr.2: 9432669-9432669 on Build GRCh38
rs906334208 Chr.2: 9432746-9432746 on Build GRCh38
rs199721257 Chr.2: 9432756-9432757 on Build GRCh38
rs533184931 Chr.2: 9432872-9432875 on Build GRCh38
rs778274437 Chr.2: 9432760-9432760 on Build GRCh38
rs144845873 Chr.2: 9432526-9432526 on Build GRCh38
rs752739808 Chr.2: 9432610-9432610 on Build GRCh38
rs886322217 Chr.2: 9432927-9432927 on Build GRCh38
rs914938600 Chr.2: 9432470-9432470 on Build GRCh38
rs762458018 Chr.2: 9432529-9432529 on Build GRCh38
rs774722784 Chr.2: 9432463-9432468 on Build GRCh38
rs746591102 Chr.2: 9432656-9432656 on Build GRCh38
rs910749992 Chr.2: 9432846-9432846 on Build GRCh38
rs376199240 Chr.2: 9432701-9432701 on Build GRCh38
rs200356690 Chr.2: 9432693-9432693 on Build GRCh38
rs139676758 Chr.2: 9432769-9432769 on Build GRCh38
rs772458862 Chr.2: 9432902-9432902 on Build GRCh38
rs751669456 Chr.2: 9432588-9432588 on Build GRCh38
rs761040737 Chr.2: 9432541-9432541 on Build GRCh38
rs978281959 Chr.2: 9432483-9432483 on Build GRCh38
rs906979879 Chr.2: 9432893-9432893 on Build GRCh38
rs758235541 Chr.2: 9432707-9432707 on Build GRCh38
rs1053994239 Chr.2: 9432581-9432581 on Build GRCh38
rs776322067 Chr.2: 9432666-9432666 on Build GRCh38
rs774167805 Chr.2: 9432485-9432485 on Build GRCh38
rs559362405 Chr.2: 9432759-9432759 on Build GRCh38
rs796805093 Chr.2: 9432848-9432849 on Build GRCh38
rs757249108 Chr.2: 9432613-9432613 on Build GRCh38
rs34003003 Chr.2: 9432799-9432799 on Build GRCh38
rs180778061 Chr.2: 9432848-9432848 on Build GRCh38
rs372969777 Chr.2: 9432677-9432677 on Build GRCh38
rs752882559 Chr.2: 9432835-9432835 on Build GRCh38
rs778804127 Chr.2: 9432619-9432619 on Build GRCh38
rs768011530 Chr.2: 9432515-9432515 on Build GRCh38
rs548820155 Chr.2: 9432676-9432676 on Build GRCh38
rs768331728 Chr.2: 9432657-9432657 on Build GRCh38
rs763418256 Chr.2: 9432683-9432683 on Build GRCh38
rs755119135 Chr.2: 9432591-9432591 on Build GRCh38
rs771765862 Chr.2: 9432503-9432503 on Build GRCh38
rs555398264 Chr.2: 9432643-9432643 on Build GRCh38
rs929309205 Chr.2: 9432901-9432901 on Build GRCh38
rs954698868 Chr.2: 9432823-9432823 on Build GRCh38
rs945086955 Chr.2: 9432471-9432471 on Build GRCh38
rs916760148 Chr.2: 9432858-9432858 on Build GRCh38
rs570669141 Chr.2: 9432867-9432867 on Build GRCh38
rs773798533 Chr.2: 9432682-9432682 on Build GRCh38
rs966399697 Chr.2: 9432800-9432800 on Build GRCh38
rs989418190 Chr.2: 9432794-9432794 on Build GRCh38
rs974819577 Chr.2: 9432774-9432774 on Build GRCh38
rs375185961 Chr.2: 9432827-9432827 on Build GRCh38
rs369409765 Chr.2: 9432705-9432705 on Build GRCh38
rs751832298 Chr.2: 9432691-9432691 on Build GRCh38
rs759900458 Chr.2: 9432470-9432470 on Build GRCh38
rs542697035 Chr.2: 9432569-9432569 on Build GRCh38
rs539578015 Chr.2: 9432932-9432932 on Build GRCh38
rs923288361 Chr.2: 9432455-9432455 on Build GRCh38
rs1044149665 Chr.2: 9432874-9432874 on Build GRCh38
rs924794992 Chr.2: 9432520-9432520 on Build GRCh38
rs771696870 Chr.2: 9432644-9432644 on Build GRCh38
rs185191169 Chr.2: 9432849-9432849 on Build GRCh38
rs561728937 Chr.2: 9432828-9432828 on Build GRCh38
rs201017284 Chr.2: 9432586-9432586 on Build GRCh38
rs375046889 Chr.2: 9432510-9432510 on Build GRCh38
rs977834749 Chr.2: 9432825-9432825 on Build GRCh38
rs544072348 Chr.2: 9432669-9432669 on Build GRCh38
rs573020802 Chr.2: 9432706-9432706 on Build GRCh38
rs938338729 Chr.2: 9432899-9432899 on Build GRCh38
rs528564401 Chr.2: 9432806-9432806 on Build GRCh38
rs369380019 Chr.2: 9432544-9432544 on Build GRCh38
rs746786393 Chr.2: 9432716-9432716 on Build GRCh38
rs12478467 Chr.2: 9432759-9432759 on Build GRCh38
rs906886599 Chr.2: 9432702-9432702 on Build GRCh38
rs964736990 Chr.2: 9432773-9432773 on Build GRCh38
rs752829836 Chr.2: 9432695-9432695 on Build GRCh38
rs948427899 Chr.2: 9432860-9432860 on Build GRCh38
rs950703808 Chr.2: 9432732-9432732 on Build GRCh38
rs943346113 Chr.2: 9432655-9432655 on Build GRCh38
rs959261671 Chr.2: 9432776-9432776 on Build GRCh38
rs767773242 Chr.2: 9432694-9432694 on Build GRCh38
rs774944629 Chr.2: 9432509-9432509 on Build GRCh38

View Full Product Details