Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 9433432-9433930 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CPSF3
Entrez Gene ID
51692
Gene Name
cleavage and polyadenylation specific factor 3
Gene Aliases
CPSF-73, CPSF73
UniGene
Hs.515972
Gene Chromosome Location
Chr.2: 9423537-9473110 on Build GRCh38
Transcripts
NM_001321833.1 NM_001321834.1 NM_001321835.1 NM_001321836.1 NM_016207.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6258920 c.525G>T p.L175F Chr.2: 9433876-9433876 on Build GRCh38
6240997 c.545G>T p.R182I Chr.2: 9433896-9433896 on Build GRCh38
4916581 c.551A>G p.E184G Chr.2: 9433902-9433902 on Build GRCh38
5449764 c.578T>A p.I193N Chr.2: 9433929-9433929 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs566650794 Chr.2: 9433651-9433651 on Build GRCh38
rs995238830 Chr.2: 9433776-9433776 on Build GRCh38
rs549066445 Chr.2: 9433693-9433693 on Build GRCh38
rs79897070 Chr.2: 9433533-9433533 on Build GRCh38
rs371153328 Chr.2: 9433868-9433868 on Build GRCh38
rs369594397 Chr.2: 9433637-9433637 on Build GRCh38
rs546956326 Chr.2: 9433617-9433617 on Build GRCh38
rs566439049 Chr.2: 9433506-9433506 on Build GRCh38
rs377648120 Chr.2: 9433626-9433626 on Build GRCh38
rs911826448 Chr.2: 9433788-9433788 on Build GRCh38
rs761479204 Chr.2: 9433829-9433837 on Build GRCh38
rs753732258 Chr.2: 9433826-9433827 on Build GRCh38
rs758290747 Chr.2: 9433593-9433593 on Build GRCh38
rs747675008 Chr.2: 9433839-9433839 on Build GRCh38
rs75117129 Chr.2: 9433457-9433457 on Build GRCh38
rs755298404 Chr.2: 9433629-9433631 on Build GRCh38
rs527262597 Chr.2: 9433699-9433699 on Build GRCh38
rs1052516770 Chr.2: 9433461-9433461 on Build GRCh38
rs774642292 Chr.2: 9433856-9433856 on Build GRCh38
rs4668620 Chr.2: 9433665-9433665 on Build GRCh38
rs1052736793 Chr.2: 9433815-9433815 on Build GRCh38
rs902369397 Chr.2: 9433649-9433649 on Build GRCh38
rs114373351 Chr.2: 9433452-9433452 on Build GRCh38
rs1043564957 Chr.2: 9433603-9433603 on Build GRCh38
rs10209595 Chr.2: 9433537-9433537 on Build GRCh38
rs999789063 Chr.2: 9433460-9433460 on Build GRCh38
rs767642975 Chr.2: 9433885-9433885 on Build GRCh38
rs372570198 Chr.2: 9433845-9433845 on Build GRCh38
rs183416767 Chr.2: 9433598-9433598 on Build GRCh38
rs370880550 Chr.2: 9433784-9433784 on Build GRCh38
rs780980419 Chr.2: 9433829-9433829 on Build GRCh38
rs1035281108 Chr.2: 9433811-9433811 on Build GRCh38
rs564357205 Chr.2: 9433550-9433550 on Build GRCh38
rs969600704 Chr.2: 9433527-9433527 on Build GRCh38
rs920829581 Chr.2: 9433504-9433504 on Build GRCh38
rs771256782 Chr.2: 9433482-9433482 on Build GRCh38
rs1055174902 Chr.2: 9433545-9433545 on Build GRCh38
rs984074332 Chr.2: 9433810-9433810 on Build GRCh38
rs950945658 Chr.2: 9433808-9433808 on Build GRCh38
rs962449886 Chr.2: 9433698-9433698 on Build GRCh38
rs759928669 Chr.2: 9433878-9433878 on Build GRCh38
rs770394516 Chr.2: 9433809-9433809 on Build GRCh38
rs769363289 Chr.2: 9433844-9433844 on Build GRCh38
rs893915807 Chr.2: 9433470-9433470 on Build GRCh38
rs1051056293 Chr.2: 9433664-9433664 on Build GRCh38
rs76962241 Chr.2: 9433853-9433853 on Build GRCh38
rs1051611067 Chr.2: 9433776-9433777 on Build GRCh38
rs960671994 Chr.2: 9433834-9433834 on Build GRCh38
rs550910793 Chr.2: 9433525-9433525 on Build GRCh38
rs374309916 Chr.2: 9433551-9433551 on Build GRCh38
rs116032989 Chr.2: 9433697-9433697 on Build GRCh38
rs893850670 Chr.2: 9433819-9433819 on Build GRCh38
rs931874310 Chr.2: 9433754-9433754 on Build GRCh38
rs773890713 Chr.2: 9433850-9433850 on Build GRCh38
rs537803125 Chr.2: 9433781-9433781 on Build GRCh38
rs927944303 Chr.2: 9433526-9433526 on Build GRCh38
rs1035295159 Chr.2: 9433556-9433556 on Build GRCh38
rs1028234097 Chr.2: 9433782-9433782 on Build GRCh38
rs951568366 Chr.2: 9433584-9433586 on Build GRCh38

View Full Product Details