Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10058747-10059181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYS1
Entrez Gene ID
192668
Gene Name
cystin 1
Gene Aliases
-
UniGene
Hs.27092
Gene Chromosome Location
Chr.2: 10056799-10080411 on Build GRCh38
Transcripts
NM_001037160.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5859772 c.452G>A p.S151N Chr.2: 10058878-10058878 on Build GRCh38
3894390 c.436G>A p.E146K Chr.2: 10058894-10058894 on Build GRCh38
3564359 c.463G>A p.E155K Chr.2: 10058867-10058867 on Build GRCh38
5354250 c.449C>T p.A150V Chr.2: 10058881-10058881 on Build GRCh38
5034044 c.437A>G p.E146G Chr.2: 10058893-10058893 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs930665069 Chr.2: 10059119-10059119 on Build GRCh38
rs372923434 Chr.2: 10058973-10058973 on Build GRCh38
rs944818562 Chr.2: 10059085-10059085 on Build GRCh38
rs867199182 Chr.2: 10059111-10059111 on Build GRCh38
rs746658735 Chr.2: 10058877-10058877 on Build GRCh38
rs755442739 Chr.2: 10058822-10058822 on Build GRCh38
rs757645657 Chr.2: 10058878-10058878 on Build GRCh38
rs967752406 Chr.2: 10059022-10059022 on Build GRCh38
rs760196585 Chr.2: 10058977-10058977 on Build GRCh38
rs772937929 Chr.2: 10058840-10058840 on Build GRCh38
rs989011052 Chr.2: 10059068-10059068 on Build GRCh38
rs539375502 Chr.2: 10059103-10059103 on Build GRCh38
rs912049007 Chr.2: 10059069-10059069 on Build GRCh38
rs774180325 Chr.2: 10058855-10058855 on Build GRCh38
rs1000035721 Chr.2: 10058890-10058890 on Build GRCh38
rs187408318 Chr.2: 10059097-10059097 on Build GRCh38
rs746101333 Chr.2: 10058898-10058898 on Build GRCh38
rs576887345 Chr.2: 10059074-10059074 on Build GRCh38
rs759217282 Chr.2: 10058858-10058858 on Build GRCh38
rs200653727 Chr.2: 10058846-10058846 on Build GRCh38
rs367569043 Chr.2: 10059004-10059004 on Build GRCh38
rs535291645 Chr.2: 10058930-10058931 on Build GRCh38
rs35785395 Chr.2: 10059063-10059064 on Build GRCh38
rs771849443 Chr.2: 10058975-10058975 on Build GRCh38
rs780173331 Chr.2: 10058923-10058923 on Build GRCh38
rs201762569 Chr.2: 10059041-10059042 on Build GRCh38
rs370575806 Chr.2: 10059042-10059043 on Build GRCh38
rs754388860 Chr.2: 10058990-10058990 on Build GRCh38
rs767151766 Chr.2: 10058859-10058859 on Build GRCh38
rs772042428 Chr.2: 10059012-10059012 on Build GRCh38
rs774232020 Chr.2: 10058957-10058957 on Build GRCh38
rs137932835 Chr.2: 10058955-10058955 on Build GRCh38
rs374907171 Chr.2: 10058976-10058976 on Build GRCh38
rs770342990 Chr.2: 10058907-10058907 on Build GRCh38
rs868468223 Chr.2: 10058805-10058805 on Build GRCh38
rs775816091 Chr.2: 10058889-10058889 on Build GRCh38
rs926193782 Chr.2: 10059041-10059041 on Build GRCh38
rs932234512 Chr.2: 10059096-10059096 on Build GRCh38
rs765242639 Chr.2: 10058995-10058995 on Build GRCh38
rs769765865 Chr.2: 10058835-10058835 on Build GRCh38
rs746963859 Chr.2: 10059033-10059033 on Build GRCh38
rs749140889 Chr.2: 10058945-10058945 on Build GRCh38
rs747097449 Chr.2: 10058926-10058926 on Build GRCh38
rs942179108 Chr.2: 10058776-10058776 on Build GRCh38
rs571719047 Chr.2: 10059077-10059077 on Build GRCh38
rs116762213 Chr.2: 10059123-10059123 on Build GRCh38
rs752187460 Chr.2: 10058866-10058866 on Build GRCh38
rs368468153 Chr.2: 10058873-10058873 on Build GRCh38
rs541619533 Chr.2: 10059142-10059142 on Build GRCh38
rs141827259 Chr.2: 10059097-10059102 on Build GRCh38
rs773192474 Chr.2: 10059014-10059014 on Build GRCh38
rs766174689 Chr.2: 10058854-10058854 on Build GRCh38
rs772158387 Chr.2: 10058815-10058815 on Build GRCh38
rs534831565 Chr.2: 10058847-10058847 on Build GRCh38
rs995219684 Chr.2: 10058962-10058962 on Build GRCh38
rs537134600 Chr.2: 10059001-10059001 on Build GRCh38
rs763468447 Chr.2: 10058985-10058985 on Build GRCh38
rs867185221 Chr.2: 10058899-10058899 on Build GRCh38
rs773169801 Chr.2: 10058942-10058942 on Build GRCh38
rs572624641 Chr.2: 10059129-10059129 on Build GRCh38
rs867721308 Chr.2: 10058941-10058941 on Build GRCh38
rs886392932 Chr.2: 10059136-10059136 on Build GRCh38
rs143968577 Chr.2: 10058870-10058870 on Build GRCh38
rs371413709 Chr.2: 10059005-10059005 on Build GRCh38
rs776446541 Chr.2: 10059104-10059104 on Build GRCh38
rs778980007 Chr.2: 10058834-10058834 on Build GRCh38
rs375786169 Chr.2: 10058808-10058808 on Build GRCh38
rs755249656 Chr.2: 10058768-10058768 on Build GRCh38
rs934832961 Chr.2: 10058833-10058833 on Build GRCh38
rs907607556 Chr.2: 10059034-10059034 on Build GRCh38
rs979482596 Chr.2: 10059037-10059037 on Build GRCh38
rs561287754 Chr.2: 10059159-10059159 on Build GRCh38
rs557042668 Chr.2: 10059042-10059042 on Build GRCh38
rs779196794 Chr.2: 10058881-10058881 on Build GRCh38
rs769667856 Chr.2: 10058931-10058931 on Build GRCh38

View Full Product Details