Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 10722733-10723241 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1C2
Entrez Gene ID
245973
Gene Name
ATPase H+ transporting V1 subunit C2
Gene Aliases
ATP6C2, VMA5
UniGene
Hs.580464
Gene Chromosome Location
Chr.2: 10720974-10785110 on Build GRCh38
Transcripts
NM_001039362.1 NM_144583.3 XM_011510339.2 XM_011510340.2 XM_011510341.2 XM_017003745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
224645 c.26C>T p.A9V Chr.2: 10722875-10722875 on Build GRCh38
2982119 c.113C>A p.A38D Chr.2: 10722962-10722962 on Build GRCh38
3565041 c.68A>C p.N23T Chr.2: 10722917-10722917 on Build GRCh38
4084025 c.32G>T p.G11V Chr.2: 10722881-10722881 on Build GRCh38
6608135 c.40G>A p.E14K Chr.2: 10722889-10722889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542240839 Chr.2: 10723046-10723046 on Build GRCh38
rs568764822 Chr.2: 10723163-10723163 on Build GRCh38
rs770358735 Chr.2: 10723023-10723035 on Build GRCh38
rs779256028 Chr.2: 10722998-10722998 on Build GRCh38
rs759296963 Chr.2: 10722938-10722938 on Build GRCh38
rs1010975066 Chr.2: 10723069-10723069 on Build GRCh38
rs752201407 Chr.2: 10722953-10722953 on Build GRCh38
rs747097954 Chr.2: 10723021-10723021 on Build GRCh38
rs115461273 Chr.2: 10722930-10722930 on Build GRCh38
rs766448798 Chr.2: 10722775-10722775 on Build GRCh38
rs973185882 Chr.2: 10723167-10723167 on Build GRCh38
rs76168119 Chr.2: 10722851-10722851 on Build GRCh38
rs774227825 Chr.2: 10722931-10722931 on Build GRCh38
rs998190926 Chr.2: 10723132-10723132 on Build GRCh38
rs767145605 Chr.2: 10722801-10722801 on Build GRCh38
rs1052196548 Chr.2: 10722947-10722947 on Build GRCh38
rs1002115404 Chr.2: 10723214-10723214 on Build GRCh38
rs764576345 Chr.2: 10722959-10722959 on Build GRCh38
rs1034335798 Chr.2: 10722901-10722901 on Build GRCh38
rs560275053 Chr.2: 10723074-10723074 on Build GRCh38
rs774510397 Chr.2: 10722796-10722796 on Build GRCh38
rs751660891 Chr.2: 10722794-10722794 on Build GRCh38
rs532972850 Chr.2: 10722802-10722802 on Build GRCh38
rs760204609 Chr.2: 10722954-10722954 on Build GRCh38
rs747060427 Chr.2: 10722790-10722790 on Build GRCh38
rs539943383 Chr.2: 10722882-10722882 on Build GRCh38
rs758640245 Chr.2: 10723013-10723013 on Build GRCh38
rs762046882 Chr.2: 10723021-10723022 on Build GRCh38
rs148726148 Chr.2: 10723135-10723135 on Build GRCh38
rs769698947 Chr.2: 10723125-10723125 on Build GRCh38
rs973080905 Chr.2: 10722821-10722821 on Build GRCh38
rs1043539731 Chr.2: 10723194-10723194 on Build GRCh38
rs777098102 Chr.2: 10723019-10723020 on Build GRCh38
rs893905462 Chr.2: 10722977-10722977 on Build GRCh38
rs962522809 Chr.2: 10722955-10722955 on Build GRCh38
rs775462278 Chr.2: 10722871-10722871 on Build GRCh38
rs969466297 Chr.2: 10723195-10723195 on Build GRCh38
rs531905196 Chr.2: 10723138-10723138 on Build GRCh38
rs369200390 Chr.2: 10722807-10722807 on Build GRCh38
rs768401093 Chr.2: 10722951-10722951 on Build GRCh38
rs1050650951 Chr.2: 10723129-10723129 on Build GRCh38
rs778324768 Chr.2: 10722830-10722830 on Build GRCh38
rs147651089 Chr.2: 10722939-10722939 on Build GRCh38
rs144221363 Chr.2: 10723009-10723009 on Build GRCh38
rs1017087355 Chr.2: 10723153-10723153 on Build GRCh38
rs73914806 Chr.2: 10723124-10723124 on Build GRCh38
rs116123470 Chr.2: 10723099-10723099 on Build GRCh38
rs574790671 Chr.2: 10723185-10723185 on Build GRCh38
rs10929693 Chr.2: 10723141-10723141 on Build GRCh38
rs199648132 Chr.2: 10722879-10722879 on Build GRCh38
rs772213749 Chr.2: 10722855-10722855 on Build GRCh38
rs1009369692 Chr.2: 10722783-10722786 on Build GRCh38
rs757614677 Chr.2: 10722842-10722842 on Build GRCh38
rs972231695 Chr.2: 10723034-10723034 on Build GRCh38
rs756508023 Chr.2: 10722952-10722952 on Build GRCh38
rs1018696128 Chr.2: 10722762-10722762 on Build GRCh38
rs149963534 Chr.2: 10722831-10722831 on Build GRCh38
rs200699798 Chr.2: 10722854-10722854 on Build GRCh38
rs201851019 Chr.2: 10722911-10722911 on Build GRCh38
rs372789883 Chr.2: 10722818-10722818 on Build GRCh38
rs757706515 Chr.2: 10722973-10722973 on Build GRCh38
rs888769264 Chr.2: 10722760-10722760 on Build GRCh38
rs769667795 Chr.2: 10722897-10722897 on Build GRCh38
rs779166260 Chr.2: 10722847-10722847 on Build GRCh38
rs780300669 Chr.2: 10723020-10723020 on Build GRCh38
rs200657466 Chr.2: 10722921-10722921 on Build GRCh38
rs374264130 Chr.2: 10722961-10722961 on Build GRCh38
rs1009832698 Chr.2: 10723193-10723193 on Build GRCh38
rs962860907 Chr.2: 10723165-10723165 on Build GRCh38

View Full Product Details