Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 11140428-11140933 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf50
Entrez Gene ID
130813
Gene Name
chromosome 2 open reading frame 50
Gene Aliases
-
UniGene
Hs.406894
Gene Chromosome Location
Chr.2: 11133053-11154564 on Build GRCh38
Transcripts
NM_182500.2 XM_006711857.2 XM_006711858.2 XM_006711859.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7152438 c.276C>T p.F92F Chr.2: 11140528-11140528 on Build GRCh38
4396919 c.270G>A p.W90* Chr.2: 11140522-11140522 on Build GRCh38
291528 c.274T>C p.F92L Chr.2: 11140526-11140526 on Build GRCh38
5508276 c.279G>A p.L93L Chr.2: 11140531-11140531 on Build GRCh38
159585 c.279G>C p.L93L Chr.2: 11140531-11140531 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs779504089 Chr.2: 11140493-11140493 on Build GRCh38
rs866415827 Chr.2: 11140514-11140514 on Build GRCh38
rs751566165 Chr.2: 11140580-11140580 on Build GRCh38
rs753392678 Chr.2: 11140592-11140592 on Build GRCh38
rs980892316 Chr.2: 11140841-11140841 on Build GRCh38
rs752546164 Chr.2: 11140587-11140587 on Build GRCh38
rs375714786 Chr.2: 11140464-11140464 on Build GRCh38
rs866749396 Chr.2: 11140638-11140638 on Build GRCh38
rs1029743570 Chr.2: 11140665-11140665 on Build GRCh38
rs1008797601 Chr.2: 11140705-11140705 on Build GRCh38
rs371275514 Chr.2: 11140586-11140586 on Build GRCh38
rs1050355820 Chr.2: 11140662-11140662 on Build GRCh38
rs755825528 Chr.2: 11140485-11140485 on Build GRCh38
rs897452710 Chr.2: 11140741-11140741 on Build GRCh38
rs374615352 Chr.2: 11140707-11140707 on Build GRCh38
rs776789494 Chr.2: 11140551-11140551 on Build GRCh38
rs761986802 Chr.2: 11140571-11140571 on Build GRCh38
rs901649035 Chr.2: 11140656-11140656 on Build GRCh38
rs564417093 Chr.2: 11140666-11140666 on Build GRCh38
rs756195313 Chr.2: 11140543-11140543 on Build GRCh38
rs904388074 Chr.2: 11140611-11140611 on Build GRCh38
rs960963132 Chr.2: 11140865-11140865 on Build GRCh38
rs755914283 Chr.2: 11140589-11140589 on Build GRCh38
rs542176930 Chr.2: 11140550-11140550 on Build GRCh38
rs952270447 Chr.2: 11140833-11140833 on Build GRCh38
rs76773051 Chr.2: 11140883-11140883 on Build GRCh38
rs957634620 Chr.2: 11140766-11140766 on Build GRCh38
rs760950239 Chr.2: 11140540-11140540 on Build GRCh38
rs770296592 Chr.2: 11140491-11140491 on Build GRCh38
rs1023100261 Chr.2: 11140852-11140852 on Build GRCh38
rs370347719 Chr.2: 11140486-11140486 on Build GRCh38
rs754348344 Chr.2: 11140535-11140535 on Build GRCh38
rs746327149 Chr.2: 11140526-11140526 on Build GRCh38
rs752834358 Chr.2: 11140478-11140479 on Build GRCh38
rs971501434 Chr.2: 11140853-11140853 on Build GRCh38
rs775815009 Chr.2: 11140539-11140539 on Build GRCh38
rs376692211 Chr.2: 11140577-11140577 on Build GRCh38
rs76093388 Chr.2: 11140864-11140864 on Build GRCh38
rs531881704 Chr.2: 11140821-11140821 on Build GRCh38
rs991670835 Chr.2: 11140797-11140797 on Build GRCh38
rs374705159 Chr.2: 11140597-11140597 on Build GRCh38
rs572226882 Chr.2: 11140620-11140620 on Build GRCh38
rs1018627851 Chr.2: 11140734-11140734 on Build GRCh38
rs777590399 Chr.2: 11140590-11140590 on Build GRCh38
rs79424178 Chr.2: 11140652-11140652 on Build GRCh38
rs1027816091 Chr.2: 11140784-11140784 on Build GRCh38
rs866633359 Chr.2: 11140876-11140876 on Build GRCh38
rs554133764 Chr.2: 11140593-11140593 on Build GRCh38
rs998853552 Chr.2: 11140671-11140671 on Build GRCh38
rs755500696 Chr.2: 11140703-11140703 on Build GRCh38
rs981638966 Chr.2: 11140863-11140863 on Build GRCh38
rs372903703 Chr.2: 11140562-11140562 on Build GRCh38
rs746440743 Chr.2: 11140599-11140599 on Build GRCh38
rs547293375 Chr.2: 11140871-11140871 on Build GRCh38
rs779205999 Chr.2: 11140800-11140800 on Build GRCh38
rs993295769 Chr.2: 11140776-11140776 on Build GRCh38

View Full Product Details