Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 16551619-16552127 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM49A
Entrez Gene ID
81553
Gene Name
family with sequence similarity 49 member A
Gene Aliases
-
UniGene
Hs.467769
Gene Chromosome Location
Chr.2: 16549459-16665866 on Build GRCh38
Transcripts
NM_030797.3 XM_005262630.3 XM_006712108.3

SNP Detail

SNP ID Chromosome Location
rs10172734 Chr.2: 16551786-16551786 on Build GRCh38
rs556325386 Chr.2: 16551693-16551693 on Build GRCh38
rs946775056 Chr.2: 16551748-16551748 on Build GRCh38
rs915336631 Chr.2: 16551736-16551736 on Build GRCh38
rs994680211 Chr.2: 16551658-16551658 on Build GRCh38
rs925115161 Chr.2: 16551854-16551854 on Build GRCh38
rs1042552640 Chr.2: 16551753-16551753 on Build GRCh38
rs767225282 Chr.2: 16551937-16551937 on Build GRCh38
rs994564074 Chr.2: 16551884-16551884 on Build GRCh38
rs956515912 Chr.2: 16552062-16552062 on Build GRCh38
rs535763140 Chr.2: 16551730-16551730 on Build GRCh38
rs140821557 Chr.2: 16552023-16552023 on Build GRCh38
rs543614969 Chr.2: 16551903-16551903 on Build GRCh38
rs1026174072 Chr.2: 16551902-16551902 on Build GRCh38
rs898633472 Chr.2: 16551953-16551954 on Build GRCh38
rs987386067 Chr.2: 16551688-16551688 on Build GRCh38
rs915220256 Chr.2: 16551979-16551979 on Build GRCh38
rs115727761 Chr.2: 16551668-16551668 on Build GRCh38
rs979185263 Chr.2: 16551790-16551790 on Build GRCh38
rs551855859 Chr.2: 16551774-16551774 on Build GRCh38
rs964953856 Chr.2: 16551953-16551953 on Build GRCh38
rs34999982 Chr.2: 16551950-16551951 on Build GRCh38
rs964741171 Chr.2: 16551711-16551711 on Build GRCh38
rs1009261740 Chr.2: 16551920-16551920 on Build GRCh38
rs924094290 Chr.2: 16551761-16551761 on Build GRCh38
rs1026294055 Chr.2: 16551961-16551961 on Build GRCh38
rs750668896 Chr.2: 16551694-16551695 on Build GRCh38
rs903727113 Chr.2: 16551662-16551662 on Build GRCh38
rs1030135350 Chr.2: 16551694-16551694 on Build GRCh38
rs1006974196 Chr.2: 16551935-16551935 on Build GRCh38
rs1056710443 Chr.2: 16551795-16551795 on Build GRCh38
rs752425578 Chr.2: 16551957-16551957 on Build GRCh38
rs933256199 Chr.2: 16551881-16551881 on Build GRCh38
rs755938738 Chr.2: 16552052-16552052 on Build GRCh38
rs935241260 Chr.2: 16551665-16551665 on Build GRCh38
rs979111815 Chr.2: 16552010-16552010 on Build GRCh38
rs777669607 Chr.2: 16552056-16552056 on Build GRCh38
rs1052752258 Chr.2: 16551870-16551870 on Build GRCh38
rs974729787 Chr.2: 16551963-16551963 on Build GRCh38
rs150555071 Chr.2: 16551877-16551877 on Build GRCh38
rs574221205 Chr.2: 16552007-16552007 on Build GRCh38
rs962908029 Chr.2: 16551732-16551732 on Build GRCh38
rs530479568 Chr.2: 16551714-16551714 on Build GRCh38
rs939532486 Chr.2: 16552094-16552094 on Build GRCh38
rs924139575 Chr.2: 16552090-16552090 on Build GRCh38
rs1044051369 Chr.2: 16551863-16551863 on Build GRCh38
rs141752407 Chr.2: 16551756-16551756 on Build GRCh38
rs968128483 Chr.2: 16552009-16552009 on Build GRCh38
rs1014283011 Chr.2: 16551982-16551982 on Build GRCh38
rs558282563 Chr.2: 16551844-16551844 on Build GRCh38
rs1023293247 Chr.2: 16552005-16552005 on Build GRCh38
rs889858502 Chr.2: 16551715-16551715 on Build GRCh38
rs569809592 Chr.2: 16551726-16551726 on Build GRCh38
rs994362120 Chr.2: 16551954-16551954 on Build GRCh38
rs181447569 Chr.2: 16551817-16551817 on Build GRCh38
rs950709452 Chr.2: 16551678-16551678 on Build GRCh38
rs532265042 Chr.2: 16551642-16551642 on Build GRCh38
rs1025905386 Chr.2: 16551749-16551749 on Build GRCh38
rs1007006719 Chr.2: 16551718-16551718 on Build GRCh38
rs895510489 Chr.2: 16551799-16551799 on Build GRCh38
rs935172557 Chr.2: 16551867-16551867 on Build GRCh38
rs563920401 Chr.2: 16551919-16551919 on Build GRCh38

View Full Product Details