Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 16552404-16552910 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM49A
Entrez Gene ID
81553
Gene Name
family with sequence similarity 49 member A
Gene Aliases
-
UniGene
Hs.467769
Gene Chromosome Location
Chr.2: 16549459-16665866 on Build GRCh38
Transcripts
NM_030797.3 XM_005262630.3 XM_006712108.3

SNP Detail

SNP ID Chromosome Location
rs147901216 Chr.2: 16552498-16552498 on Build GRCh38
rs930069625 Chr.2: 16552482-16552482 on Build GRCh38
rs78495239 Chr.2: 16552700-16552700 on Build GRCh38
rs568794036 Chr.2: 16552868-16552868 on Build GRCh38
rs747037538 Chr.2: 16552457-16552457 on Build GRCh38
rs1031959812 Chr.2: 16552630-16552630 on Build GRCh38
rs999490355 Chr.2: 16552717-16552717 on Build GRCh38
rs188384201 Chr.2: 16552674-16552674 on Build GRCh38
rs1023831542 Chr.2: 16552807-16552807 on Build GRCh38
rs7552 Chr.2: 16552660-16552660 on Build GRCh38
rs13404748 Chr.2: 16552885-16552885 on Build GRCh38
rs974306797 Chr.2: 16552648-16552648 on Build GRCh38
rs190112266 Chr.2: 16552520-16552520 on Build GRCh38
rs936919823 Chr.2: 16552848-16552848 on Build GRCh38
rs28490426 Chr.2: 16552707-16552707 on Build GRCh38
rs900320744 Chr.2: 16552693-16552693 on Build GRCh38
rs537571322 Chr.2: 16552699-16552699 on Build GRCh38
rs944431425 Chr.2: 16552688-16552688 on Build GRCh38
rs905433624 Chr.2: 16552594-16552594 on Build GRCh38
rs960839553 Chr.2: 16552744-16552744 on Build GRCh38
rs1045266185 Chr.2: 16552573-16552573 on Build GRCh38
rs924383055 Chr.2: 16552862-16552862 on Build GRCh38
rs992350773 Chr.2: 16552601-16552601 on Build GRCh38
rs767999050 Chr.2: 16552653-16552653 on Build GRCh38
rs186132487 Chr.2: 16552445-16552445 on Build GRCh38
rs916710277 Chr.2: 16552620-16552620 on Build GRCh38
rs190729471 Chr.2: 16552675-16552675 on Build GRCh38
rs920175910 Chr.2: 16552833-16552833 on Build GRCh38
rs930290951 Chr.2: 16552665-16552665 on Build GRCh38
rs949519543 Chr.2: 16552822-16552822 on Build GRCh38
rs139923046 Chr.2: 16552689-16552689 on Build GRCh38
rs942655827 Chr.2: 16552427-16552427 on Build GRCh38
rs919955224 Chr.2: 16552450-16552450 on Build GRCh38
rs558546769 Chr.2: 16552808-16552808 on Build GRCh38
rs542891822 Chr.2: 16552809-16552809 on Build GRCh38
rs896614688 Chr.2: 16552548-16552548 on Build GRCh38
rs918012448 Chr.2: 16552813-16552813 on Build GRCh38
rs1031390911 Chr.2: 16552727-16552727 on Build GRCh38
rs781353218 Chr.2: 16552513-16552513 on Build GRCh38
rs929971153 Chr.2: 16552772-16552772 on Build GRCh38
rs141668569 Chr.2: 16552668-16552671 on Build GRCh38
rs992633199 Chr.2: 16552790-16552790 on Build GRCh38
rs1014180099 Chr.2: 16552549-16552549 on Build GRCh38
rs1045197305 Chr.2: 16552827-16552827 on Build GRCh38
rs559615335 Chr.2: 16552854-16552854 on Build GRCh38
rs948306301 Chr.2: 16552632-16552632 on Build GRCh38

View Full Product Details