Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 23707118-23707629 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD2B
Entrez Gene ID
54454
Gene Name
ATPase family, AAA domain containing 2B
Gene Aliases
-
UniGene
Hs.467862
Gene Chromosome Location
Chr.2: 23689710-23927114 on Build GRCh38
Transcripts
XM_006712030.3 NM_052920.1 XM_006711929.3 XM_011532501.2 XM_017003264.1

Gene Symbol
KLHL29
Entrez Gene ID
114818
Gene Name
kelch like family member 29
Gene Aliases
KBTBD9
UniGene
Hs.130593
Gene Chromosome Location
Chr.2: 23385419-23708613 on Build GRCh38
Transcripts
XM_006712030.3 NM_052920.1 XM_006711929.3 XM_011532501.2 XM_017003264.1

SNP Detail

SNP ID Chromosome Location
rs763804939 Chr.2: 23707326-23707326 on Build GRCh38
rs191482944 Chr.2: 23707272-23707272 on Build GRCh38
rs576077256 Chr.2: 23707189-23707189 on Build GRCh38
rs908286085 Chr.2: 23707231-23707231 on Build GRCh38
rs912801697 Chr.2: 23707554-23707554 on Build GRCh38
rs980658655 Chr.2: 23707169-23707169 on Build GRCh38
rs934249611 Chr.2: 23707503-23707503 on Build GRCh38
rs932333780 Chr.2: 23707258-23707258 on Build GRCh38
rs537321093 Chr.2: 23707225-23707225 on Build GRCh38
rs979167324 Chr.2: 23707457-23707457 on Build GRCh38
rs1037986172 Chr.2: 23707255-23707255 on Build GRCh38
rs924992332 Chr.2: 23707461-23707461 on Build GRCh38
rs560696209 Chr.2: 23707155-23707155 on Build GRCh38
rs960695668 Chr.2: 23707424-23707424 on Build GRCh38
rs546424750 Chr.2: 23707329-23707329 on Build GRCh38
rs114880940 Chr.2: 23707482-23707482 on Build GRCh38
rs542784086 Chr.2: 23707328-23707328 on Build GRCh38
rs562963431 Chr.2: 23707409-23707409 on Build GRCh38
rs201028215 Chr.2: 23707453-23707453 on Build GRCh38
rs945014195 Chr.2: 23707440-23707442 on Build GRCh38
rs1011689552 Chr.2: 23707334-23707334 on Build GRCh38
rs992392440 Chr.2: 23707462-23707462 on Build GRCh38
rs1017008966 Chr.2: 23707324-23707324 on Build GRCh38
rs980597083 Chr.2: 23707339-23707339 on Build GRCh38
rs367616671 Chr.2: 23707540-23707540 on Build GRCh38
rs908399004 Chr.2: 23707467-23707467 on Build GRCh38
rs904712247 Chr.2: 23707240-23707240 on Build GRCh38
rs960653094 Chr.2: 23707417-23707417 on Build GRCh38
rs559235568 Chr.2: 23707226-23707226 on Build GRCh38
rs577331090 Chr.2: 23707236-23707236 on Build GRCh38
rs752272977 Chr.2: 23707381-23707381 on Build GRCh38
rs1029647111 Chr.2: 23707298-23707299 on Build GRCh38
rs951473440 Chr.2: 23707303-23707303 on Build GRCh38
rs547582260 Chr.2: 23707605-23707605 on Build GRCh38
rs762453504 Chr.2: 23707325-23707325 on Build GRCh38
rs1008811942 Chr.2: 23707280-23707280 on Build GRCh38
rs1021735529 Chr.2: 23707347-23707347 on Build GRCh38
rs750426143 Chr.2: 23707471-23707471 on Build GRCh38
rs1021666042 Chr.2: 23707158-23707158 on Build GRCh38
rs754222734 Chr.2: 23707310-23707310 on Build GRCh38
rs532161949 Chr.2: 23707585-23707585 on Build GRCh38
rs898574999 Chr.2: 23707599-23707599 on Build GRCh38
rs116187684 Chr.2: 23707256-23707256 on Build GRCh38
rs531370122 Chr.2: 23707420-23707420 on Build GRCh38
rs913679564 Chr.2: 23707171-23707171 on Build GRCh38
rs148928427 Chr.2: 23707270-23707270 on Build GRCh38
rs944333938 Chr.2: 23707555-23707555 on Build GRCh38
rs967936731 Chr.2: 23707160-23707160 on Build GRCh38
rs549774554 Chr.2: 23707436-23707436 on Build GRCh38
rs1039863851 Chr.2: 23707470-23707470 on Build GRCh38
rs973765332 Chr.2: 23707488-23707488 on Build GRCh38
rs960786186 Chr.2: 23707191-23707191 on Build GRCh38
rs1017038417 Chr.2: 23707568-23707568 on Build GRCh38
rs1056126368 Chr.2: 23707152-23707152 on Build GRCh38
rs932281481 Chr.2: 23707506-23707506 on Build GRCh38
rs559166380 Chr.2: 23707410-23707411 on Build GRCh38
rs913556485 Chr.2: 23707433-23707433 on Build GRCh38
rs1004176935 Chr.2: 23707555-23707555 on Build GRCh38
rs34591683 Chr.2: 23707182-23707183 on Build GRCh38
rs926570824 Chr.2: 23707172-23707172 on Build GRCh38
rs755832997 Chr.2: 23707581-23707581 on Build GRCh38
rs919685944 Chr.2: 23707497-23707497 on Build GRCh38
rs939806108 Chr.2: 23707235-23707235 on Build GRCh38

View Full Product Details