Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 23707501-23707997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD2B
Entrez Gene ID
54454
Gene Name
ATPase family, AAA domain containing 2B
Gene Aliases
-
UniGene
Hs.467862
Gene Chromosome Location
Chr.2: 23689710-23927114 on Build GRCh38
Transcripts
XM_006712030.3 NM_052920.1 XM_006711929.3 XM_011532501.2 XM_017003264.1

Gene Symbol
KLHL29
Entrez Gene ID
114818
Gene Name
kelch like family member 29
Gene Aliases
KBTBD9
UniGene
Hs.130593
Gene Chromosome Location
Chr.2: 23385419-23708613 on Build GRCh38
Transcripts
XM_006712030.3 NM_052920.1 XM_006711929.3 XM_011532501.2 XM_017003264.1

SNP Detail

SNP ID Chromosome Location
rs779949632 Chr.2: 23707688-23707688 on Build GRCh38
rs534865185 Chr.2: 23707894-23707894 on Build GRCh38
rs1053680262 Chr.2: 23707607-23707607 on Build GRCh38
rs565582236 Chr.2: 23707629-23707629 on Build GRCh38
rs921457729 Chr.2: 23707697-23707697 on Build GRCh38
rs1051356221 Chr.2: 23707746-23707746 on Build GRCh38
rs779495418 Chr.2: 23707963-23707963 on Build GRCh38
rs548583147 Chr.2: 23707751-23707751 on Build GRCh38
rs1043126335 Chr.2: 23707634-23707634 on Build GRCh38
rs912801697 Chr.2: 23707554-23707554 on Build GRCh38
rs1013795540 Chr.2: 23707905-23707905 on Build GRCh38
rs749112578 Chr.2: 23707784-23707784 on Build GRCh38
rs553593992 Chr.2: 23707915-23707915 on Build GRCh38
rs532161949 Chr.2: 23707585-23707585 on Build GRCh38
rs898574999 Chr.2: 23707599-23707599 on Build GRCh38
rs925066065 Chr.2: 23707676-23707676 on Build GRCh38
rs966589459 Chr.2: 23707943-23707943 on Build GRCh38
rs879364813 Chr.2: 23707768-23707768 on Build GRCh38
rs976017037 Chr.2: 23707692-23707692 on Build GRCh38
rs779251356 Chr.2: 23707884-23707884 on Build GRCh38
rs552618513 Chr.2: 23707899-23707899 on Build GRCh38
rs142747711 Chr.2: 23707632-23707632 on Build GRCh38
rs376739156 Chr.2: 23707893-23707893 on Build GRCh38
rs548305637 Chr.2: 23707651-23707651 on Build GRCh38
rs878998498 Chr.2: 23707771-23707771 on Build GRCh38
rs961162595 Chr.2: 23707870-23707870 on Build GRCh38
rs944333938 Chr.2: 23707555-23707555 on Build GRCh38
rs960935665 Chr.2: 23707619-23707619 on Build GRCh38
rs74364521 Chr.2: 23707719-23707719 on Build GRCh38
rs1017038417 Chr.2: 23707568-23707568 on Build GRCh38
rs1013526703 Chr.2: 23707815-23707815 on Build GRCh38
rs367616671 Chr.2: 23707540-23707540 on Build GRCh38
rs558955211 Chr.2: 23707787-23707787 on Build GRCh38
rs1031956602 Chr.2: 23707968-23707968 on Build GRCh38
rs763688125 Chr.2: 23707797-23707797 on Build GRCh38
rs2712098 Chr.2: 23707844-23707844 on Build GRCh38
rs898604484 Chr.2: 23707854-23707854 on Build GRCh38
rs183897344 Chr.2: 23707690-23707690 on Build GRCh38
rs1004176935 Chr.2: 23707555-23707555 on Build GRCh38
rs940037589 Chr.2: 23707764-23707764 on Build GRCh38
rs891944638 Chr.2: 23707631-23707631 on Build GRCh38
rs547582260 Chr.2: 23707605-23707605 on Build GRCh38
rs992102237 Chr.2: 23707622-23707622 on Build GRCh38
rs780958270 Chr.2: 23707852-23707852 on Build GRCh38
rs756843104 Chr.2: 23707869-23707869 on Build GRCh38
rs755832997 Chr.2: 23707581-23707581 on Build GRCh38

View Full Product Details