Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24174996-24175488 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM228A
Entrez Gene ID
653140
Gene Name
family with sequence similarity 228 member A
Gene Aliases
C2orf84
UniGene
Hs.741736
Gene Chromosome Location
Chr.2: 24175043-24191698 on Build GRCh38
Transcripts
NM_001040710.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5150501 c.5C>T p.A2V Chr.2: 24175485-24175485 on Build GRCh38
4093162 c.6T>C p.A2A Chr.2: 24175486-24175486 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs372054514 Chr.2: 24175458-24175458 on Build GRCh38
rs555102677 Chr.2: 24175325-24175325 on Build GRCh38
rs905053303 Chr.2: 24175141-24175141 on Build GRCh38
rs987763894 Chr.2: 24175266-24175266 on Build GRCh38
rs944449711 Chr.2: 24175241-24175241 on Build GRCh38
rs1052056154 Chr.2: 24175078-24175078 on Build GRCh38
rs761169252 Chr.2: 24175441-24175441 on Build GRCh38
rs994864575 Chr.2: 24175398-24175398 on Build GRCh38
rs573544890 Chr.2: 24175346-24175346 on Build GRCh38
rs780125982 Chr.2: 24175114-24175114 on Build GRCh38
rs530012573 Chr.2: 24175046-24175046 on Build GRCh38
rs946497423 Chr.2: 24175090-24175090 on Build GRCh38
rs956933452 Chr.2: 24175467-24175467 on Build GRCh38
rs952926404 Chr.2: 24175117-24175117 on Build GRCh38
rs1029451022 Chr.2: 24175407-24175407 on Build GRCh38
rs1003392229 Chr.2: 24175157-24175157 on Build GRCh38
rs1005746698 Chr.2: 24175391-24175391 on Build GRCh38
rs935295559 Chr.2: 24175206-24175206 on Build GRCh38
rs207461591 Chr.2: 24175132-24175132 on Build GRCh38
rs888890942 Chr.2: 24175239-24175239 on Build GRCh38
rs988097492 Chr.2: 24175070-24175070 on Build GRCh38
rs749143489 Chr.2: 24175306-24175306 on Build GRCh38
rs963455948 Chr.2: 24175218-24175218 on Build GRCh38
rs973590690 Chr.2: 24175036-24175036 on Build GRCh38
rs982148497 Chr.2: 24175160-24175160 on Build GRCh38
rs927979110 Chr.2: 24175195-24175195 on Build GRCh38
rs1004853865 Chr.2: 24175209-24175209 on Build GRCh38
rs571754600 Chr.2: 24175083-24175083 on Build GRCh38
rs897491484 Chr.2: 24175263-24175263 on Build GRCh38
rs1026275919 Chr.2: 24175100-24175100 on Build GRCh38
rs546868481 Chr.2: 24175071-24175072 on Build GRCh38
rs887028132 Chr.2: 24175198-24175198 on Build GRCh38
rs533618777 Chr.2: 24175288-24175288 on Build GRCh38
rs368132241 Chr.2: 24175451-24175451 on Build GRCh38
rs912161680 Chr.2: 24175074-24175074 on Build GRCh38
rs577851822 Chr.2: 24175408-24175408 on Build GRCh38
rs772387377 Chr.2: 24175435-24175435 on Build GRCh38
rs188029883 Chr.2: 24175375-24175375 on Build GRCh38
rs2042467 Chr.2: 24175412-24175412 on Build GRCh38
rs956985076 Chr.2: 24175264-24175264 on Build GRCh38
rs994435748 Chr.2: 24175084-24175084 on Build GRCh38
rs987658381 Chr.2: 24175250-24175250 on Build GRCh38
rs1008549678 Chr.2: 24175066-24175066 on Build GRCh38
rs914931193 Chr.2: 24175276-24175276 on Build GRCh38
rs553934950 Chr.2: 24175210-24175210 on Build GRCh38
rs571409515 Chr.2: 24175077-24175077 on Build GRCh38
rs963465708 Chr.2: 24175019-24175019 on Build GRCh38
rs1011118460 Chr.2: 24175020-24175020 on Build GRCh38
rs376569182 Chr.2: 24175461-24175461 on Build GRCh38
rs953645632 Chr.2: 24175235-24175235 on Build GRCh38
rs372043993 Chr.2: 24175171-24175171 on Build GRCh38
rs912109469 Chr.2: 24175251-24175251 on Build GRCh38
rs1029151701 Chr.2: 24175233-24175233 on Build GRCh38
rs775546054 Chr.2: 24175438-24175438 on Build GRCh38
rs55986600 Chr.2: 24175318-24175318 on Build GRCh38
rs867015604 Chr.2: 24175230-24175230 on Build GRCh38
rs769129979 Chr.2: 24175443-24175443 on Build GRCh38
rs1044886966 Chr.2: 24175129-24175129 on Build GRCh38
rs538810117 Chr.2: 24175140-24175140 on Build GRCh38
rs946566833 Chr.2: 24175211-24175211 on Build GRCh38
rs771086625 Chr.2: 24175435-24175435 on Build GRCh38
rs371489525 Chr.2: 24175122-24175122 on Build GRCh38
rs926411291 Chr.2: 24175305-24175305 on Build GRCh38
rs556376130 Chr.2: 24175394-24175394 on Build GRCh38
rs375721925 Chr.2: 24175229-24175229 on Build GRCh38
rs765801249 Chr.2: 24175464-24175464 on Build GRCh38
rs980900374 Chr.2: 24175302-24175302 on Build GRCh38
rs1056761209 Chr.2: 24175158-24175158 on Build GRCh38
rs953638346 Chr.2: 24175064-24175064 on Build GRCh38
rs375306135 Chr.2: 24175430-24175430 on Build GRCh38
rs922136274 Chr.2: 24175046-24175046 on Build GRCh38
rs768541444 Chr.2: 24175355-24175355 on Build GRCh38
rs888682352 Chr.2: 24175350-24175350 on Build GRCh38

View Full Product Details