Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24943528-24944075 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_017004269.1 XM_017004270.1

SNP Detail

SNP ID Chromosome Location
rs34922018 Chr.2: 24943829-24943830 on Build GRCh38
rs753650156 Chr.2: 24944028-24944028 on Build GRCh38
rs758708433 Chr.2: 24943761-24943761 on Build GRCh38
rs907670934 Chr.2: 24943632-24943632 on Build GRCh38
rs535943143 Chr.2: 24943988-24943988 on Build GRCh38
rs892432874 Chr.2: 24943846-24943846 on Build GRCh38
rs764271989 Chr.2: 24943877-24943877 on Build GRCh38
rs150529083 Chr.2: 24943569-24943569 on Build GRCh38
rs763071831 Chr.2: 24943825-24943825 on Build GRCh38
rs1001846142 Chr.2: 24944007-24944007 on Build GRCh38
rs111759308 Chr.2: 24943906-24943906 on Build GRCh38
rs983158287 Chr.2: 24943617-24943617 on Build GRCh38
rs775537102 Chr.2: 24943771-24943771 on Build GRCh38
rs943744123 Chr.2: 24943559-24943559 on Build GRCh38
rs932915950 Chr.2: 24943750-24943750 on Build GRCh38
rs891112684 Chr.2: 24943784-24943784 on Build GRCh38
rs191683185 Chr.2: 24943778-24943778 on Build GRCh38
rs528969365 Chr.2: 24943905-24943905 on Build GRCh38
rs976429887 Chr.2: 24943966-24943966 on Build GRCh38
rs147346699 Chr.2: 24943912-24943912 on Build GRCh38
rs1050035094 Chr.2: 24943775-24943775 on Build GRCh38
rs1045178180 Chr.2: 24943639-24943639 on Build GRCh38
rs766209784 Chr.2: 24943931-24943931 on Build GRCh38
rs573195660 Chr.2: 24943587-24943587 on Build GRCh38
rs575961236 Chr.2: 24944036-24944036 on Build GRCh38
rs1011203704 Chr.2: 24943847-24943847 on Build GRCh38
rs940156084 Chr.2: 24943637-24943637 on Build GRCh38
rs1022547261 Chr.2: 24943898-24943898 on Build GRCh38
rs148863492 Chr.2: 24943608-24943609 on Build GRCh38
rs965517130 Chr.2: 24943932-24943932 on Build GRCh38
rs959729144 Chr.2: 24943594-24943594 on Build GRCh38
rs974219782 Chr.2: 24943638-24943638 on Build GRCh38
rs920162057 Chr.2: 24943741-24943741 on Build GRCh38
rs906627679 Chr.2: 24943694-24943695 on Build GRCh38
rs139523732 Chr.2: 24943901-24943901 on Build GRCh38

View Full Product Details