Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24944307-24944845 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_017004269.1 XM_017004270.1

SNP Detail

SNP ID Chromosome Location
rs951074180 Chr.2: 24944375-24944375 on Build GRCh38
rs976503352 Chr.2: 24944364-24944364 on Build GRCh38
rs545931812 Chr.2: 24944647-24944647 on Build GRCh38
rs965635436 Chr.2: 24944473-24944473 on Build GRCh38
rs777524640 Chr.2: 24944570-24944570 on Build GRCh38
rs567099232 Chr.2: 24944353-24944356 on Build GRCh38
rs573158826 Chr.2: 24944607-24944607 on Build GRCh38
rs142491128 Chr.2: 24944471-24944471 on Build GRCh38
rs769278564 Chr.2: 24944797-24944797 on Build GRCh38
rs781745626 Chr.2: 24944556-24944556 on Build GRCh38
rs573530701 Chr.2: 24944689-24944689 on Build GRCh38
rs1026416543 Chr.2: 24944662-24944662 on Build GRCh38
rs927405181 Chr.2: 24944533-24944533 on Build GRCh38
rs940594822 Chr.2: 24944602-24944602 on Build GRCh38
rs147748312 Chr.2: 24944470-24944470 on Build GRCh38
rs373733107 Chr.2: 24944367-24944367 on Build GRCh38
rs192683025 Chr.2: 24944341-24944341 on Build GRCh38
rs951147083 Chr.2: 24944695-24944695 on Build GRCh38
rs947445453 Chr.2: 24944499-24944499 on Build GRCh38
rs866199311 Chr.2: 24944481-24944481 on Build GRCh38
rs1017013509 Chr.2: 24944729-24944729 on Build GRCh38
rs185644756 Chr.2: 24944775-24944775 on Build GRCh38
rs1014562899 Chr.2: 24944339-24944339 on Build GRCh38
rs994514015 Chr.2: 24944670-24944670 on Build GRCh38
rs756691083 Chr.2: 24944685-24944699 on Build GRCh38
rs145654874 Chr.2: 24944373-24944373 on Build GRCh38
rs969078691 Chr.2: 24944779-24944781 on Build GRCh38
rs989084613 Chr.2: 24944489-24944489 on Build GRCh38
rs550209303 Chr.2: 24944400-24944400 on Build GRCh38
rs981131979 Chr.2: 24944804-24944804 on Build GRCh38
rs913221092 Chr.2: 24944492-24944492 on Build GRCh38
rs902046424 Chr.2: 24944603-24944603 on Build GRCh38
rs969717884 Chr.2: 24944468-24944468 on Build GRCh38
rs1036266682 Chr.2: 24944631-24944631 on Build GRCh38
rs954314645 Chr.2: 24944382-24944382 on Build GRCh38
rs1005267271 Chr.2: 24944712-24944712 on Build GRCh38
rs527583605 Chr.2: 24944352-24944353 on Build GRCh38
rs994983918 Chr.2: 24944343-24944343 on Build GRCh38
rs889090847 Chr.2: 24944552-24944552 on Build GRCh38
rs895714494 Chr.2: 24944580-24944580 on Build GRCh38
rs78554489 Chr.2: 24944755-24944755 on Build GRCh38
rs558010972 Chr.2: 24944543-24944543 on Build GRCh38
rs983769590 Chr.2: 24944376-24944376 on Build GRCh38
rs1043534781 Chr.2: 24944530-24944530 on Build GRCh38
rs555396160 Chr.2: 24944682-24944682 on Build GRCh38

View Full Product Details