Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24944734-24945232 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_017004269.1 XM_017004270.1

SNP Detail

SNP ID Chromosome Location
rs946794004 Chr.2: 24944926-24944926 on Build GRCh38
rs142411584 Chr.2: 24945089-24945089 on Build GRCh38
rs376951036 Chr.2: 24944959-24944959 on Build GRCh38
rs969483640 Chr.2: 24945102-24945102 on Build GRCh38
rs888884509 Chr.2: 24944826-24944826 on Build GRCh38
rs899087653 Chr.2: 24945170-24945170 on Build GRCh38
rs769278564 Chr.2: 24944797-24944797 on Build GRCh38
rs1019773011 Chr.2: 24944896-24944896 on Build GRCh38
rs771138840 Chr.2: 24944880-24944880 on Build GRCh38
rs1172293 Chr.2: 24945051-24945051 on Build GRCh38
rs527976989 Chr.2: 24945067-24945067 on Build GRCh38
rs180899285 Chr.2: 24945071-24945071 on Build GRCh38
rs577693841 Chr.2: 24944967-24944967 on Build GRCh38
rs978795520 Chr.2: 24945017-24945017 on Build GRCh38
rs992898648 Chr.2: 24945070-24945070 on Build GRCh38
rs888803193 Chr.2: 24945192-24945192 on Build GRCh38
rs185644756 Chr.2: 24944775-24944775 on Build GRCh38
rs190222039 Chr.2: 24945038-24945038 on Build GRCh38
rs902118255 Chr.2: 24944972-24944972 on Build GRCh38
rs1016681143 Chr.2: 24945095-24945095 on Build GRCh38
rs969078691 Chr.2: 24944779-24944781 on Build GRCh38
rs531366311 Chr.2: 24945187-24945187 on Build GRCh38
rs943369385 Chr.2: 24944965-24944965 on Build GRCh38
rs927853132 Chr.2: 24944831-24944831 on Build GRCh38
rs981131979 Chr.2: 24944804-24944804 on Build GRCh38
rs549928113 Chr.2: 24945150-24945150 on Build GRCh38
rs1034655008 Chr.2: 24945145-24945145 on Build GRCh38
rs987993088 Chr.2: 24944892-24944892 on Build GRCh38
rs746692285 Chr.2: 24944850-24944850 on Build GRCh38
rs1007443321 Chr.2: 24944886-24944886 on Build GRCh38
rs559446692 Chr.2: 24945162-24945162 on Build GRCh38
rs543882813 Chr.2: 24944957-24944957 on Build GRCh38
rs960554616 Chr.2: 24944840-24944840 on Build GRCh38
rs531695648 Chr.2: 24945099-24945099 on Build GRCh38
rs1050114650 Chr.2: 24945188-24945188 on Build GRCh38
rs969085993 Chr.2: 24945008-24945008 on Build GRCh38
rs937462172 Chr.2: 24945068-24945068 on Build GRCh38
rs534679408 Chr.2: 24945107-24945108 on Build GRCh38
rs1036196285 Chr.2: 24945121-24945121 on Build GRCh38

View Full Product Details