Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24945132-24945644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_017004269.1 XM_017004270.1

SNP Detail

SNP ID Chromosome Location
rs976129162 Chr.2: 24945283-24945283 on Build GRCh38
rs77112774 Chr.2: 24945412-24945412 on Build GRCh38
rs566737157 Chr.2: 24945539-24945539 on Build GRCh38
rs1041927077 Chr.2: 24945569-24945569 on Build GRCh38
rs571691391 Chr.2: 24945217-24945217 on Build GRCh38
rs1038555689 Chr.2: 24945396-24945396 on Build GRCh38
rs774500023 Chr.2: 24945451-24945451 on Build GRCh38
rs1034686042 Chr.2: 24945438-24945438 on Build GRCh38
rs555593490 Chr.2: 24945373-24945373 on Build GRCh38
rs546745563 Chr.2: 24945528-24945528 on Build GRCh38
rs971882704 Chr.2: 24945508-24945508 on Build GRCh38
rs1035051277 Chr.2: 24945585-24945585 on Build GRCh38
rs1000441176 Chr.2: 24945584-24945584 on Build GRCh38
rs376382857 Chr.2: 24945421-24945421 on Build GRCh38
rs73920628 Chr.2: 24945532-24945532 on Build GRCh38
rs756924445 Chr.2: 24945612-24945612 on Build GRCh38
rs888803193 Chr.2: 24945192-24945192 on Build GRCh38
rs1047959734 Chr.2: 24945353-24945354 on Build GRCh38
rs542829557 Chr.2: 24945516-24945516 on Build GRCh38
rs151308563 Chr.2: 24945224-24945224 on Build GRCh38
rs969011292 Chr.2: 24945343-24945343 on Build GRCh38
rs1050800508 Chr.2: 24945518-24945518 on Build GRCh38
rs1009519116 Chr.2: 24945481-24945481 on Build GRCh38
rs777431285 Chr.2: 24945593-24945593 on Build GRCh38
rs11886240 Chr.2: 24945527-24945527 on Build GRCh38
rs559446692 Chr.2: 24945162-24945162 on Build GRCh38
rs1050114650 Chr.2: 24945188-24945188 on Build GRCh38
rs765233140 Chr.2: 24945533-24945533 on Build GRCh38
rs767698230 Chr.2: 24945526-24945526 on Build GRCh38
rs923236200 Chr.2: 24945340-24945340 on Build GRCh38
rs904715706 Chr.2: 24945583-24945583 on Build GRCh38
rs552500362 Chr.2: 24945519-24945519 on Build GRCh38
rs543784508 Chr.2: 24945604-24945604 on Build GRCh38
rs899087653 Chr.2: 24945170-24945170 on Build GRCh38
rs892229690 Chr.2: 24945538-24945538 on Build GRCh38
rs1044585837 Chr.2: 24945600-24945600 on Build GRCh38
rs555923064 Chr.2: 24945413-24945413 on Build GRCh38
rs140485746 Chr.2: 24945229-24945229 on Build GRCh38
rs964828916 Chr.2: 24945519-24945519 on Build GRCh38
rs573876535 Chr.2: 24945383-24945383 on Build GRCh38
rs373186522 Chr.2: 24945420-24945420 on Build GRCh38
rs566717855 Chr.2: 24945250-24945250 on Build GRCh38
rs150441011 Chr.2: 24945486-24945486 on Build GRCh38
rs531366311 Chr.2: 24945187-24945187 on Build GRCh38
rs1020514553 Chr.2: 24945313-24945313 on Build GRCh38
rs749737032 Chr.2: 24945324-24945324 on Build GRCh38
rs552278178 Chr.2: 24945620-24945620 on Build GRCh38
rs762981861 Chr.2: 24945336-24945336 on Build GRCh38
rs757928266 Chr.2: 24945589-24945589 on Build GRCh38
rs17046751 Chr.2: 24945608-24945608 on Build GRCh38
rs75112641 Chr.2: 24945509-24945509 on Build GRCh38
rs533977630 Chr.2: 24945273-24945273 on Build GRCh38
rs943420851 Chr.2: 24945270-24945270 on Build GRCh38
rs145509182 Chr.2: 24945614-24945614 on Build GRCh38
rs1024313421 Chr.2: 24945611-24945611 on Build GRCh38

View Full Product Details