Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24957732-24958243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_011532902.1 XM_017004269.1 XM_017004270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1019204 c.380T>G p.I127S Chr.2: 24957835-24957835 on Build GRCh38
442672 c.349G>T p.E117* Chr.2: 24957866-24957866 on Build GRCh38
2911629 c.245G>A p.R82Q Chr.2: 24957970-24957970 on Build GRCh38
4414415 c.361C>A p.H121N Chr.2: 24957854-24957854 on Build GRCh38
4164471 c.380T>A p.I127N Chr.2: 24957835-24957835 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766150158 Chr.2: 24957938-24957938 on Build GRCh38
rs564044426 Chr.2: 24957777-24957777 on Build GRCh38
rs192396063 Chr.2: 24958176-24958176 on Build GRCh38
rs756430253 Chr.2: 24957820-24957820 on Build GRCh38
rs897386439 Chr.2: 24958125-24958125 on Build GRCh38
rs900821566 Chr.2: 24958126-24958126 on Build GRCh38
rs371054022 Chr.2: 24958129-24958129 on Build GRCh38
rs954240646 Chr.2: 24957863-24957863 on Build GRCh38
rs375468318 Chr.2: 24957939-24957939 on Build GRCh38
rs949558113 Chr.2: 24958028-24958028 on Build GRCh38
rs557304396 Chr.2: 24958165-24958165 on Build GRCh38
rs746429741 Chr.2: 24957860-24957860 on Build GRCh38
rs758848351 Chr.2: 24958003-24958003 on Build GRCh38
rs115525250 Chr.2: 24958087-24958087 on Build GRCh38
rs988678274 Chr.2: 24957909-24957909 on Build GRCh38
rs747474254 Chr.2: 24957832-24957834 on Build GRCh38
rs376643367 Chr.2: 24958021-24958021 on Build GRCh38
rs777336562 Chr.2: 24957900-24957900 on Build GRCh38
rs534801055 Chr.2: 24958058-24958058 on Build GRCh38
rs183505121 Chr.2: 24958037-24958037 on Build GRCh38
rs182876550 Chr.2: 24957916-24957916 on Build GRCh38
rs550555089 Chr.2: 24957928-24957928 on Build GRCh38
rs780808213 Chr.2: 24958022-24958022 on Build GRCh38
rs769244220 Chr.2: 24957919-24957919 on Build GRCh38
rs200146548 Chr.2: 24957817-24957817 on Build GRCh38
rs773195940 Chr.2: 24957924-24957924 on Build GRCh38
rs1025507465 Chr.2: 24957857-24957857 on Build GRCh38
rs889834114 Chr.2: 24958209-24958209 on Build GRCh38
rs746614892 Chr.2: 24957889-24957889 on Build GRCh38
rs995784795 Chr.2: 24958155-24958155 on Build GRCh38
rs190369752 Chr.2: 24957819-24957819 on Build GRCh38
rs777964244 Chr.2: 24957835-24957835 on Build GRCh38
rs148479643 Chr.2: 24957917-24957917 on Build GRCh38
rs866684094 Chr.2: 24957893-24957893 on Build GRCh38
rs769267080 Chr.2: 24957959-24957959 on Build GRCh38
rs925696215 Chr.2: 24958090-24958090 on Build GRCh38
rs760359122 Chr.2: 24957763-24957763 on Build GRCh38
rs551426209 Chr.2: 24958009-24958009 on Build GRCh38
rs901713032 Chr.2: 24957958-24957967 on Build GRCh38
rs768328690 Chr.2: 24957897-24957897 on Build GRCh38
rs781464483 Chr.2: 24958016-24958020 on Build GRCh38
rs562389145 Chr.2: 24957844-24957844 on Build GRCh38
rs752948585 Chr.2: 24957815-24957815 on Build GRCh38
rs762791930 Chr.2: 24957779-24957779 on Build GRCh38
rs775205957 Chr.2: 24957762-24957762 on Build GRCh38
rs112617303 Chr.2: 24958061-24958061 on Build GRCh38
rs540104495 Chr.2: 24957888-24957888 on Build GRCh38
rs765275307 Chr.2: 24957973-24957973 on Build GRCh38
rs753976312 Chr.2: 24957839-24957839 on Build GRCh38
rs1016736639 Chr.2: 24958210-24958210 on Build GRCh38
rs942431321 Chr.2: 24958120-24958120 on Build GRCh38
rs545876669 Chr.2: 24957766-24957766 on Build GRCh38
rs374801528 Chr.2: 24957970-24957970 on Build GRCh38
rs754175162 Chr.2: 24957955-24957955 on Build GRCh38
rs988332282 Chr.2: 24958018-24958018 on Build GRCh38
rs150854407 Chr.2: 24957803-24957803 on Build GRCh38
rs887002132 Chr.2: 24958207-24958207 on Build GRCh38
rs946897788 Chr.2: 24958080-24958080 on Build GRCh38
rs1005802914 Chr.2: 24958208-24958208 on Build GRCh38
rs760914183 Chr.2: 24957943-24957943 on Build GRCh38
rs751432309 Chr.2: 24957941-24957941 on Build GRCh38
rs370182747 Chr.2: 24957771-24957771 on Build GRCh38
rs187777438 Chr.2: 24958067-24958067 on Build GRCh38
rs926988258 Chr.2: 24957974-24957974 on Build GRCh38
rs969462392 Chr.2: 24957956-24957956 on Build GRCh38
rs191551912 Chr.2: 24958004-24958004 on Build GRCh38
rs866327110 Chr.2: 24957843-24957843 on Build GRCh38
rs764429779 Chr.2: 24957946-24957946 on Build GRCh38
rs868089801 Chr.2: 24957841-24957841 on Build GRCh38
rs188835816 Chr.2: 24957984-24957984 on Build GRCh38
rs772453638 Chr.2: 24957886-24957886 on Build GRCh38

View Full Product Details