Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 24963273-24963815 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNAJC27
Entrez Gene ID
51277
Gene Name
DnaJ heat shock protein family (Hsp40) member C27
Gene Aliases
RBJ, RabJS
UniGene
Hs.434993
Gene Chromosome Location
Chr.2: 24943636-24972094 on Build GRCh38
Transcripts
NM_001198559.1 NM_016544.2 XM_011532902.1 XM_017004269.1 XM_017004270.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1407076 c.179T>G p.V60G Chr.2: 24963466-24963466 on Build GRCh38
2911631 c.178G>A p.V60I Chr.2: 24963467-24963467 on Build GRCh38
7324194 c.221G>T p.G74V Chr.2: 24963424-24963424 on Build GRCh38
5862261 c.175C>T p.H59Y Chr.2: 24963470-24963470 on Build GRCh38
274853 c.207C>A p.I69I Chr.2: 24963438-24963438 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760083290 Chr.2: 24963475-24963475 on Build GRCh38
rs548797143 Chr.2: 24963363-24963363 on Build GRCh38
rs371271613 Chr.2: 24963506-24963506 on Build GRCh38
rs368306040 Chr.2: 24963430-24963430 on Build GRCh38
rs777278893 Chr.2: 24963501-24963501 on Build GRCh38
rs748761795 Chr.2: 24963395-24963395 on Build GRCh38
rs774230113 Chr.2: 24963359-24963359 on Build GRCh38
rs199534250 Chr.2: 24963494-24963494 on Build GRCh38
rs747789379 Chr.2: 24963387-24963387 on Build GRCh38
rs750497850 Chr.2: 24963366-24963366 on Build GRCh38
rs889263029 Chr.2: 24963702-24963702 on Build GRCh38
rs755755538 Chr.2: 24963394-24963394 on Build GRCh38
rs1002730681 Chr.2: 24963300-24963300 on Build GRCh38
rs773425774 Chr.2: 24963501-24963504 on Build GRCh38
rs956995632 Chr.2: 24963405-24963405 on Build GRCh38
rs752118391 Chr.2: 24963372-24963372 on Build GRCh38
rs980886471 Chr.2: 24963500-24963500 on Build GRCh38
rs762927821 Chr.2: 24963454-24963454 on Build GRCh38
rs1009823993 Chr.2: 24963779-24963779 on Build GRCh38
rs752028592 Chr.2: 24963468-24963468 on Build GRCh38
rs908427866 Chr.2: 24963557-24963557 on Build GRCh38
rs1019124164 Chr.2: 24963732-24963732 on Build GRCh38
rs932392855 Chr.2: 24963675-24963675 on Build GRCh38
rs898203453 Chr.2: 24963641-24963641 on Build GRCh38
rs771973715 Chr.2: 24963423-24963423 on Build GRCh38
rs753598583 Chr.2: 24963502-24963502 on Build GRCh38
rs766511358 Chr.2: 24963467-24963467 on Build GRCh38
rs918843368 Chr.2: 24963718-24963718 on Build GRCh38
rs190480393 Chr.2: 24963505-24963505 on Build GRCh38
rs948193821 Chr.2: 24963771-24963771 on Build GRCh38
rs753010475 Chr.2: 24963495-24963495 on Build GRCh38
rs892676614 Chr.2: 24963761-24963761 on Build GRCh38
rs996557509 Chr.2: 24963352-24963352 on Build GRCh38
rs748084533 Chr.2: 24963601-24963601 on Build GRCh38
rs772164162 Chr.2: 24963508-24963508 on Build GRCh38
rs1019636850 Chr.2: 24963390-24963390 on Build GRCh38
rs1010517648 Chr.2: 24963577-24963577 on Build GRCh38
rs144719716 Chr.2: 24963565-24963565 on Build GRCh38
rs796922155 Chr.2: 24963533-24963533 on Build GRCh38
rs1038054427 Chr.2: 24963308-24963308 on Build GRCh38
rs546720430 Chr.2: 24963625-24963625 on Build GRCh38
rs774430161 Chr.2: 24963417-24963417 on Build GRCh38
rs765379265 Chr.2: 24963443-24963443 on Build GRCh38
rs9636440 Chr.2: 24963607-24963607 on Build GRCh38
rs369273131 Chr.2: 24963499-24963499 on Build GRCh38
rs372317934 Chr.2: 24963446-24963446 on Build GRCh38
rs1018344678 Chr.2: 24963661-24963661 on Build GRCh38
rs770199798 Chr.2: 24963473-24963473 on Build GRCh38
rs550281862 Chr.2: 24963763-24963763 on Build GRCh38
rs985531003 Chr.2: 24963649-24963649 on Build GRCh38
rs775167708 Chr.2: 24963425-24963425 on Build GRCh38
rs759395277 Chr.2: 24963361-24963361 on Build GRCh38
rs773281094 Chr.2: 24963450-24963450 on Build GRCh38
rs765308624 Chr.2: 24963365-24963365 on Build GRCh38
rs568213260 Chr.2: 24963654-24963654 on Build GRCh38
rs911209058 Chr.2: 24963433-24963433 on Build GRCh38
rs1004264434 Chr.2: 24963299-24963299 on Build GRCh38
rs746789441 Chr.2: 24963520-24963520 on Build GRCh38
rs535003523 Chr.2: 24963588-24963588 on Build GRCh38
rs748235035 Chr.2: 24963410-24963412 on Build GRCh38
rs766881144 Chr.2: 24963371-24963371 on Build GRCh38
rs770627067 Chr.2: 24963396-24963396 on Build GRCh38
rs976741057 Chr.2: 24963707-24963707 on Build GRCh38
rs867268241 Chr.2: 24963682-24963682 on Build GRCh38
rs944302774 Chr.2: 24963441-24963441 on Build GRCh38
rs1021454348 Chr.2: 24963582-24963582 on Build GRCh38
rs768550478 Chr.2: 24963524-24963524 on Build GRCh38
rs368864997 Chr.2: 24963362-24963362 on Build GRCh38
rs755330276 Chr.2: 24963378-24963378 on Build GRCh38
rs772062294 Chr.2: 24963686-24963686 on Build GRCh38
rs984735965 Chr.2: 24963725-24963725 on Build GRCh38
rs535512554 Chr.2: 24963719-24963719 on Build GRCh38
rs770997037 Chr.2: 24963358-24963358 on Build GRCh38
rs772694840 Chr.2: 24963780-24963780 on Build GRCh38
rs184871179 Chr.2: 24963370-24963370 on Build GRCh38
rs1035583253 Chr.2: 24963352-24963352 on Build GRCh38
rs898219519 Chr.2: 24963318-24963318 on Build GRCh38
rs779829340 Chr.2: 24963509-24963509 on Build GRCh38
rs756800290 Chr.2: 24963503-24963503 on Build GRCh38
rs142148817 Chr.2: 24963421-24963421 on Build GRCh38

View Full Product Details