Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25379198-25379707 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DTNB
Entrez Gene ID
1838
Gene Name
dystrobrevin beta
Gene Aliases
-
UniGene
Hs.307720
Gene Chromosome Location
Chr.2: 25377220-25673647 on Build GRCh38
Transcripts
NM_001256303.1 NM_001256304.1 NM_001256308.2 NM_001320932.1 NM_001320933.1 NM_001320934.1 NM_001320935.1 NM_021907.4 NM_033147.3 NM_033148.3 NM_183360.2 NM_183361.2 XM_005264182.1 XM_011532671.1 XM_011532673.1 XM_011532674.1 XM_011532675.1 XM_011532676.1 XM_011532677.2 XM_011532680.2 XM_011532681.2 XM_017003529.1 XM_017003532.1 XM_017003533.1 XM_017003534.1 XM_017003535.1 XM_017003537.1 XM_017003538.1 XM_017003539.1 XM_017003540.1 XM_017003541.1 XM_017003542.1 XM_017003543.1 XM_017003544.1 XM_017003545.1

SNP Detail

SNP ID Chromosome Location
rs769536748 Chr.2: 25379485-25379485 on Build GRCh38
rs972050239 Chr.2: 25379327-25379327 on Build GRCh38
rs909255169 Chr.2: 25379422-25379422 on Build GRCh38
rs769002206 Chr.2: 25379611-25379611 on Build GRCh38
rs1016473845 Chr.2: 25379683-25379683 on Build GRCh38
rs1016754373 Chr.2: 25379547-25379549 on Build GRCh38
rs374398348 Chr.2: 25379352-25379353 on Build GRCh38
rs763851120 Chr.2: 25379420-25379420 on Build GRCh38
rs950891611 Chr.2: 25379353-25379353 on Build GRCh38
rs556212820 Chr.2: 25379421-25379421 on Build GRCh38
rs913685125 Chr.2: 25379419-25379419 on Build GRCh38
rs376657937 Chr.2: 25379492-25379492 on Build GRCh38
rs576237680 Chr.2: 25379380-25379380 on Build GRCh38
rs933213817 Chr.2: 25379468-25379468 on Build GRCh38
rs752562117 Chr.2: 25379487-25379488 on Build GRCh38
rs751042756 Chr.2: 25379337-25379337 on Build GRCh38
rs913549567 Chr.2: 25379612-25379612 on Build GRCh38
rs377478255 Chr.2: 25379496-25379496 on Build GRCh38
rs201977400 Chr.2: 25379309-25379309 on Build GRCh38
rs375407461 Chr.2: 25379286-25379286 on Build GRCh38
rs370598266 Chr.2: 25379255-25379255 on Build GRCh38
rs1026882813 Chr.2: 25379335-25379335 on Build GRCh38
rs889273683 Chr.2: 25379449-25379449 on Build GRCh38
rs769793472 Chr.2: 25379367-25379367 on Build GRCh38
rs969642699 Chr.2: 25379562-25379562 on Build GRCh38
rs993489273 Chr.2: 25379224-25379224 on Build GRCh38
rs987843157 Chr.2: 25379607-25379607 on Build GRCh38
rs745417523 Chr.2: 25379463-25379463 on Build GRCh38
rs923552450 Chr.2: 25379677-25379677 on Build GRCh38
rs577089212 Chr.2: 25379628-25379628 on Build GRCh38
rs894896315 Chr.2: 25379537-25379537 on Build GRCh38
rs910278616 Chr.2: 25379642-25379642 on Build GRCh38
rs2304428 Chr.2: 25379581-25379581 on Build GRCh38
rs538651436 Chr.2: 25379241-25379241 on Build GRCh38
rs576895740 Chr.2: 25379259-25379259 on Build GRCh38
rs887902966 Chr.2: 25379539-25379539 on Build GRCh38
rs779712885 Chr.2: 25379515-25379515 on Build GRCh38
rs984952581 Chr.2: 25379670-25379670 on Build GRCh38
rs371116808 Chr.2: 25379277-25379277 on Build GRCh38
rs764618654 Chr.2: 25379281-25379281 on Build GRCh38
rs2304427 Chr.2: 25379441-25379441 on Build GRCh38
rs765665995 Chr.2: 25379301-25379301 on Build GRCh38
rs754471327 Chr.2: 25379346-25379346 on Build GRCh38
rs772009218 Chr.2: 25379360-25379361 on Build GRCh38
rs981748027 Chr.2: 25379334-25379334 on Build GRCh38
rs554904685 Chr.2: 25379347-25379347 on Build GRCh38
rs376612696 Chr.2: 25379230-25379230 on Build GRCh38
rs927876383 Chr.2: 25379387-25379387 on Build GRCh38
rs775486056 Chr.2: 25379278-25379278 on Build GRCh38
rs951034465 Chr.2: 25379634-25379634 on Build GRCh38
rs943535244 Chr.2: 25379662-25379662 on Build GRCh38
rs868653534 Chr.2: 25379404-25379404 on Build GRCh38
rs967818636 Chr.2: 25379621-25379621 on Build GRCh38
rs756706719 Chr.2: 25379572-25379572 on Build GRCh38
rs923324836 Chr.2: 25379467-25379467 on Build GRCh38
rs939034728 Chr.2: 25379400-25379400 on Build GRCh38
rs780833157 Chr.2: 25379448-25379448 on Build GRCh38
rs1011986043 Chr.2: 25379563-25379563 on Build GRCh38
rs762429872 Chr.2: 25379299-25379299 on Build GRCh38
rs2304426 Chr.2: 25379273-25379273 on Build GRCh38
rs984962453 Chr.2: 25379418-25379418 on Build GRCh38
rs375720562 Chr.2: 25379260-25379260 on Build GRCh38
rs547350901 Chr.2: 25379248-25379248 on Build GRCh38
rs1040511725 Chr.2: 25379666-25379669 on Build GRCh38
rs373766114 Chr.2: 25379487-25379487 on Build GRCh38
rs35126842 Chr.2: 25379399-25379399 on Build GRCh38
rs906224195 Chr.2: 25379578-25379578 on Build GRCh38
rs1053553767 Chr.2: 25379518-25379518 on Build GRCh38
rs779857589 Chr.2: 25379323-25379323 on Build GRCh38
rs760673390 Chr.2: 25379280-25379280 on Build GRCh38
rs774270522 Chr.2: 25379277-25379280 on Build GRCh38
rs1024585156 Chr.2: 25379564-25379564 on Build GRCh38
rs748009282 Chr.2: 25379362-25379362 on Build GRCh38
rs368628317 Chr.2: 25379308-25379308 on Build GRCh38
rs987769569 Chr.2: 25379415-25379415 on Build GRCh38
rs1006245338 Chr.2: 25379546-25379546 on Build GRCh38
rs993914833 Chr.2: 25379505-25379505 on Build GRCh38

View Full Product Details