Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 25742095-25742598 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASXL2
Entrez Gene ID
55252
Gene Name
additional sex combs like 2, transcriptional regulator
Gene Aliases
ASXH2
UniGene
Hs.119815
Gene Chromosome Location
Chr.2: 25733758-25878485 on Build GRCh38
Transcripts
NM_018263.4 XM_006712039.3 XM_006712040.2 XM_011532950.2 XM_011532951.2 XM_017004429.1 XM_017004430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6969940 c.3990G>A p.M1330I Chr.2: 25742347-25742347 on Build GRCh38
6936843 c.3791A>G p.D1264G Chr.2: 25742546-25742546 on Build GRCh38
6976737 c.2467G>A p.D823N Chr.2: 25742319-25742319 on Build GRCh38
6446613 c.4165G>T p.E1389* Chr.2: 25742172-25742172 on Build GRCh38
2911944 c.3926C>T p.P1309L Chr.2: 25742411-25742411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769817520 Chr.2: 25742293-25742293 on Build GRCh38
rs764200522 Chr.2: 25742134-25742134 on Build GRCh38
rs771593827 Chr.2: 25742398-25742398 on Build GRCh38
rs757071976 Chr.2: 25742274-25742274 on Build GRCh38
rs369161602 Chr.2: 25742184-25742184 on Build GRCh38
rs372536765 Chr.2: 25742371-25742371 on Build GRCh38
rs955754916 Chr.2: 25742347-25742347 on Build GRCh38
rs150720719 Chr.2: 25742379-25742379 on Build GRCh38
rs61744558 Chr.2: 25742421-25742421 on Build GRCh38
rs774857577 Chr.2: 25742335-25742335 on Build GRCh38
rs374945043 Chr.2: 25742143-25742143 on Build GRCh38
rs572342520 Chr.2: 25742489-25742489 on Build GRCh38
rs769337369 Chr.2: 25742533-25742533 on Build GRCh38
rs776161059 Chr.2: 25742521-25742521 on Build GRCh38
rs776010496 Chr.2: 25742437-25742437 on Build GRCh38
rs763425141 Chr.2: 25742314-25742314 on Build GRCh38
rs553349404 Chr.2: 25742462-25742462 on Build GRCh38
rs375656900 Chr.2: 25742514-25742514 on Build GRCh38
rs752902849 Chr.2: 25742463-25742463 on Build GRCh38
rs772369923 Chr.2: 25742129-25742129 on Build GRCh38
rs996436134 Chr.2: 25742269-25742269 on Build GRCh38
rs373233840 Chr.2: 25742220-25742220 on Build GRCh38
rs757459663 Chr.2: 25742488-25742488 on Build GRCh38
rs548047376 Chr.2: 25742248-25742248 on Build GRCh38
rs914399385 Chr.2: 25742376-25742376 on Build GRCh38
rs537141280 Chr.2: 25742389-25742389 on Build GRCh38
rs756079519 Chr.2: 25742170-25742170 on Build GRCh38
rs750253621 Chr.2: 25742375-25742375 on Build GRCh38
rs570805684 Chr.2: 25742396-25742396 on Build GRCh38
rs756290348 Chr.2: 25742478-25742478 on Build GRCh38
rs780904802 Chr.2: 25742168-25742168 on Build GRCh38
rs775732218 Chr.2: 25742493-25742493 on Build GRCh38
rs780551383 Chr.2: 25742180-25742180 on Build GRCh38
rs781373928 Chr.2: 25742384-25742384 on Build GRCh38
rs772604191 Chr.2: 25742425-25742425 on Build GRCh38
rs534744008 Chr.2: 25742419-25742419 on Build GRCh38
rs370518430 Chr.2: 25742428-25742428 on Build GRCh38
rs910114091 Chr.2: 25742460-25742460 on Build GRCh38
rs190294224 Chr.2: 25742144-25742144 on Build GRCh38
rs61745462 Chr.2: 25742408-25742408 on Build GRCh38
rs773133219 Chr.2: 25742566-25742566 on Build GRCh38
rs370555517 Chr.2: 25742366-25742366 on Build GRCh38
rs191895729 Chr.2: 25742337-25742337 on Build GRCh38
rs768709532 Chr.2: 25742202-25742202 on Build GRCh38
rs775329935 Chr.2: 25742342-25742342 on Build GRCh38
rs752284764 Chr.2: 25742250-25742250 on Build GRCh38
rs773402627 Chr.2: 25742219-25742219 on Build GRCh38
rs376210436 Chr.2: 25742338-25742338 on Build GRCh38
rs771264308 Chr.2: 25742322-25742322 on Build GRCh38
rs1021520950 Chr.2: 25742279-25742279 on Build GRCh38
rs546352636 Chr.2: 25742130-25742130 on Build GRCh38
rs770704442 Chr.2: 25742287-25742294 on Build GRCh38
rs375737665 Chr.2: 25742441-25742441 on Build GRCh38
rs186221992 Chr.2: 25742179-25742179 on Build GRCh38
rs367860439 Chr.2: 25742512-25742512 on Build GRCh38
rs757158204 Chr.2: 25742364-25742364 on Build GRCh38
rs780114874 Chr.2: 25742216-25742216 on Build GRCh38
rs765184636 Chr.2: 25742443-25742443 on Build GRCh38
rs371375230 Chr.2: 25742173-25742173 on Build GRCh38
rs61744557 Chr.2: 25742423-25742423 on Build GRCh38
rs558833597 Chr.2: 25742395-25742395 on Build GRCh38
rs548343899 Chr.2: 25742363-25742363 on Build GRCh38
rs746010291 Chr.2: 25742120-25742120 on Build GRCh38
rs777391361 Chr.2: 25742172-25742172 on Build GRCh38
rs750411348 Chr.2: 25742446-25742446 on Build GRCh38
rs984403631 Chr.2: 25742457-25742457 on Build GRCh38
rs377592465 Chr.2: 25742405-25742405 on Build GRCh38
rs113503588 Chr.2: 25742149-25742149 on Build GRCh38
rs138300375 Chr.2: 25742530-25742530 on Build GRCh38
rs61745463 Chr.2: 25742407-25742407 on Build GRCh38
rs749734270 Chr.2: 25742397-25742397 on Build GRCh38
rs200059955 Chr.2: 25742507-25742507 on Build GRCh38
rs61745460 Chr.2: 25742415-25742415 on Build GRCh38
rs369921227 Chr.2: 25742272-25742272 on Build GRCh38
rs760200576 Chr.2: 25742344-25742344 on Build GRCh38
rs755954518 Chr.2: 25742270-25742270 on Build GRCh38
rs115221221 Chr.2: 25742226-25742226 on Build GRCh38
rs751241845 Chr.2: 25742230-25742230 on Build GRCh38
rs986882403 Chr.2: 25742321-25742321 on Build GRCh38
rs750809033 Chr.2: 25742150-25742150 on Build GRCh38
rs759270666 Chr.2: 25742235-25742235 on Build GRCh38
rs753758339 Chr.2: 25742273-25742273 on Build GRCh38
rs768871277 Chr.2: 25742290-25742290 on Build GRCh38
rs778738425 Chr.2: 25742276-25742276 on Build GRCh38
rs776705808 Chr.2: 25742204-25742204 on Build GRCh38
rs749347152 Chr.2: 25742487-25742487 on Build GRCh38
rs762477855 Chr.2: 25742160-25742160 on Build GRCh38
rs184151788 Chr.2: 25742362-25742362 on Build GRCh38
rs955113689 Chr.2: 25742517-25742517 on Build GRCh38
rs762244436 Chr.2: 25742215-25742215 on Build GRCh38
rs777510322 Chr.2: 25742285-25742285 on Build GRCh38
rs61744554 Chr.2: 25742435-25742435 on Build GRCh38
rs773742264 Chr.2: 25742312-25742312 on Build GRCh38
rs377609636 Chr.2: 25742490-25742490 on Build GRCh38
rs1004952488 Chr.2: 25742187-25742187 on Build GRCh38
rs777867553 Chr.2: 25742479-25742479 on Build GRCh38
rs965157437 Chr.2: 25742370-25742370 on Build GRCh38
rs1004025763 Chr.2: 25742206-25742206 on Build GRCh38
rs766080550 Chr.2: 25742164-25742164 on Build GRCh38
rs762887744 Chr.2: 25742567-25742567 on Build GRCh38
rs373784825 Chr.2: 25742291-25742291 on Build GRCh38
rs752619832 Chr.2: 25742169-25742169 on Build GRCh38
rs758162529 Chr.2: 25742455-25742455 on Build GRCh38
rs61745461 Chr.2: 25742409-25742409 on Build GRCh38
rs763602575 Chr.2: 25742436-25742436 on Build GRCh38
rs751309437 Chr.2: 25742167-25742167 on Build GRCh38
rs760303756 Chr.2: 25742433-25742433 on Build GRCh38
rs1029216859 Chr.2: 25742509-25742509 on Build GRCh38

View Full Product Details