Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26176261-26176763 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAREM2
Entrez Gene ID
150946
Gene Name
GRB2 associated regulator of MAPK1 subtype 2
Gene Aliases
FAM59B, GAREML
UniGene
Hs.187912
Gene Chromosome Location
Chr.2: 26173091-26202405 on Build GRCh38
Transcripts
NM_001168241.1 XM_006711951.3 XM_011532564.2 XM_011532565.2 XM_011532566.2 XM_011532567.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4915025 c.214G>T p.V72F Chr.2: 26176445-26176445 on Build GRCh38
6326339 c.126G>A p.E42E Chr.2: 26176357-26176357 on Build GRCh38
1614682 c.243G>A p.L81L Chr.2: 26176474-26176474 on Build GRCh38
3695426 c.179C>T p.T60M Chr.2: 26176410-26176410 on Build GRCh38
1019303 c.136G>A p.E46K Chr.2: 26176367-26176367 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs973754386 Chr.2: 26176596-26176596 on Build GRCh38
rs890942672 Chr.2: 26176391-26176391 on Build GRCh38
rs932326263 Chr.2: 26176604-26176604 on Build GRCh38
rs912492992 Chr.2: 26176621-26176621 on Build GRCh38
rs892382059 Chr.2: 26176496-26176496 on Build GRCh38
rs1011575685 Chr.2: 26176509-26176509 on Build GRCh38
rs761676685 Chr.2: 26176289-26176289 on Build GRCh38
rs751173063 Chr.2: 26176556-26176556 on Build GRCh38
rs575966670 Chr.2: 26176572-26176572 on Build GRCh38
rs747343441 Chr.2: 26176336-26176336 on Build GRCh38
rs878911763 Chr.2: 26176349-26176349 on Build GRCh38
rs752438282 Chr.2: 26176462-26176462 on Build GRCh38
rs908245781 Chr.2: 26176582-26176582 on Build GRCh38
rs569858299 Chr.2: 26176640-26176640 on Build GRCh38
rs965545109 Chr.2: 26176341-26176341 on Build GRCh38
rs751459214 Chr.2: 26176397-26176397 on Build GRCh38
rs774271327 Chr.2: 26176304-26176307 on Build GRCh38
rs147118036 Chr.2: 26176309-26176310 on Build GRCh38
rs1038445659 Chr.2: 26176353-26176353 on Build GRCh38
rs762500060 Chr.2: 26176369-26176369 on Build GRCh38
rs777501978 Chr.2: 26176352-26176352 on Build GRCh38
rs893213208 Chr.2: 26176669-26176669 on Build GRCh38
rs1052791678 Chr.2: 26176479-26176479 on Build GRCh38
rs779332966 Chr.2: 26176501-26176501 on Build GRCh38
rs901340696 Chr.2: 26176577-26176577 on Build GRCh38
rs11675057 Chr.2: 26176612-26176612 on Build GRCh38
rs928836896 Chr.2: 26176306-26176306 on Build GRCh38
rs185450095 Chr.2: 26176332-26176332 on Build GRCh38
rs563491136 Chr.2: 26176371-26176371 on Build GRCh38
rs969060339 Chr.2: 26176661-26176661 on Build GRCh38
rs1035921407 Chr.2: 26176554-26176554 on Build GRCh38
rs528463260 Chr.2: 26176481-26176481 on Build GRCh38
rs983773099 Chr.2: 26176643-26176643 on Build GRCh38
rs970270866 Chr.2: 26176541-26176541 on Build GRCh38
rs4665830 Chr.2: 26176345-26176345 on Build GRCh38
rs142877591 Chr.2: 26176601-26176601 on Build GRCh38
rs190551610 Chr.2: 26176390-26176390 on Build GRCh38
rs553375469 Chr.2: 26176319-26176319 on Build GRCh38
rs879911965 Chr.2: 26176716-26176717 on Build GRCh38
rs772698775 Chr.2: 26176355-26176355 on Build GRCh38
rs182331449 Chr.2: 26176685-26176685 on Build GRCh38
rs755174696 Chr.2: 26176539-26176539 on Build GRCh38
rs138312040 Chr.2: 26176398-26176398 on Build GRCh38
rs776876537 Chr.2: 26176351-26176351 on Build GRCh38
rs1003103453 Chr.2: 26176551-26176551 on Build GRCh38
rs373270167 Chr.2: 26176411-26176411 on Build GRCh38
rs548415657 Chr.2: 26176634-26176634 on Build GRCh38
rs1016539661 Chr.2: 26176658-26176658 on Build GRCh38
rs796143321 Chr.2: 26176450-26176450 on Build GRCh38
rs374428862 Chr.2: 26176471-26176471 on Build GRCh38
rs764867918 Chr.2: 26176354-26176354 on Build GRCh38
rs1053403072 Chr.2: 26176665-26176665 on Build GRCh38
rs1043061689 Chr.2: 26176565-26176565 on Build GRCh38
rs899743762 Chr.2: 26176360-26176360 on Build GRCh38
rs962815301 Chr.2: 26176585-26176585 on Build GRCh38
rs1007400397 Chr.2: 26176576-26176576 on Build GRCh38
rs776185018 Chr.2: 26176313-26176313 on Build GRCh38

View Full Product Details