Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26575823-26576330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C2orf70
Entrez Gene ID
339778
Gene Name
chromosome 2 open reading frame 70
Gene Aliases
-
UniGene
Hs.407083
Gene Chromosome Location
Chr.2: 26562559-26581166 on Build GRCh38
Transcripts
NM_001105519.2 NM_001322426.1 XM_011532810.2 XM_011532811.2 XM_017003962.1 XM_017003963.1 XM_017003964.1 XM_017003965.1 XM_017003966.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
139380 c.148C>T p.Q50* Chr.2: 26575975-26575975 on Build GRCh38
6615905 c.231delC p.N79fs*4 Chr.2: 26576058-26576058 on Build GRCh38
6615904 c.254G>A p.R85H Chr.2: 26576081-26576081 on Build GRCh38
6615903 c.114C>T p.G38G Chr.2: 26575941-26575941 on Build GRCh38
4001632 c.336G>A p.T112T Chr.2: 26576163-26576163 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761937059 Chr.2: 26575853-26575853 on Build GRCh38
rs759830210 Chr.2: 26575891-26575891 on Build GRCh38
rs376892936 Chr.2: 26575890-26575890 on Build GRCh38
rs771408978 Chr.2: 26575955-26575955 on Build GRCh38
rs766366811 Chr.2: 26576196-26576196 on Build GRCh38
rs762536045 Chr.2: 26576189-26576189 on Build GRCh38
rs377310271 Chr.2: 26576219-26576219 on Build GRCh38
rs138550936 Chr.2: 26576099-26576099 on Build GRCh38
rs763609891 Chr.2: 26575878-26575878 on Build GRCh38
rs1014229954 Chr.2: 26576280-26576280 on Build GRCh38
rs749380596 Chr.2: 26575951-26575951 on Build GRCh38
rs931060743 Chr.2: 26575884-26575884 on Build GRCh38
rs937645131 Chr.2: 26576274-26576274 on Build GRCh38
rs534721835 Chr.2: 26575959-26575959 on Build GRCh38
rs376431029 Chr.2: 26576270-26576270 on Build GRCh38
rs531598350 Chr.2: 26576292-26576292 on Build GRCh38
rs770944861 Chr.2: 26576057-26576057 on Build GRCh38
rs767468540 Chr.2: 26576217-26576217 on Build GRCh38
rs780554569 Chr.2: 26576128-26576128 on Build GRCh38
rs972606761 Chr.2: 26576204-26576204 on Build GRCh38
rs778722509 Chr.2: 26575918-26575918 on Build GRCh38
rs1021612309 Chr.2: 26575928-26575928 on Build GRCh38
rs763590692 Chr.2: 26575984-26575984 on Build GRCh38
rs913003230 Chr.2: 26576255-26576255 on Build GRCh38
rs773829464 Chr.2: 26575864-26575864 on Build GRCh38
rs778151436 Chr.2: 26576028-26576028 on Build GRCh38
rs758331071 Chr.2: 26576098-26576098 on Build GRCh38
rs2272464 Chr.2: 26576163-26576163 on Build GRCh38
rs1038073441 Chr.2: 26576232-26576232 on Build GRCh38
rs376790545 Chr.2: 26576080-26576080 on Build GRCh38
rs904732072 Chr.2: 26576269-26576269 on Build GRCh38
rs969144882 Chr.2: 26575938-26575938 on Build GRCh38
rs759085052 Chr.2: 26576231-26576231 on Build GRCh38
rs764698232 Chr.2: 26576092-26576092 on Build GRCh38
rs867201385 Chr.2: 26576146-26576146 on Build GRCh38
rs766644577 Chr.2: 26575885-26575885 on Build GRCh38
rs757337747 Chr.2: 26576124-26576124 on Build GRCh38
rs764726900 Chr.2: 26576075-26576076 on Build GRCh38
rs763680722 Chr.2: 26576081-26576081 on Build GRCh38
rs542880558 Chr.2: 26576088-26576088 on Build GRCh38
rs112873448 Chr.2: 26576059-26576059 on Build GRCh38
rs748424847 Chr.2: 26575945-26575945 on Build GRCh38
rs776830826 Chr.2: 26575897-26575897 on Build GRCh38
rs903308216 Chr.2: 26576278-26576279 on Build GRCh38
rs561356267 Chr.2: 26575942-26575942 on Build GRCh38
rs754839442 Chr.2: 26576108-26576108 on Build GRCh38
rs6740750 Chr.2: 26576181-26576181 on Build GRCh38
rs374971635 Chr.2: 26575915-26575915 on Build GRCh38
rs1036650911 Chr.2: 26575847-26575847 on Build GRCh38
rs2272465 Chr.2: 26576223-26576223 on Build GRCh38
rs765175546 Chr.2: 26576004-26576004 on Build GRCh38
rs749494116 Chr.2: 26575950-26575950 on Build GRCh38
rs372202665 Chr.2: 26576006-26576006 on Build GRCh38
rs750814998 Chr.2: 26576209-26576210 on Build GRCh38
rs200703707 Chr.2: 26576162-26576162 on Build GRCh38
rs753762077 Chr.2: 26575914-26575914 on Build GRCh38
rs367622220 Chr.2: 26575985-26575985 on Build GRCh38
rs1041225425 Chr.2: 26576082-26576082 on Build GRCh38
rs1043236567 Chr.2: 26576258-26576258 on Build GRCh38
rs531808422 Chr.2: 26575998-26575998 on Build GRCh38
rs764434980 Chr.2: 26576238-26576238 on Build GRCh38
rs61742175 Chr.2: 26576029-26576029 on Build GRCh38
rs771809134 Chr.2: 26576016-26576016 on Build GRCh38
rs1014323345 Chr.2: 26576172-26576172 on Build GRCh38
rs370346487 Chr.2: 26575902-26575902 on Build GRCh38
rs374799691 Chr.2: 26576121-26576121 on Build GRCh38
rs1033045187 Chr.2: 26576302-26576302 on Build GRCh38
rs201327242 Chr.2: 26576109-26576109 on Build GRCh38
rs777315280 Chr.2: 26575908-26575908 on Build GRCh38
rs753279641 Chr.2: 26575994-26575994 on Build GRCh38
rs1040838618 Chr.2: 26576254-26576254 on Build GRCh38
rs758184713 Chr.2: 26576015-26576015 on Build GRCh38
rs776735083 Chr.2: 26575851-26575851 on Build GRCh38
rs200725438 Chr.2: 26576218-26576218 on Build GRCh38
rs369849488 Chr.2: 26576187-26576187 on Build GRCh38
rs1020705394 Chr.2: 26576262-26576262 on Build GRCh38
rs768973565 Chr.2: 26575862-26575862 on Build GRCh38
rs181202795 Chr.2: 26575919-26575919 on Build GRCh38
rs1148953 Chr.2: 26576256-26576256 on Build GRCh38
rs910923973 Chr.2: 26576228-26576228 on Build GRCh38
rs943754953 Chr.2: 26576239-26576239 on Build GRCh38
rs201740466 Chr.2: 26576093-26576093 on Build GRCh38
rs866402113 Chr.2: 26576052-26576052 on Build GRCh38
rs767148167 Chr.2: 26575896-26575896 on Build GRCh38
rs567506070 Chr.2: 26575941-26575941 on Build GRCh38
rs769042927 Chr.2: 26576160-26576160 on Build GRCh38
rs756944732 Chr.2: 26575929-26575931 on Build GRCh38
rs759903840 Chr.2: 26575961-26575961 on Build GRCh38
rs780049642 Chr.2: 26576019-26576019 on Build GRCh38
rs543956755 Chr.2: 26576165-26576165 on Build GRCh38
rs572327407 Chr.2: 26576083-26576083 on Build GRCh38
rs961172721 Chr.2: 26576198-26576198 on Build GRCh38
rs767810976 Chr.2: 26575978-26575978 on Build GRCh38
rs778916888 Chr.2: 26576125-26576125 on Build GRCh38
rs368935437 Chr.2: 26576126-26576126 on Build GRCh38
rs749510808 Chr.2: 26576123-26576123 on Build GRCh38
rs553972095 Chr.2: 26576076-26576076 on Build GRCh38
rs895845623 Chr.2: 26576279-26576279 on Build GRCh38
rs13002673 Chr.2: 26576025-26576025 on Build GRCh38
rs760379580 Chr.2: 26575979-26575979 on Build GRCh38
rs954044078 Chr.2: 26576265-26576265 on Build GRCh38
rs201943227 Chr.2: 26575903-26575903 on Build GRCh38
rs747327595 Chr.2: 26576144-26576144 on Build GRCh38
rs1056188652 Chr.2: 26576277-26576277 on Build GRCh38
rs559552247 Chr.2: 26576213-26576213 on Build GRCh38
rs751338382 Chr.2: 26576106-26576106 on Build GRCh38
rs772471808 Chr.2: 26576073-26576073 on Build GRCh38

View Full Product Details