Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 26777924-26778413 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35F6
Entrez Gene ID
54978
Gene Name
solute carrier family 35 member F6
Gene Aliases
ANT2BP, C2orf18, TANGO9
UniGene
Hs.1594
Gene Chromosome Location
Chr.2: 26764274-26781231 on Build GRCh38
Transcripts
NM_017877.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291527 c.865G>A p.A289T Chr.2: 26778260-26778260 on Build GRCh38
4950009 c.917T>C p.L306P Chr.2: 26778312-26778312 on Build GRCh38
2912873 c.754G>A p.A252T Chr.2: 26778149-26778149 on Build GRCh38
3426341 c.834C>T p.F278F Chr.2: 26778229-26778229 on Build GRCh38
317072 c.702C>T p.P234P Chr.2: 26778097-26778097 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs892894795 Chr.2: 26778110-26778110 on Build GRCh38
rs929049668 Chr.2: 26777959-26777959 on Build GRCh38
rs201724027 Chr.2: 26778113-26778113 on Build GRCh38
rs764426025 Chr.2: 26778269-26778269 on Build GRCh38
rs765125230 Chr.2: 26778051-26778051 on Build GRCh38
rs1032864092 Chr.2: 26778239-26778239 on Build GRCh38
rs139136213 Chr.2: 26778166-26778166 on Build GRCh38
rs71441043 Chr.2: 26778194-26778194 on Build GRCh38
rs183157533 Chr.2: 26777958-26777958 on Build GRCh38
rs761269148 Chr.2: 26778121-26778121 on Build GRCh38
rs1011034634 Chr.2: 26778112-26778112 on Build GRCh38
rs772591280 Chr.2: 26778242-26778242 on Build GRCh38
rs373652138 Chr.2: 26778295-26778295 on Build GRCh38
rs758988437 Chr.2: 26778301-26778301 on Build GRCh38
rs766262923 Chr.2: 26778169-26778169 on Build GRCh38
rs202137789 Chr.2: 26778029-26778029 on Build GRCh38
rs373308346 Chr.2: 26778001-26778001 on Build GRCh38
rs902548312 Chr.2: 26778225-26778225 on Build GRCh38
rs777392115 Chr.2: 26778373-26778373 on Build GRCh38
rs150784252 Chr.2: 26778148-26778148 on Build GRCh38
rs941734916 Chr.2: 26778168-26778168 on Build GRCh38
rs565856734 Chr.2: 26778229-26778229 on Build GRCh38
rs751979502 Chr.2: 26778308-26778308 on Build GRCh38
rs757870921 Chr.2: 26778101-26778101 on Build GRCh38
rs759769490 Chr.2: 26778027-26778031 on Build GRCh38
rs773470057 Chr.2: 26777972-26777972 on Build GRCh38
rs778429096 Chr.2: 26778003-26778003 on Build GRCh38
rs866575859 Chr.2: 26778304-26778304 on Build GRCh38
rs760111445 Chr.2: 26778032-26778032 on Build GRCh38
rs761682671 Chr.2: 26778253-26778253 on Build GRCh38
rs536311747 Chr.2: 26778260-26778260 on Build GRCh38
rs769224906 Chr.2: 26778231-26778231 on Build GRCh38
rs775716080 Chr.2: 26778116-26778116 on Build GRCh38
rs200993409 Chr.2: 26778123-26778123 on Build GRCh38
rs966697898 Chr.2: 26778096-26778096 on Build GRCh38
rs770752397 Chr.2: 26778259-26778259 on Build GRCh38
rs530029652 Chr.2: 26778125-26778125 on Build GRCh38
rs530838038 Chr.2: 26778092-26778092 on Build GRCh38
rs756159127 Chr.2: 26777994-26777994 on Build GRCh38
rs981799189 Chr.2: 26778358-26778358 on Build GRCh38
rs200150725 Chr.2: 26778172-26778172 on Build GRCh38
rs768324836 Chr.2: 26778199-26778199 on Build GRCh38
rs569966708 Chr.2: 26778356-26778356 on Build GRCh38
rs774327484 Chr.2: 26778230-26778230 on Build GRCh38
rs746233457 Chr.2: 26778109-26778109 on Build GRCh38
rs1039231247 Chr.2: 26778174-26778174 on Build GRCh38
rs756285954 Chr.2: 26778097-26778097 on Build GRCh38
rs200205090 Chr.2: 26778009-26778009 on Build GRCh38
rs754048659 Chr.2: 26778271-26778271 on Build GRCh38
rs893911910 Chr.2: 26778249-26778249 on Build GRCh38
rs762659301 Chr.2: 26778142-26778142 on Build GRCh38
rs948678619 Chr.2: 26778047-26778048 on Build GRCh38
rs919357412 Chr.2: 26777980-26777980 on Build GRCh38
rs981341794 Chr.2: 26778122-26778122 on Build GRCh38
rs747211994 Chr.2: 26778052-26778052 on Build GRCh38
rs900772490 Chr.2: 26778061-26778061 on Build GRCh38
rs570029630 Chr.2: 26778151-26778151 on Build GRCh38
rs930472035 Chr.2: 26778006-26778006 on Build GRCh38
rs780828722 Chr.2: 26778326-26778326 on Build GRCh38
rs762124888 Chr.2: 26778290-26778290 on Build GRCh38
rs368930041 Chr.2: 26778108-26778108 on Build GRCh38
rs552403890 Chr.2: 26778105-26778105 on Build GRCh38
rs375888353 Chr.2: 26778016-26778016 on Build GRCh38
rs950455705 Chr.2: 26778389-26778389 on Build GRCh38
rs983634596 Chr.2: 26778156-26778156 on Build GRCh38
rs769424795 Chr.2: 26778366-26778366 on Build GRCh38
rs750451559 Chr.2: 26778296-26778296 on Build GRCh38
rs187626078 Chr.2: 26778185-26778185 on Build GRCh38
rs753995299 Chr.2: 26778177-26778177 on Build GRCh38
rs997737574 Chr.2: 26778084-26778084 on Build GRCh38
rs746205640 Chr.2: 26778019-26778019 on Build GRCh38
rs894356599 Chr.2: 26778036-26778036 on Build GRCh38
rs771647754 Chr.2: 26778023-26778023 on Build GRCh38
rs1035599437 Chr.2: 26778367-26778367 on Build GRCh38
rs146289512 Chr.2: 26778100-26778100 on Build GRCh38
rs559842319 Chr.2: 26778098-26778098 on Build GRCh38
rs747864074 Chr.2: 26778347-26778347 on Build GRCh38
rs780381802 Chr.2: 26778302-26778302 on Build GRCh38
rs373175788 Chr.2: 26778120-26778120 on Build GRCh38
rs114985478 Chr.2: 26778175-26778175 on Build GRCh38
rs746737214 Chr.2: 26778118-26778118 on Build GRCh38
rs761018960 Chr.2: 26778173-26778173 on Build GRCh38
rs755280937 Chr.2: 26778320-26778320 on Build GRCh38
rs773482867 Chr.2: 26778254-26778254 on Build GRCh38
rs878963965 Chr.2: 26778272-26778272 on Build GRCh38
rs1006378175 Chr.2: 26778044-26778044 on Build GRCh38
rs930732057 Chr.2: 26778247-26778247 on Build GRCh38
rs761784077 Chr.2: 26778039-26778039 on Build GRCh38
rs780521211 Chr.2: 26778186-26778186 on Build GRCh38
rs763664512 Chr.2: 26778034-26778034 on Build GRCh38
rs1036478013 Chr.2: 26777966-26777966 on Build GRCh38
rs748706622 Chr.2: 26778382-26778382 on Build GRCh38
rs759609073 Chr.2: 26778178-26778178 on Build GRCh38
rs747300363 Chr.2: 26778197-26778197 on Build GRCh38
rs928979208 Chr.2: 26778143-26778143 on Build GRCh38
rs757357044 Chr.2: 26778183-26778183 on Build GRCh38
rs767645176 Chr.2: 26778077-26778077 on Build GRCh38
rs758160255 Chr.2: 26778064-26778064 on Build GRCh38

View Full Product Details